Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93534741-93535268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
UniGene
Hs.134060
Gene Chromosome Location
Chr.1: 93448067-93569671 on Build GRCh38
Transcripts
NM_001024948.2 NM_001164473.2 NM_017737.4 XM_011541625.2 XM_017001533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344964 c.824T>G p.F275C Chr.1: 93534742-93534742 on Build GRCh38
4615288 c.907delA p.K303fs*13 Chr.1: 93534825-93534825 on Build GRCh38
5789528 c.977G>A p.G326E Chr.1: 93534895-93534895 on Build GRCh38
1285022 c.934delA p.T313fs*3 Chr.1: 93534852-93534852 on Build GRCh38
3977986 c.850G>T p.E284* Chr.1: 93534768-93534768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766654341 Chr.1: 93534932-93534934 on Build GRCh38
rs903246963 Chr.1: 93535002-93535002 on Build GRCh38
rs1021188544 Chr.1: 93535178-93535178 on Build GRCh38
rs377145504 Chr.1: 93534936-93534936 on Build GRCh38
rs968174504 Chr.1: 93534907-93534907 on Build GRCh38
rs986185526 Chr.1: 93534974-93534974 on Build GRCh38
rs1038999160 Chr.1: 93535230-93535230 on Build GRCh38
rs556573464 Chr.1: 93535058-93535058 on Build GRCh38
rs530561746 Chr.1: 93535233-93535233 on Build GRCh38
rs147331550 Chr.1: 93534972-93534972 on Build GRCh38
rs369621739 Chr.1: 93534833-93534833 on Build GRCh38
rs545705434 Chr.1: 93535110-93535110 on Build GRCh38
rs944375953 Chr.1: 93535025-93535025 on Build GRCh38
rs541371013 Chr.1: 93535038-93535038 on Build GRCh38
rs182265334 Chr.1: 93535159-93535159 on Build GRCh38
rs534587208 Chr.1: 93534812-93534812 on Build GRCh38
rs769511126 Chr.1: 93534928-93534928 on Build GRCh38
rs758718137 Chr.1: 93534881-93534881 on Build GRCh38
rs764162971 Chr.1: 93535141-93535141 on Build GRCh38
rs867493343 Chr.1: 93535048-93535048 on Build GRCh38
rs770373855 Chr.1: 93534783-93534783 on Build GRCh38
rs911594497 Chr.1: 93534998-93534998 on Build GRCh38
rs991734591 Chr.1: 93535099-93535099 on Build GRCh38
rs1021219358 Chr.1: 93534858-93534858 on Build GRCh38
rs201882774 Chr.1: 93534829-93534829 on Build GRCh38
rs979609413 Chr.1: 93534924-93534924 on Build GRCh38
rs762328668 Chr.1: 93535070-93535071 on Build GRCh38
rs973068270 Chr.1: 93535138-93535138 on Build GRCh38
rs367764338 Chr.1: 93534804-93534804 on Build GRCh38
rs372656799 Chr.1: 93534826-93534826 on Build GRCh38
rs766553618 Chr.1: 93534834-93534834 on Build GRCh38
rs778253994 Chr.1: 93534890-93534890 on Build GRCh38
rs769520098 Chr.1: 93534786-93534786 on Build GRCh38
rs376532621 Chr.1: 93535037-93535037 on Build GRCh38
rs1028522580 Chr.1: 93535202-93535202 on Build GRCh38
rs780725529 Chr.1: 93534922-93534922 on Build GRCh38
rs371983971 Chr.1: 93534873-93534873 on Build GRCh38
rs369143570 Chr.1: 93534957-93534957 on Build GRCh38
rs139322860 Chr.1: 93535055-93535055 on Build GRCh38
rs551530629 Chr.1: 93535008-93535008 on Build GRCh38
rs756176007 Chr.1: 93535140-93535140 on Build GRCh38
rs899866536 Chr.1: 93534941-93534941 on Build GRCh38
rs747505942 Chr.1: 93534899-93534899 on Build GRCh38
rs879260753 Chr.1: 93535139-93535139 on Build GRCh38
rs745559320 Chr.1: 93534925-93534925 on Build GRCh38
rs1001014346 Chr.1: 93535194-93535194 on Build GRCh38
rs748083464 Chr.1: 93535006-93535014 on Build GRCh38
rs1384298 Chr.1: 93535101-93535101 on Build GRCh38
rs565178376 Chr.1: 93534962-93534962 on Build GRCh38
rs778795633 Chr.1: 93535188-93535188 on Build GRCh38
rs373661130 Chr.1: 93534784-93534784 on Build GRCh38
rs548407914 Chr.1: 93535116-93535116 on Build GRCh38
rs755194767 Chr.1: 93534839-93534839 on Build GRCh38
rs756831876 Chr.1: 93534905-93534905 on Build GRCh38
rs778199083 Chr.1: 93534773-93534773 on Build GRCh38
rs565398423 Chr.1: 93535150-93535150 on Build GRCh38
rs746549667 Chr.1: 93534782-93534782 on Build GRCh38
rs779191301 Chr.1: 93534871-93534871 on Build GRCh38
rs753125613 Chr.1: 93534875-93534875 on Build GRCh38
rs1032735843 Chr.1: 93534952-93534952 on Build GRCh38
rs929036888 Chr.1: 93535143-93535147 on Build GRCh38
rs754072630 Chr.1: 93534835-93534835 on Build GRCh38
rs953239434 Chr.1: 93535242-93535242 on Build GRCh38
rs527889357 Chr.1: 93535154-93535154 on Build GRCh38

View Full Product Details