Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161228117-161228715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOMM40L
Entrez Gene ID
84134
Gene Name
translocase of outer mitochondrial membrane 40 like
Gene Aliases
TOMM40B
UniGene
Hs.321653
Gene Chromosome Location
Chr.1: 161225939-161230746 on Build GRCh38
Transcripts
NM_001286373.1 NM_001286374.1 NM_032174.5 XM_006711572.2 XM_011510057.2 XM_017002481.1 XM_017002482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1645075 c.577G>C p.A193P Chr.1: 161228278-161228278 on Build GRCh38
6989785 c.523C>T p.R175W Chr.1: 161228224-161228224 on Build GRCh38
5768218 c.568G>A p.E190K Chr.1: 161228269-161228269 on Build GRCh38
3705186 c.543G>A p.E181E Chr.1: 161228244-161228244 on Build GRCh38
3705187 c.544C>A p.L182I Chr.1: 161228245-161228245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761489741 Chr.1: 161228339-161228339 on Build GRCh38
rs755639135 Chr.1: 161228448-161228448 on Build GRCh38
rs777341360 Chr.1: 161228453-161228453 on Build GRCh38
rs751751970 Chr.1: 161228536-161228536 on Build GRCh38
rs745554102 Chr.1: 161228466-161228466 on Build GRCh38
rs142987556 Chr.1: 161228257-161228257 on Build GRCh38
rs921897699 Chr.1: 161228447-161228447 on Build GRCh38
rs776371436 Chr.1: 161228636-161228636 on Build GRCh38
rs202137061 Chr.1: 161228231-161228231 on Build GRCh38
rs777522046 Chr.1: 161228665-161228665 on Build GRCh38
rs769354698 Chr.1: 161228245-161228245 on Build GRCh38
rs564568531 Chr.1: 161228241-161228241 on Build GRCh38
rs146347518 Chr.1: 161228598-161228598 on Build GRCh38
rs766533700 Chr.1: 161228425-161228425 on Build GRCh38
rs137990738 Chr.1: 161228268-161228268 on Build GRCh38
rs904710194 Chr.1: 161228607-161228607 on Build GRCh38
rs764636511 Chr.1: 161228269-161228269 on Build GRCh38
rs933544110 Chr.1: 161228254-161228254 on Build GRCh38
rs946312463 Chr.1: 161228586-161228586 on Build GRCh38
rs761070402 Chr.1: 161228346-161228346 on Build GRCh38
rs549698861 Chr.1: 161228387-161228387 on Build GRCh38
rs756884199 Chr.1: 161228412-161228412 on Build GRCh38
rs753224917 Chr.1: 161228200-161228200 on Build GRCh38
rs1026090940 Chr.1: 161228599-161228599 on Build GRCh38
rs377605741 Chr.1: 161228685-161228685 on Build GRCh38
rs201442921 Chr.1: 161228470-161228470 on Build GRCh38
rs1033742937 Chr.1: 161228321-161228321 on Build GRCh38
rs767331843 Chr.1: 161228549-161228549 on Build GRCh38
rs1004780405 Chr.1: 161228695-161228695 on Build GRCh38
rs779919169 Chr.1: 161228576-161228576 on Build GRCh38
rs762973886 Chr.1: 161228525-161228525 on Build GRCh38
rs368132279 Chr.1: 161228574-161228574 on Build GRCh38
rs752366819 Chr.1: 161228551-161228551 on Build GRCh38
rs753403494 Chr.1: 161228317-161228317 on Build GRCh38
rs754730047 Chr.1: 161228437-161228437 on Build GRCh38
rs34197420 Chr.1: 161228458-161228459 on Build GRCh38
rs1043230793 Chr.1: 161228606-161228606 on Build GRCh38
rs762380913 Chr.1: 161228474-161228474 on Build GRCh38
rs945559799 Chr.1: 161228677-161228677 on Build GRCh38
rs199979621 Chr.1: 161228339-161228339 on Build GRCh38
rs751884586 Chr.1: 161228172-161228172 on Build GRCh38
rs372039948 Chr.1: 161228306-161228306 on Build GRCh38
rs201833813 Chr.1: 161228152-161228152 on Build GRCh38
rs1024851836 Chr.1: 161228343-161228343 on Build GRCh38
rs61731576 Chr.1: 161228307-161228307 on Build GRCh38
rs368013374 Chr.1: 161228181-161228181 on Build GRCh38
rs751824628 Chr.1: 161228224-161228224 on Build GRCh38
rs539138809 Chr.1: 161228548-161228548 on Build GRCh38
rs1054891603 Chr.1: 161228613-161228613 on Build GRCh38
rs753887040 Chr.1: 161228218-161228218 on Build GRCh38
rs757431559 Chr.1: 161228219-161228219 on Build GRCh38
rs775915514 Chr.1: 161228325-161228327 on Build GRCh38
rs936390503 Chr.1: 161228609-161228609 on Build GRCh38
rs767979892 Chr.1: 161228259-161228259 on Build GRCh38
rs1013203855 Chr.1: 161228620-161228620 on Build GRCh38
rs772789513 Chr.1: 161228337-161228337 on Build GRCh38
rs369169431 Chr.1: 161228694-161228694 on Build GRCh38
rs116664724 Chr.1: 161228623-161228623 on Build GRCh38
rs376405584 Chr.1: 161228314-161228314 on Build GRCh38
rs190278807 Chr.1: 161228404-161228404 on Build GRCh38
rs776125727 Chr.1: 161228238-161228238 on Build GRCh38
rs373013630 Chr.1: 161228441-161228441 on Build GRCh38
rs761370161 Chr.1: 161228156-161228156 on Build GRCh38
rs777995111 Chr.1: 161228211-161228211 on Build GRCh38
rs750681272 Chr.1: 161228413-161228413 on Build GRCh38
rs768226600 Chr.1: 161228233-161228233 on Build GRCh38
rs915494063 Chr.1: 161228626-161228626 on Build GRCh38
rs148537960 Chr.1: 161228274-161228274 on Build GRCh38
rs769316999 Chr.1: 161228333-161228333 on Build GRCh38
rs771819011 Chr.1: 161228225-161228225 on Build GRCh38
rs753883387 Chr.1: 161228276-161228276 on Build GRCh38
rs765601025 Chr.1: 161228496-161228496 on Build GRCh38
rs142881710 Chr.1: 161228462-161228462 on Build GRCh38
rs780012355 Chr.1: 161228229-161228229 on Build GRCh38
rs748837835 Chr.1: 161228666-161228666 on Build GRCh38
rs200820654 Chr.1: 161228202-161228202 on Build GRCh38
rs181535066 Chr.1: 161228577-161228577 on Build GRCh38
rs201092540 Chr.1: 161228446-161228446 on Build GRCh38
rs372859200 Chr.1: 161228331-161228331 on Build GRCh38
rs755542977 Chr.1: 161228235-161228235 on Build GRCh38
rs756089823 Chr.1: 161228664-161228664 on Build GRCh38
rs756319764 Chr.1: 161228208-161228208 on Build GRCh38
rs186166605 Chr.1: 161228318-161228318 on Build GRCh38
rs142138143 Chr.1: 161228552-161228552 on Build GRCh38
rs534049072 Chr.1: 161228258-161228258 on Build GRCh38
rs758250541 Chr.1: 161228309-161228309 on Build GRCh38
rs757317383 Chr.1: 161228299-161228299 on Build GRCh38
rs764880653 Chr.1: 161228162-161228162 on Build GRCh38
rs569307435 Chr.1: 161228558-161228558 on Build GRCh38
rs781486410 Chr.1: 161228284-161228284 on Build GRCh38
rs764844453 Chr.1: 161228339-161228340 on Build GRCh38
rs886345569 Chr.1: 161228652-161228652 on Build GRCh38
rs751381557 Chr.1: 161228428-161228428 on Build GRCh38
rs761844271 Chr.1: 161228408-161228408 on Build GRCh38
rs777058436 Chr.1: 161228473-161228473 on Build GRCh38
rs200010139 Chr.1: 161228467-161228467 on Build GRCh38
rs771826742 Chr.1: 161228556-161228556 on Build GRCh38
rs774104502 Chr.1: 161228463-161228463 on Build GRCh38
rs369177277 Chr.1: 161228424-161228424 on Build GRCh38
rs776008781 Chr.1: 161228340-161228340 on Build GRCh38
rs781033930 Chr.1: 161228443-161228443 on Build GRCh38
rs773358077 Chr.1: 161228519-161228519 on Build GRCh38
rs766619867 Chr.1: 161228535-161228535 on Build GRCh38
rs759898024 Chr.1: 161228178-161228178 on Build GRCh38
rs747415327 Chr.1: 161228465-161228465 on Build GRCh38
rs374950980 Chr.1: 161228179-161228179 on Build GRCh38
rs779145031 Chr.1: 161228222-161228222 on Build GRCh38
rs950676338 Chr.1: 161228344-161228344 on Build GRCh38
rs113211519 Chr.1: 161228228-161228228 on Build GRCh38
rs746741835 Chr.1: 161228315-161228315 on Build GRCh38
rs983241571 Chr.1: 161228369-161228369 on Build GRCh38
rs753322527 Chr.1: 161228263-161228263 on Build GRCh38
rs746738371 Chr.1: 161228619-161228619 on Build GRCh38

View Full Product Details