Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62132450-62133044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRG
Entrez Gene ID
5793
Gene Name
protein tyrosine phosphatase, receptor type G
Gene Aliases
HPTPG, PTPG, R-PTP-GAMMA, RPTPG
UniGene
Hs.595541
Gene Chromosome Location
Chr.3: 61561569-62297609 on Build GRCh38
Transcripts
NM_002841.3 XM_005265353.3 XM_017006961.1 XM_017006962.1 XM_017006963.1 XM_017006964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904774 c.628G>A p.D210N Chr.3: 62132614-62132614 on Build GRCh38
222816 c.659G>A p.G220E Chr.3: 62132645-62132645 on Build GRCh38
3945650 c.654C>G p.I218M Chr.3: 62132640-62132640 on Build GRCh38
6165285 c.677A>G p.H226R Chr.3: 62132663-62132663 on Build GRCh38
6282196 c.658G>A p.G220R Chr.3: 62132644-62132644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777665676 Chr.3: 62132618-62132618 on Build GRCh38
rs768326759 Chr.3: 62132642-62132642 on Build GRCh38
rs940827043 Chr.3: 62132782-62132782 on Build GRCh38
rs766431772 Chr.3: 62132599-62132599 on Build GRCh38
rs746834721 Chr.3: 62132619-62132619 on Build GRCh38
rs145318908 Chr.3: 62132640-62132640 on Build GRCh38
rs987718245 Chr.3: 62132990-62132990 on Build GRCh38
rs757061905 Chr.3: 62132702-62132702 on Build GRCh38
rs761579975 Chr.3: 62132787-62132787 on Build GRCh38
rs144784343 Chr.3: 62132652-62132652 on Build GRCh38
rs1040589029 Chr.3: 62132784-62132784 on Build GRCh38
rs1056091582 Chr.3: 62132789-62132789 on Build GRCh38
rs372255585 Chr.3: 62132506-62132506 on Build GRCh38
rs61757808 Chr.3: 62132609-62132609 on Build GRCh38
rs895609830 Chr.3: 62132625-62132625 on Build GRCh38
rs779981819 Chr.3: 62132612-62132612 on Build GRCh38
rs768236996 Chr.3: 62132720-62132720 on Build GRCh38
rs754085336 Chr.3: 62132606-62132606 on Build GRCh38
rs552552724 Chr.3: 62132758-62132758 on Build GRCh38
rs765562785 Chr.3: 62132696-62132696 on Build GRCh38
rs114254859 Chr.3: 62132705-62132705 on Build GRCh38
rs904561632 Chr.3: 62132520-62132520 on Build GRCh38
rs926944080 Chr.3: 62132734-62132734 on Build GRCh38
rs994566271 Chr.3: 62132501-62132501 on Build GRCh38
rs905542156 Chr.3: 62132472-62132472 on Build GRCh38
rs147897609 Chr.3: 62132988-62132988 on Build GRCh38
rs780917324 Chr.3: 62132704-62132704 on Build GRCh38
rs768070313 Chr.3: 62132582-62132582 on Build GRCh38
rs753995594 Chr.3: 62132658-62132658 on Build GRCh38
rs1005094198 Chr.3: 62132577-62132577 on Build GRCh38
rs1031030091 Chr.3: 62132825-62132825 on Build GRCh38
rs1001839249 Chr.3: 62132549-62132549 on Build GRCh38
rs886567295 Chr.3: 62132558-62132558 on Build GRCh38
rs762248211 Chr.3: 62132644-62132644 on Build GRCh38
rs913286014 Chr.3: 62132737-62132737 on Build GRCh38
rs375539164 Chr.3: 62132701-62132701 on Build GRCh38
rs1004593919 Chr.3: 62132937-62132937 on Build GRCh38
rs968467831 Chr.3: 62132641-62132641 on Build GRCh38
rs548286261 Chr.3: 62132712-62132712 on Build GRCh38
rs149393059 Chr.3: 62132643-62132643 on Build GRCh38
rs1005288986 Chr.3: 62132918-62132918 on Build GRCh38
rs533835628 Chr.3: 62132610-62132610 on Build GRCh38
rs200790873 Chr.3: 62132629-62132629 on Build GRCh38
rs757076816 Chr.3: 62132621-62132621 on Build GRCh38
rs1508392 Chr.3: 62132910-62132910 on Build GRCh38
rs180851782 Chr.3: 62132646-62132646 on Build GRCh38
rs371016927 Chr.3: 62132637-62132637 on Build GRCh38
rs150201805 Chr.3: 62132716-62132716 on Build GRCh38
rs922221885 Chr.3: 62132677-62132677 on Build GRCh38
rs971644568 Chr.3: 62132940-62132940 on Build GRCh38
rs751942035 Chr.3: 62132570-62132570 on Build GRCh38
rs996416296 Chr.3: 62132794-62132794 on Build GRCh38
rs756201759 Chr.3: 62132595-62132595 on Build GRCh38
rs996544138 Chr.3: 62132944-62132944 on Build GRCh38
rs575608589 Chr.3: 62132498-62132498 on Build GRCh38
rs750410527 Chr.3: 62132590-62132590 on Build GRCh38
rs956808463 Chr.3: 62132864-62132864 on Build GRCh38
rs13082242 Chr.3: 62132816-62132816 on Build GRCh38
rs907608229 Chr.3: 62133006-62133006 on Build GRCh38
rs766549529 Chr.3: 62132656-62132656 on Build GRCh38
rs186087981 Chr.3: 62132738-62132738 on Build GRCh38
rs769343658 Chr.3: 62132635-62132635 on Build GRCh38
rs1019199980 Chr.3: 62132604-62132604 on Build GRCh38
rs946013605 Chr.3: 62132510-62132510 on Build GRCh38
rs569354996 Chr.3: 62132984-62132984 on Build GRCh38
rs762266838 Chr.3: 62132576-62132576 on Build GRCh38
rs886223781 Chr.3: 62132770-62132770 on Build GRCh38
rs1016715649 Chr.3: 62132931-62132931 on Build GRCh38
rs74649851 Chr.3: 62132956-62132956 on Build GRCh38
rs949972628 Chr.3: 62132707-62132707 on Build GRCh38
rs925933730 Chr.3: 62132785-62132785 on Build GRCh38
rs754342635 Chr.3: 62132926-62132926 on Build GRCh38
rs1029668557 Chr.3: 62132800-62132800 on Build GRCh38
rs761014301 Chr.3: 62132718-62132718 on Build GRCh38
rs748800005 Chr.3: 62132719-62132719 on Build GRCh38
rs755214551 Chr.3: 62132695-62132695 on Build GRCh38
rs903816278 Chr.3: 62132805-62132805 on Build GRCh38
rs73096543 Chr.3: 62132994-62132994 on Build GRCh38
rs927241167 Chr.3: 62132986-62132986 on Build GRCh38
rs151015230 Chr.3: 62132518-62132518 on Build GRCh38
rs140916981 Chr.3: 62132630-62132630 on Build GRCh38
rs188991589 Chr.3: 62132645-62132645 on Build GRCh38

View Full Product Details