Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 79087812-79088348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REG1B
Entrez Gene ID
5968
Gene Name
regenerating family member 1 beta
Gene Aliases
PSPS2, REGH, REGI-BETA, REGL
UniGene
Hs.4158
Gene Chromosome Location
Chr.2: 79085023-79088024 on Build GRCh38
Transcripts
NM_006507.3

SNP Detail

SNP ID Chromosome Location
rs148994971 Chr.2: 79088173-79088173 on Build GRCh38
rs762821161 Chr.2: 79088211-79088211 on Build GRCh38
rs893111442 Chr.2: 79087888-79087888 on Build GRCh38
rs769068586 Chr.2: 79088154-79088154 on Build GRCh38
rs1053621759 Chr.2: 79088008-79088008 on Build GRCh38
rs899430560 Chr.2: 79088120-79088120 on Build GRCh38
rs554638420 Chr.2: 79088040-79088040 on Build GRCh38
rs893143786 Chr.2: 79088178-79088178 on Build GRCh38
rs573837463 Chr.2: 79088050-79088050 on Build GRCh38
rs937922762 Chr.2: 79088170-79088170 on Build GRCh38
rs758010726 Chr.2: 79087920-79087924 on Build GRCh38
rs563730478 Chr.2: 79087872-79087872 on Build GRCh38
rs571860917 Chr.2: 79087894-79087894 on Build GRCh38
rs776320994 Chr.2: 79088094-79088094 on Build GRCh38
rs957940562 Chr.2: 79088226-79088226 on Build GRCh38
rs994976957 Chr.2: 79087899-79087899 on Build GRCh38
rs553552425 Chr.2: 79087980-79087980 on Build GRCh38
rs1015151619 Chr.2: 79087876-79087876 on Build GRCh38
rs947939196 Chr.2: 79088238-79088238 on Build GRCh38
rs544900163 Chr.2: 79088196-79088196 on Build GRCh38
rs1027158198 Chr.2: 79088245-79088245 on Build GRCh38
rs746627305 Chr.2: 79087924-79087924 on Build GRCh38
rs768304448 Chr.2: 79088029-79088029 on Build GRCh38
rs534552727 Chr.2: 79087967-79087967 on Build GRCh38
rs949286327 Chr.2: 79088130-79088130 on Build GRCh38
rs905108268 Chr.2: 79087849-79087849 on Build GRCh38
rs981852967 Chr.2: 79088289-79088289 on Build GRCh38
rs937787126 Chr.2: 79088303-79088303 on Build GRCh38
rs562503572 Chr.2: 79088071-79088071 on Build GRCh38
rs967580900 Chr.2: 79087922-79087922 on Build GRCh38
rs947790783 Chr.2: 79088012-79088012 on Build GRCh38
rs572220386 Chr.2: 79087911-79087911 on Build GRCh38
rs982559277 Chr.2: 79088097-79088097 on Build GRCh38
rs1019906596 Chr.2: 79087932-79087932 on Build GRCh38
rs532337738 Chr.2: 79088307-79088307 on Build GRCh38
rs930888072 Chr.2: 79088323-79088323 on Build GRCh38
rs1046177040 Chr.2: 79088051-79088051 on Build GRCh38
rs2073443 Chr.2: 79088159-79088159 on Build GRCh38
rs982852454 Chr.2: 79088262-79088262 on Build GRCh38
rs1041842701 Chr.2: 79088182-79088182 on Build GRCh38
rs570017325 Chr.2: 79088295-79088295 on Build GRCh38
rs975077712 Chr.2: 79087987-79087987 on Build GRCh38
rs1035987655 Chr.2: 79088283-79088283 on Build GRCh38
rs927746872 Chr.2: 79088270-79088270 on Build GRCh38
rs1011678604 Chr.2: 79087897-79087897 on Build GRCh38
rs575983990 Chr.2: 79088101-79088101 on Build GRCh38
rs977429046 Chr.2: 79088219-79088219 on Build GRCh38
rs964245516 Chr.2: 79087951-79087951 on Build GRCh38
rs952445321 Chr.2: 79088250-79088250 on Build GRCh38
rs114386614 Chr.2: 79088280-79088280 on Build GRCh38
rs911872115 Chr.2: 79087950-79087950 on Build GRCh38
rs1055788818 Chr.2: 79087887-79087887 on Build GRCh38
rs143823866 Chr.2: 79088180-79088180 on Build GRCh38
rs542599472 Chr.2: 79088070-79088070 on Build GRCh38
rs187216467 Chr.2: 79087837-79087837 on Build GRCh38
rs970868222 Chr.2: 79088284-79088284 on Build GRCh38
rs116737973 Chr.2: 79088231-79088231 on Build GRCh38
rs926459812 Chr.2: 79088294-79088294 on Build GRCh38
rs2073442 Chr.2: 79088225-79088225 on Build GRCh38
rs761842040 Chr.2: 79088202-79088202 on Build GRCh38
rs533707974 Chr.2: 79087963-79087963 on Build GRCh38
rs750732819 Chr.2: 79088230-79088230 on Build GRCh38
rs916459276 Chr.2: 79088011-79088011 on Build GRCh38
rs952359392 Chr.2: 79088087-79088087 on Build GRCh38
rs1019051475 Chr.2: 79088190-79088190 on Build GRCh38
rs1036543017 Chr.2: 79088107-79088107 on Build GRCh38
rs10193627 Chr.2: 79088309-79088309 on Build GRCh38
rs577247923 Chr.2: 79087858-79087858 on Build GRCh38
rs1053712898 Chr.2: 79088306-79088306 on Build GRCh38

View Full Product Details