Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 70881896-70882485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC49
Entrez Gene ID
54839
Gene Name
leucine rich repeat containing 49
Gene Aliases
PGs4
UniGene
Hs.12692
Gene Chromosome Location
Chr.15: 70852671-71050097 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Gene Symbol
THAP10
Entrez Gene ID
56906
Gene Name
THAP domain containing 10
Gene Aliases
-
UniGene
Hs.591123
Gene Chromosome Location
Chr.15: 70881342-70892433 on Build GRCh38
Transcripts
NM_001284357.1 XM_017022357.1 XM_017022359.1 NM_020147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6801024 c.748G>A p.V250I Chr.15: 70882480-70882480 on Build GRCh38
5383757 c.759A>G p.K253K Chr.15: 70882469-70882469 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023653349 Chr.15: 70882234-70882235 on Build GRCh38
rs868370589 Chr.15: 70882247-70882247 on Build GRCh38
rs1031166361 Chr.15: 70882207-70882207 on Build GRCh38
rs531067507 Chr.15: 70882344-70882344 on Build GRCh38
rs1000137025 Chr.15: 70882124-70882124 on Build GRCh38
rs560018074 Chr.15: 70882091-70882091 on Build GRCh38
rs763405112 Chr.15: 70882200-70882200 on Build GRCh38
rs1037390702 Chr.15: 70882399-70882399 on Build GRCh38
rs969352608 Chr.15: 70882235-70882235 on Build GRCh38
rs775801004 Chr.15: 70882442-70882442 on Build GRCh38
rs955425092 Chr.15: 70882177-70882177 on Build GRCh38
rs1010402327 Chr.15: 70882156-70882156 on Build GRCh38
rs929794394 Chr.15: 70881948-70881948 on Build GRCh38
rs563940504 Chr.15: 70882436-70882436 on Build GRCh38
rs892087884 Chr.15: 70882143-70882143 on Build GRCh38
rs764288615 Chr.15: 70882446-70882446 on Build GRCh38
rs1030564814 Chr.15: 70882108-70882108 on Build GRCh38
rs918490157 Chr.15: 70882383-70882383 on Build GRCh38
rs996403892 Chr.15: 70882083-70882083 on Build GRCh38
rs759989555 Chr.15: 70882431-70882431 on Build GRCh38
rs1044690026 Chr.15: 70882028-70882028 on Build GRCh38
rs551828351 Chr.15: 70882264-70882264 on Build GRCh38
rs372078971 Chr.15: 70882444-70882444 on Build GRCh38
rs1033684166 Chr.15: 70882134-70882134 on Build GRCh38
rs903532301 Chr.15: 70882059-70882059 on Build GRCh38
rs776682503 Chr.15: 70882448-70882449 on Build GRCh38
rs145846479 Chr.15: 70882460-70882460 on Build GRCh38
rs774956626 Chr.15: 70882429-70882429 on Build GRCh38
rs747705301 Chr.15: 70882423-70882423 on Build GRCh38
rs900243836 Chr.15: 70882443-70882443 on Build GRCh38
rs1056259391 Chr.15: 70881985-70881985 on Build GRCh38
rs896224914 Chr.15: 70882015-70882015 on Build GRCh38
rs111981438 Chr.15: 70882241-70882241 on Build GRCh38
rs890701096 Chr.15: 70882115-70882115 on Build GRCh38
rs550573394 Chr.15: 70882427-70882427 on Build GRCh38
rs773603470 Chr.15: 70882076-70882076 on Build GRCh38
rs1029865679 Chr.15: 70882164-70882164 on Build GRCh38
rs540462822 Chr.15: 70882240-70882240 on Build GRCh38
rs928563426 Chr.15: 70882269-70882269 on Build GRCh38
rs983695018 Chr.15: 70882298-70882298 on Build GRCh38
rs528670818 Chr.15: 70882214-70882214 on Build GRCh38
rs941789415 Chr.15: 70882380-70882380 on Build GRCh38
rs569059408 Chr.15: 70881966-70881966 on Build GRCh38
rs1018240653 Chr.15: 70882206-70882206 on Build GRCh38
rs767922286 Chr.15: 70882434-70882434 on Build GRCh38
rs776059168 Chr.15: 70882406-70882406 on Build GRCh38
rs185958880 Chr.15: 70882309-70882309 on Build GRCh38
rs559810721 Chr.15: 70882381-70882381 on Build GRCh38
rs947828174 Chr.15: 70882016-70882016 on Build GRCh38
rs540295060 Chr.15: 70881978-70881978 on Build GRCh38
rs1000419796 Chr.15: 70882123-70882123 on Build GRCh38
rs938601929 Chr.15: 70882297-70882297 on Build GRCh38
rs958365639 Chr.15: 70882208-70882208 on Build GRCh38

View Full Product Details