Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10433510-10434100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP2
Entrez Gene ID
80063
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
MCAF2
UniGene
Hs.513343
Gene Chromosome Location
Chr.16: 10386055-10483640 on Build GRCh38
Transcripts
NM_001256160.1 NM_024997.3 NR_045815.1 NR_045816.1 XM_006720953.3 XM_006720954.3 XM_017023705.1 XM_017023706.1 XM_017023707.1 XM_017023708.1 XM_017023710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641984 c.887G>A p.R296H Chr.16: 10433576-10433576 on Build GRCh38
4939014 c.873G>A p.E291E Chr.16: 10433562-10433562 on Build GRCh38
3957099 c.910A>G p.N304D Chr.16: 10433599-10433599 on Build GRCh38
6446064 c.891G>A p.M297I Chr.16: 10433580-10433580 on Build GRCh38
6420314 c.879T>A p.N293K Chr.16: 10433568-10433568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565265744 Chr.16: 10433794-10433794 on Build GRCh38
rs764674729 Chr.16: 10433595-10433595 on Build GRCh38
rs974663323 Chr.16: 10433879-10433879 on Build GRCh38
rs547287704 Chr.16: 10433972-10433972 on Build GRCh38
rs986710248 Chr.16: 10433834-10433834 on Build GRCh38
rs902782007 Chr.16: 10433746-10433746 on Build GRCh38
rs200252356 Chr.16: 10433559-10433559 on Build GRCh38
rs12930874 Chr.16: 10434017-10434017 on Build GRCh38
rs1010709029 Chr.16: 10433795-10433795 on Build GRCh38
rs12918266 Chr.16: 10434011-10434011 on Build GRCh38
rs950255152 Chr.16: 10433950-10433950 on Build GRCh38
rs963093427 Chr.16: 10433791-10433791 on Build GRCh38
rs201540502 Chr.16: 10433695-10433695 on Build GRCh38
rs951277814 Chr.16: 10433816-10433816 on Build GRCh38
rs528806582 Chr.16: 10433897-10433898 on Build GRCh38
rs369030187 Chr.16: 10433688-10433688 on Build GRCh38
rs1025613939 Chr.16: 10433805-10433805 on Build GRCh38
rs1039424608 Chr.16: 10433711-10433711 on Build GRCh38
rs59023972 Chr.16: 10433975-10433975 on Build GRCh38
rs199775004 Chr.16: 10433902-10433903 on Build GRCh38
rs868173880 Chr.16: 10434038-10434038 on Build GRCh38
rs1045958786 Chr.16: 10433736-10433736 on Build GRCh38
rs942450432 Chr.16: 10433702-10433702 on Build GRCh38
rs75255978 Chr.16: 10433739-10433739 on Build GRCh38
rs774126998 Chr.16: 10433701-10433706 on Build GRCh38
rs927468189 Chr.16: 10433954-10433954 on Build GRCh38
rs930352319 Chr.16: 10433686-10433686 on Build GRCh38
rs753582134 Chr.16: 10433949-10433949 on Build GRCh38
rs374397904 Chr.16: 10433564-10433564 on Build GRCh38
rs896644175 Chr.16: 10433769-10433769 on Build GRCh38
rs768459665 Chr.16: 10433543-10433543 on Build GRCh38
rs554619131 Chr.16: 10434010-10434010 on Build GRCh38
rs183712422 Chr.16: 10434065-10434065 on Build GRCh38
rs538897121 Chr.16: 10433977-10433977 on Build GRCh38
rs539670131 Chr.16: 10433728-10433729 on Build GRCh38
rs75181877 Chr.16: 10433759-10433759 on Build GRCh38
rs559085869 Chr.16: 10434001-10434001 on Build GRCh38
rs117042339 Chr.16: 10433953-10433953 on Build GRCh38
rs779055275 Chr.16: 10433611-10433611 on Build GRCh38
rs986668073 Chr.16: 10433670-10433670 on Build GRCh38
rs774125134 Chr.16: 10433675-10433675 on Build GRCh38
rs186293150 Chr.16: 10433631-10433631 on Build GRCh38
rs201156120 Chr.16: 10433597-10433597 on Build GRCh38
rs144646219 Chr.16: 10433578-10433578 on Build GRCh38
rs141089693 Chr.16: 10433547-10433547 on Build GRCh38
rs975282115 Chr.16: 10434076-10434076 on Build GRCh38
rs752493796 Chr.16: 10433786-10433786 on Build GRCh38
rs896669592 Chr.16: 10433989-10433989 on Build GRCh38
rs774257467 Chr.16: 10433668-10433668 on Build GRCh38
rs778512908 Chr.16: 10433968-10433968 on Build GRCh38
rs757388716 Chr.16: 10433609-10433609 on Build GRCh38
rs865955838 Chr.16: 10433585-10433585 on Build GRCh38
rs760405876 Chr.16: 10433849-10433849 on Build GRCh38
rs987783161 Chr.16: 10433598-10433598 on Build GRCh38
rs151108987 Chr.16: 10433545-10433545 on Build GRCh38
rs753791558 Chr.16: 10433608-10433608 on Build GRCh38
rs918532933 Chr.16: 10433616-10433616 on Build GRCh38
rs556546358 Chr.16: 10434029-10434029 on Build GRCh38
rs765883753 Chr.16: 10433560-10433560 on Build GRCh38
rs763812784 Chr.16: 10433939-10433939 on Build GRCh38
rs1012772960 Chr.16: 10433787-10433787 on Build GRCh38
rs190799715 Chr.16: 10433633-10433633 on Build GRCh38
rs1020397357 Chr.16: 10433821-10433821 on Build GRCh38
rs7205853 Chr.16: 10433756-10433756 on Build GRCh38
rs181592635 Chr.16: 10433866-10433866 on Build GRCh38
rs950267143 Chr.16: 10433733-10433733 on Build GRCh38
rs867732781 Chr.16: 10433588-10433588 on Build GRCh38
rs746766010 Chr.16: 10433541-10433541 on Build GRCh38
rs920226877 Chr.16: 10433880-10433880 on Build GRCh38
rs772561259 Chr.16: 10433689-10433689 on Build GRCh38
rs935619724 Chr.16: 10433984-10433984 on Build GRCh38
rs112669070 Chr.16: 10433640-10433640 on Build GRCh38
rs367653001 Chr.16: 10433652-10433652 on Build GRCh38
rs117331770 Chr.16: 10433955-10433955 on Build GRCh38
rs1033048803 Chr.16: 10433536-10433536 on Build GRCh38
rs983586781 Chr.16: 10433916-10433916 on Build GRCh38
rs571301918 Chr.16: 10434005-10434005 on Build GRCh38
rs1012434355 Chr.16: 10433996-10433996 on Build GRCh38
rs1056628195 Chr.16: 10433985-10433985 on Build GRCh38
rs550740570 Chr.16: 10433904-10433904 on Build GRCh38
rs754734275 Chr.16: 10433626-10433626 on Build GRCh38
rs951350017 Chr.16: 10434048-10434048 on Build GRCh38
rs768043303 Chr.16: 10433575-10433575 on Build GRCh38
rs372313756 Chr.16: 10433638-10433638 on Build GRCh38
rs1023700533 Chr.16: 10433790-10433790 on Build GRCh38
rs545497476 Chr.16: 10433770-10433770 on Build GRCh38
rs373265497 Chr.16: 10433538-10433538 on Build GRCh38
rs770497574 Chr.16: 10433667-10433667 on Build GRCh38
rs774180158 Chr.16: 10433561-10433561 on Build GRCh38
rs879107236 Chr.16: 10433777-10433777 on Build GRCh38
rs748710667 Chr.16: 10433660-10433660 on Build GRCh38
rs886592213 Chr.16: 10433710-10433710 on Build GRCh38
rs141109455 Chr.16: 10434065-10434066 on Build GRCh38
rs1008172966 Chr.16: 10434051-10434051 on Build GRCh38
rs770659563 Chr.16: 10433667-10433667 on Build GRCh38
rs375592507 Chr.16: 10433614-10433614 on Build GRCh38
rs1018867031 Chr.16: 10433860-10433860 on Build GRCh38
rs372156472 Chr.16: 10433579-10433579 on Build GRCh38
rs769345893 Chr.16: 10433554-10433554 on Build GRCh38
rs191078712 Chr.16: 10434058-10434058 on Build GRCh38
rs12930870 Chr.16: 10434002-10434002 on Build GRCh38
rs966928803 Chr.16: 10433826-10433826 on Build GRCh38
rs759459067 Chr.16: 10433743-10433743 on Build GRCh38
rs1025687368 Chr.16: 10434031-10434031 on Build GRCh38

View Full Product Details