Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67229254-67229810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2 NM_001318217.1 NM_014187.3 NR_134524.1

Gene Symbol
TMEM208
Entrez Gene ID
29100
Gene Name
transmembrane protein 208
Gene Aliases
HSPC171
UniGene
Hs.433203
Gene Chromosome Location
Chr.16: 67227113-67229279 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2 NM_001318217.1 NM_014187.3 NR_134524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3387535 c.3399C>A p.G1133G Chr.16: 67229806-67229806 on Build GRCh38
7106031 c.3445G>A p.D1149N Chr.16: 67229686-67229686 on Build GRCh38
7250491 c.3463G>A p.G1155R Chr.16: 67229668-67229668 on Build GRCh38
6663960 c.3444T>C p.D1148D Chr.16: 67229687-67229687 on Build GRCh38
3818249 c.3426G>C p.L1142F Chr.16: 67229705-67229705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750859651 Chr.16: 67229752-67229752 on Build GRCh38
rs943089666 Chr.16: 67229479-67229479 on Build GRCh38
rs796682373 Chr.16: 67229295-67229295 on Build GRCh38
rs895789130 Chr.16: 67229541-67229541 on Build GRCh38
rs757800624 Chr.16: 67229650-67229650 on Build GRCh38
rs764857793 Chr.16: 67229624-67229624 on Build GRCh38
rs577717314 Chr.16: 67229726-67229726 on Build GRCh38
rs768077398 Chr.16: 67229746-67229746 on Build GRCh38
rs373847429 Chr.16: 67229594-67229594 on Build GRCh38
rs538035714 Chr.16: 67229569-67229569 on Build GRCh38
rs755457226 Chr.16: 67229573-67229576 on Build GRCh38
rs748689233 Chr.16: 67229731-67229731 on Build GRCh38
rs752089928 Chr.16: 67229640-67229640 on Build GRCh38
rs763232262 Chr.16: 67229630-67229630 on Build GRCh38
rs750557765 Chr.16: 67229662-67229662 on Build GRCh38
rs578181551 Chr.16: 67229516-67229516 on Build GRCh38
rs749473379 Chr.16: 67229670-67229670 on Build GRCh38
rs201725796 Chr.16: 67229610-67229610 on Build GRCh38
rs922908489 Chr.16: 67229786-67229786 on Build GRCh38
rs754043657 Chr.16: 67229779-67229779 on Build GRCh38
rs769282334 Chr.16: 67229707-67229707 on Build GRCh38
rs970605667 Chr.16: 67229369-67229369 on Build GRCh38
rs568989262 Chr.16: 67229317-67229317 on Build GRCh38
rs745916248 Chr.16: 67229591-67229591 on Build GRCh38
rs559733281 Chr.16: 67229478-67229478 on Build GRCh38
rs372303748 Chr.16: 67229595-67229595 on Build GRCh38
rs1053116375 Chr.16: 67229519-67229519 on Build GRCh38
rs12447426 Chr.16: 67229422-67229422 on Build GRCh38
rs191057325 Chr.16: 67229730-67229730 on Build GRCh38
rs982454664 Chr.16: 67229424-67229424 on Build GRCh38
rs777341518 Chr.16: 67229586-67229588 on Build GRCh38
rs1014365261 Chr.16: 67229546-67229546 on Build GRCh38
rs187031352 Chr.16: 67229473-67229473 on Build GRCh38
rs1023151388 Chr.16: 67229365-67229365 on Build GRCh38
rs771041970 Chr.16: 67229689-67229689 on Build GRCh38
rs920877703 Chr.16: 67229416-67229416 on Build GRCh38
rs929011044 Chr.16: 67229488-67229488 on Build GRCh38
rs1023077764 Chr.16: 67229547-67229547 on Build GRCh38
rs199940256 Chr.16: 67229759-67229759 on Build GRCh38
rs763703372 Chr.16: 67229712-67229712 on Build GRCh38
rs1006117998 Chr.16: 67229407-67229407 on Build GRCh38
rs140881591 Chr.16: 67229663-67229663 on Build GRCh38
rs950990975 Chr.16: 67229420-67229420 on Build GRCh38
rs144815381 Chr.16: 67229671-67229671 on Build GRCh38
rs993332820 Chr.16: 67229353-67229353 on Build GRCh38
rs756157624 Chr.16: 67229754-67229754 on Build GRCh38
rs958184211 Chr.16: 67229472-67229475 on Build GRCh38
rs1047612437 Chr.16: 67229500-67229500 on Build GRCh38
rs932500610 Chr.16: 67229487-67229487 on Build GRCh38
rs150522467 Chr.16: 67229572-67229572 on Build GRCh38
rs1040404151 Chr.16: 67229485-67229486 on Build GRCh38
rs781646540 Chr.16: 67229658-67229658 on Build GRCh38
rs748689976 Chr.16: 67229668-67229669 on Build GRCh38
rs200812024 Chr.16: 67229708-67229708 on Build GRCh38
rs1033995073 Chr.16: 67229412-67229412 on Build GRCh38
rs773299731 Chr.16: 67229617-67229617 on Build GRCh38
rs71659690 Chr.16: 67229476-67229477 on Build GRCh38
rs574100635 Chr.16: 67229375-67229375 on Build GRCh38
rs941828768 Chr.16: 67229504-67229504 on Build GRCh38
rs976443028 Chr.16: 67229477-67229477 on Build GRCh38
rs746773202 Chr.16: 67229683-67229683 on Build GRCh38
rs3178090 Chr.16: 67229402-67229402 on Build GRCh38
rs925434040 Chr.16: 67229463-67229464 on Build GRCh38
rs766020302 Chr.16: 67229621-67229621 on Build GRCh38
rs12448025 Chr.16: 67229582-67229582 on Build GRCh38
rs531215761 Chr.16: 67229577-67229577 on Build GRCh38
rs923053258 Chr.16: 67229485-67229485 on Build GRCh38
rs925773671 Chr.16: 67229745-67229745 on Build GRCh38
rs905931381 Chr.16: 67229550-67229550 on Build GRCh38
rs943799273 Chr.16: 67229426-67229426 on Build GRCh38
rs745380813 Chr.16: 67229473-67229492 on Build GRCh38
rs759065927 Chr.16: 67229622-67229622 on Build GRCh38
rs778956538 Chr.16: 67229788-67229788 on Build GRCh38
rs908181106 Chr.16: 67229425-67229425 on Build GRCh38
rs545316719 Chr.16: 67229551-67229551 on Build GRCh38
rs761295490 Chr.16: 67229450-67229455 on Build GRCh38
rs1042887562 Chr.16: 67229492-67229492 on Build GRCh38
rs895051108 Chr.16: 67229523-67229523 on Build GRCh38
rs752389945 Chr.16: 67229628-67229628 on Build GRCh38
rs779259038 Chr.16: 67229784-67229784 on Build GRCh38
rs777674042 Chr.16: 67229673-67229673 on Build GRCh38
rs760644630 Chr.16: 67229619-67229619 on Build GRCh38
rs376778624 Chr.16: 67229692-67229692 on Build GRCh38
rs762626115 Chr.16: 67229709-67229709 on Build GRCh38
rs771017040 Chr.16: 67229705-67229705 on Build GRCh38
rs904477147 Chr.16: 67229503-67229503 on Build GRCh38
rs756994428 Chr.16: 67229454-67229454 on Build GRCh38
rs549419870 Chr.16: 67229605-67229605 on Build GRCh38
rs764469417 Chr.16: 67229633-67229633 on Build GRCh38
rs901587790 Chr.16: 67229302-67229302 on Build GRCh38
rs985760429 Chr.16: 67229476-67229479 on Build GRCh38

View Full Product Details