Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8190061-8190599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS6
Entrez Gene ID
54785
Gene Name
BLOC-1 related complex subunit 6
Gene Aliases
C17orf59, PRO2472
UniGene
Hs.129563
Gene Chromosome Location
Chr.17: 8188333-8190246 on Build GRCh38
Transcripts
NM_017622.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829931 c.13C>T p.R5W Chr.17: 8190128-8190128 on Build GRCh38
3692059 c.67C>A p.L23I Chr.17: 8190074-8190074 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960987107 Chr.17: 8190223-8190223 on Build GRCh38
rs992761189 Chr.17: 8190226-8190226 on Build GRCh38
rs993806108 Chr.17: 8190491-8190491 on Build GRCh38
rs372143345 Chr.17: 8190450-8190450 on Build GRCh38
rs780744831 Chr.17: 8190419-8190419 on Build GRCh38
rs113766009 Chr.17: 8190301-8190301 on Build GRCh38
rs1022227342 Chr.17: 8190133-8190133 on Build GRCh38
rs1038343274 Chr.17: 8190488-8190488 on Build GRCh38
rs543822329 Chr.17: 8190310-8190310 on Build GRCh38
rs749708322 Chr.17: 8190166-8190166 on Build GRCh38
rs1031178535 Chr.17: 8190209-8190209 on Build GRCh38
rs1025376338 Chr.17: 8190305-8190305 on Build GRCh38
rs918828465 Chr.17: 8190542-8190542 on Build GRCh38
rs1055889472 Chr.17: 8190345-8190345 on Build GRCh38
rs966338039 Chr.17: 8190137-8190137 on Build GRCh38
rs889276586 Chr.17: 8190485-8190485 on Build GRCh38
rs1033962336 Chr.17: 8190099-8190099 on Build GRCh38
rs562226848 Chr.17: 8190455-8190455 on Build GRCh38
rs941541255 Chr.17: 8190224-8190224 on Build GRCh38
rs958687520 Chr.17: 8190389-8190389 on Build GRCh38
rs192809903 Chr.17: 8190284-8190284 on Build GRCh38
rs1034684811 Chr.17: 8190373-8190373 on Build GRCh38
rs974268563 Chr.17: 8190238-8190238 on Build GRCh38
rs537803711 Chr.17: 8190105-8190105 on Build GRCh38
rs977491678 Chr.17: 8190418-8190418 on Build GRCh38
rs554069484 Chr.17: 8190237-8190237 on Build GRCh38
rs914603684 Chr.17: 8190403-8190403 on Build GRCh38
rs978178702 Chr.17: 8190145-8190145 on Build GRCh38
rs902451452 Chr.17: 8190434-8190434 on Build GRCh38
rs779254592 Chr.17: 8190494-8190494 on Build GRCh38
rs202045414 Chr.17: 8190110-8190110 on Build GRCh38
rs747642911 Chr.17: 8190493-8190493 on Build GRCh38
rs1006883590 Chr.17: 8190259-8190259 on Build GRCh38
rs902041308 Chr.17: 8190116-8190116 on Build GRCh38
rs1041770600 Chr.17: 8190104-8190104 on Build GRCh38
rs756897380 Chr.17: 8190180-8190180 on Build GRCh38
rs544976491 Chr.17: 8190363-8190363 on Build GRCh38
rs778576787 Chr.17: 8190187-8190187 on Build GRCh38
rs577759256 Chr.17: 8190341-8190341 on Build GRCh38
rs990464645 Chr.17: 8190394-8190394 on Build GRCh38
rs779850055 Chr.17: 8190111-8190111 on Build GRCh38
rs895058967 Chr.17: 8190109-8190109 on Build GRCh38
rs755161699 Chr.17: 8190098-8190098 on Build GRCh38
rs561183643 Chr.17: 8190489-8190489 on Build GRCh38
rs933497401 Chr.17: 8190453-8190453 on Build GRCh38
rs955328605 Chr.17: 8190169-8190169 on Build GRCh38
rs758616015 Chr.17: 8190506-8190506 on Build GRCh38
rs1010755761 Chr.17: 8190410-8190410 on Build GRCh38
rs746342404 Chr.17: 8190495-8190495 on Build GRCh38
rs1010788257 Chr.17: 8190125-8190125 on Build GRCh38
rs545302259 Chr.17: 8190350-8190350 on Build GRCh38
rs753205201 Chr.17: 8190108-8190108 on Build GRCh38
rs911350774 Chr.17: 8190186-8190186 on Build GRCh38
rs946169180 Chr.17: 8190102-8190102 on Build GRCh38
rs572292493 Chr.17: 8190241-8190241 on Build GRCh38
rs550050730 Chr.17: 8190548-8190548 on Build GRCh38
rs962360952 Chr.17: 8190291-8190291 on Build GRCh38
rs555143691 Chr.17: 8190289-8190289 on Build GRCh38
rs937459319 Chr.17: 8190328-8190328 on Build GRCh38
rs942328337 Chr.17: 8190487-8190488 on Build GRCh38
rs974237651 Chr.17: 8190536-8190536 on Build GRCh38
rs538941740 Chr.17: 8190202-8190202 on Build GRCh38
rs971232473 Chr.17: 8190324-8190324 on Build GRCh38
rs751184238 Chr.17: 8190286-8190286 on Build GRCh38
rs1046179123 Chr.17: 8190300-8190300 on Build GRCh38
rs562495675 Chr.17: 8190338-8190338 on Build GRCh38
rs531820273 Chr.17: 8190507-8190507 on Build GRCh38
rs549162190 Chr.17: 8190477-8190477 on Build GRCh38
rs955070998 Chr.17: 8190488-8190488 on Build GRCh38
rs188696819 Chr.17: 8190161-8190161 on Build GRCh38

View Full Product Details