Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32860495-32861023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A9
Entrez Gene ID
11136
Gene Name
solute carrier family 7 member 9
Gene Aliases
BAT1, CSNU3
UniGene
Hs.743345
Gene Chromosome Location
Chr.19: 32830511-32870958 on Build GRCh38
Transcripts
NM_001126335.1 NM_001243036.1 NM_014270.4 XM_006722992.1 XM_011526402.2 XM_017026230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6539125 c.737G>C p.R246T Chr.19: 32860618-32860618 on Build GRCh38
1680872 c.709C>G p.Q237E Chr.19: 32860646-32860646 on Build GRCh38
5577750 c.711A>G p.Q237Q Chr.19: 32860644-32860644 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879096200 Chr.19: 32860787-32860787 on Build GRCh38
rs911728155 Chr.19: 32860698-32860698 on Build GRCh38
rs374200108 Chr.19: 32860578-32860578 on Build GRCh38
rs867047808 Chr.19: 32860993-32860993 on Build GRCh38
rs559399074 Chr.19: 32860921-32860921 on Build GRCh38
rs936703669 Chr.19: 32860821-32860821 on Build GRCh38
rs759350035 Chr.19: 32860579-32860579 on Build GRCh38
rs867949699 Chr.19: 32860714-32860714 on Build GRCh38
rs763899265 Chr.19: 32860687-32860687 on Build GRCh38
rs777371504 Chr.19: 32860633-32860633 on Build GRCh38
rs546022826 Chr.19: 32860810-32860810 on Build GRCh38
rs1016193217 Chr.19: 32860551-32860551 on Build GRCh38
rs572964075 Chr.19: 32860983-32860983 on Build GRCh38
rs750421080 Chr.19: 32860601-32860601 on Build GRCh38
rs545404042 Chr.19: 32860551-32860555 on Build GRCh38
rs766529640 Chr.19: 32860606-32860606 on Build GRCh38
rs769163411 Chr.19: 32860555-32860555 on Build GRCh38
rs1025228673 Chr.19: 32860890-32860890 on Build GRCh38
rs1024777760 Chr.19: 32860878-32860878 on Build GRCh38
rs368278789 Chr.19: 32860590-32860590 on Build GRCh38
rs548987068 Chr.19: 32860569-32860569 on Build GRCh38
rs371514939 Chr.19: 32860924-32860924 on Build GRCh38
rs1012022005 Chr.19: 32860873-32860873 on Build GRCh38
rs1032558543 Chr.19: 32860515-32860515 on Build GRCh38
rs2287878 Chr.19: 32860789-32860789 on Build GRCh38
rs148824276 Chr.19: 32860646-32860646 on Build GRCh38
rs1053875894 Chr.19: 32860847-32860847 on Build GRCh38
rs565790816 Chr.19: 32860572-32860572 on Build GRCh38
rs760729290 Chr.19: 32860699-32860699 on Build GRCh38
rs761842364 Chr.19: 32860597-32860597 on Build GRCh38
rs748662662 Chr.19: 32860627-32860627 on Build GRCh38
rs965933960 Chr.19: 32860986-32860986 on Build GRCh38
rs370792507 Chr.19: 32860573-32860573 on Build GRCh38
rs755331994 Chr.19: 32860618-32860618 on Build GRCh38
rs966707549 Chr.19: 32860943-32860943 on Build GRCh38
rs775339719 Chr.19: 32860674-32860674 on Build GRCh38
rs765789681 Chr.19: 32860629-32860630 on Build GRCh38
rs923502374 Chr.19: 32860729-32860729 on Build GRCh38
rs886079401 Chr.19: 32860808-32860808 on Build GRCh38
rs1003635332 Chr.19: 32860580-32860580 on Build GRCh38
rs959230142 Chr.19: 32860525-32860525 on Build GRCh38
rs989921765 Chr.19: 32860599-32860599 on Build GRCh38
rs753450871 Chr.19: 32860639-32860643 on Build GRCh38
rs779376859 Chr.19: 32860557-32860557 on Build GRCh38
rs754802320 Chr.19: 32860647-32860648 on Build GRCh38
rs374602349 Chr.19: 32860563-32860563 on Build GRCh38
rs114421824 Chr.19: 32860600-32860600 on Build GRCh38
rs999858855 Chr.19: 32860962-32860962 on Build GRCh38
rs760452532 Chr.19: 32860625-32860625 on Build GRCh38
rs943544729 Chr.19: 32860710-32860710 on Build GRCh38
rs542618537 Chr.19: 32860919-32860919 on Build GRCh38
rs537325259 Chr.19: 32860765-32860765 on Build GRCh38
rs994046423 Chr.19: 32860889-32860889 on Build GRCh38
rs371669498 Chr.19: 32860648-32860648 on Build GRCh38
rs534802508 Chr.19: 32860581-32860581 on Build GRCh38
rs145079475 Chr.19: 32860879-32860879 on Build GRCh38
rs773347847 Chr.19: 32860693-32860693 on Build GRCh38
rs895244722 Chr.19: 32860865-32860865 on Build GRCh38
rs1012536083 Chr.19: 32860541-32860541 on Build GRCh38
rs201306812 Chr.19: 32860673-32860673 on Build GRCh38
rs906668882 Chr.19: 32860575-32860575 on Build GRCh38
rs542721996 Chr.19: 32860775-32860790 on Build GRCh38
rs765230948 Chr.19: 32860692-32860693 on Build GRCh38
rs754257458 Chr.19: 32860614-32860614 on Build GRCh38
rs762970140 Chr.19: 32860684-32860684 on Build GRCh38
rs146682721 Chr.19: 32860632-32860632 on Build GRCh38
rs779435584 Chr.19: 32860619-32860619 on Build GRCh38
rs775874610 Chr.19: 32860806-32860806 on Build GRCh38
rs186323275 Chr.19: 32860516-32860516 on Build GRCh38
rs368670810 Chr.19: 32860653-32860653 on Build GRCh38
rs760090711 Chr.19: 32860740-32860740 on Build GRCh38
rs749693176 Chr.19: 32860746-32860747 on Build GRCh38
rs534514201 Chr.19: 32860715-32860715 on Build GRCh38
rs921726894 Chr.19: 32860751-32860751 on Build GRCh38

View Full Product Details