Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44118109-44118604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF225
Entrez Gene ID
7768
Gene Name
zinc finger protein 225
Gene Aliases
-
UniGene
Hs.279567
Gene Chromosome Location
Chr.19: 44111426-44134816 on Build GRCh38
Transcripts
NM_001321685.1 NM_013362.3 XM_011527285.2 XM_011527286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5114811 c.140T>C p.V47A Chr.19: 44118312-44118312 on Build GRCh38
4617936 c.100C>T p.R34* Chr.19: 44118272-44118272 on Build GRCh38
6264136 c.105A>T p.E35D Chr.19: 44118277-44118277 on Build GRCh38
6264135 c.36C>T p.D12D Chr.19: 44118208-44118208 on Build GRCh38
4079076 c.70C>G p.L24V Chr.19: 44118242-44118242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370346059 Chr.19: 44118454-44118454 on Build GRCh38
rs773921352 Chr.19: 44118519-44118519 on Build GRCh38
rs915111389 Chr.19: 44118148-44118148 on Build GRCh38
rs777399992 Chr.19: 44118272-44118272 on Build GRCh38
rs745804099 Chr.19: 44118485-44118485 on Build GRCh38
rs760215963 Chr.19: 44118329-44118329 on Build GRCh38
rs748227060 Chr.19: 44118542-44118543 on Build GRCh38
rs202046565 Chr.19: 44118195-44118195 on Build GRCh38
rs762208576 Chr.19: 44118507-44118507 on Build GRCh38
rs1031298277 Chr.19: 44118491-44118491 on Build GRCh38
rs906634382 Chr.19: 44118361-44118361 on Build GRCh38
rs778285902 Chr.19: 44118144-44118144 on Build GRCh38
rs765768361 Chr.19: 44118437-44118437 on Build GRCh38
rs558322207 Chr.19: 44118395-44118395 on Build GRCh38
rs751544140 Chr.19: 44118554-44118554 on Build GRCh38
rs919505701 Chr.19: 44118307-44118307 on Build GRCh38
rs762873425 Chr.19: 44118191-44118191 on Build GRCh38
rs1051886406 Chr.19: 44118406-44118407 on Build GRCh38
rs754413257 Chr.19: 44118233-44118233 on Build GRCh38
rs940180003 Chr.19: 44118522-44118522 on Build GRCh38
rs1033572008 Chr.19: 44118132-44118132 on Build GRCh38
rs959901402 Chr.19: 44118160-44118160 on Build GRCh38
rs758561332 Chr.19: 44118461-44118461 on Build GRCh38
rs764363712 Chr.19: 44118223-44118223 on Build GRCh38
rs76696522 Chr.19: 44118527-44118527 on Build GRCh38
rs185929255 Chr.19: 44118137-44118137 on Build GRCh38
rs766362247 Chr.19: 44118462-44118462 on Build GRCh38
rs989450031 Chr.19: 44118404-44118404 on Build GRCh38
rs974374861 Chr.19: 44118486-44118486 on Build GRCh38
rs115124536 Chr.19: 44118135-44118135 on Build GRCh38
rs62640901 Chr.19: 44118305-44118305 on Build GRCh38
rs761162937 Chr.19: 44118215-44118215 on Build GRCh38
rs373963033 Chr.19: 44118474-44118474 on Build GRCh38
rs771471890 Chr.19: 44118323-44118323 on Build GRCh38
rs768150427 Chr.19: 44118495-44118495 on Build GRCh38
rs34863330 Chr.19: 44118488-44118488 on Build GRCh38
rs199991881 Chr.19: 44118176-44118176 on Build GRCh38
rs764573428 Chr.19: 44118330-44118330 on Build GRCh38
rs114271963 Chr.19: 44118489-44118489 on Build GRCh38
rs147545000 Chr.19: 44118260-44118260 on Build GRCh38
rs770267059 Chr.19: 44118514-44118514 on Build GRCh38
rs748611472 Chr.19: 44118396-44118396 on Build GRCh38
rs773257562 Chr.19: 44118178-44118178 on Build GRCh38
rs764549796 Chr.19: 44118550-44118551 on Build GRCh38
rs563327487 Chr.19: 44118545-44118545 on Build GRCh38
rs749271387 Chr.19: 44118278-44118278 on Build GRCh38
rs868601142 Chr.19: 44118403-44118403 on Build GRCh38
rs377676634 Chr.19: 44118161-44118161 on Build GRCh38
rs944469371 Chr.19: 44118483-44118483 on Build GRCh38
rs747907664 Chr.19: 44118174-44118174 on Build GRCh38
rs757201822 Chr.19: 44118479-44118479 on Build GRCh38
rs911646377 Chr.19: 44118173-44118173 on Build GRCh38
rs773802408 Chr.19: 44118197-44118197 on Build GRCh38
rs540946151 Chr.19: 44118480-44118480 on Build GRCh38
rs548588769 Chr.19: 44118164-44118164 on Build GRCh38
rs371799405 Chr.19: 44118578-44118578 on Build GRCh38
rs775282283 Chr.19: 44118326-44118326 on Build GRCh38
rs752903565 Chr.19: 44118572-44118572 on Build GRCh38
rs747492231 Chr.19: 44118504-44118504 on Build GRCh38
rs751846827 Chr.19: 44118473-44118473 on Build GRCh38
rs374098472 Chr.19: 44118199-44118199 on Build GRCh38
rs552879055 Chr.19: 44118159-44118159 on Build GRCh38
rs750485489 Chr.19: 44118256-44118256 on Build GRCh38
rs569516660 Chr.19: 44118268-44118268 on Build GRCh38
rs765426722 Chr.19: 44118244-44118244 on Build GRCh38
rs772503035 Chr.19: 44118549-44118550 on Build GRCh38
rs755213333 Chr.19: 44118473-44118473 on Build GRCh38
rs1003505968 Chr.19: 44118367-44118367 on Build GRCh38
rs888708572 Chr.19: 44118517-44118517 on Build GRCh38
rs767133516 Chr.19: 44118208-44118208 on Build GRCh38
rs780261904 Chr.19: 44118267-44118267 on Build GRCh38
rs775732314 Chr.19: 44118209-44118209 on Build GRCh38
rs767670071 Chr.19: 44118569-44118569 on Build GRCh38
rs745326237 Chr.19: 44118319-44118319 on Build GRCh38
rs778925210 Chr.19: 44118516-44118516 on Build GRCh38
rs895745901 Chr.19: 44118499-44118499 on Build GRCh38
rs191225974 Chr.19: 44118476-44118476 on Build GRCh38
rs374196454 Chr.19: 44118150-44118150 on Build GRCh38
rs746700398 Chr.19: 44118493-44118493 on Build GRCh38
rs373559946 Chr.19: 44118286-44118286 on Build GRCh38
rs1002319676 Chr.19: 44118538-44118538 on Build GRCh38
rs759809853 Chr.19: 44118557-44118557 on Build GRCh38
rs777091355 Chr.19: 44118356-44118356 on Build GRCh38
rs762437672 Chr.19: 44118360-44118360 on Build GRCh38
rs780878493 Chr.19: 44118171-44118171 on Build GRCh38
rs376939625 Chr.19: 44118241-44118241 on Build GRCh38
rs568335959 Chr.19: 44118177-44118177 on Build GRCh38

View Full Product Details