Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 82364266-82364850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPBG
Entrez Gene ID
7162
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, 5T4AG, M6P1, WAIF1
UniGene
Hs.71947
Gene Chromosome Location
Chr.6: 82363206-82370828 on Build GRCh38
Transcripts
NM_001166392.1 NM_006670.4 XM_011536097.2

SNP Detail

SNP ID Chromosome Location
rs957844988 Chr.6: 82364618-82364618 on Build GRCh38
rs904816133 Chr.6: 82364473-82364473 on Build GRCh38
rs942021546 Chr.6: 82364352-82364352 on Build GRCh38
rs757951052 Chr.6: 82364607-82364607 on Build GRCh38
rs561344214 Chr.6: 82364371-82364371 on Build GRCh38
rs1017280045 Chr.6: 82364497-82364497 on Build GRCh38
rs1020912567 Chr.6: 82364805-82364805 on Build GRCh38
rs920778206 Chr.6: 82364715-82364715 on Build GRCh38
rs1025126268 Chr.6: 82364292-82364292 on Build GRCh38
rs893670724 Chr.6: 82364771-82364771 on Build GRCh38
rs891984186 Chr.6: 82364410-82364410 on Build GRCh38
rs899298723 Chr.6: 82364484-82364484 on Build GRCh38
rs1030067215 Chr.6: 82364518-82364518 on Build GRCh38
rs976280064 Chr.6: 82364330-82364331 on Build GRCh38
rs964799160 Chr.6: 82364705-82364705 on Build GRCh38
rs906287524 Chr.6: 82364592-82364592 on Build GRCh38
rs1039080802 Chr.6: 82364699-82364699 on Build GRCh38
rs1054785701 Chr.6: 82364761-82364761 on Build GRCh38
rs1016053239 Chr.6: 82364768-82364768 on Build GRCh38
rs908111693 Chr.6: 82364723-82364723 on Build GRCh38
rs938784277 Chr.6: 82364368-82364368 on Build GRCh38
rs1010440042 Chr.6: 82364464-82364464 on Build GRCh38
rs957950065 Chr.6: 82364339-82364339 on Build GRCh38
rs983285455 Chr.6: 82364574-82364574 on Build GRCh38
rs1010849890 Chr.6: 82364641-82364641 on Build GRCh38
rs745560221 Chr.6: 82364497-82364497 on Build GRCh38
rs538458401 Chr.6: 82364464-82364464 on Build GRCh38
rs778833888 Chr.6: 82364436-82364436 on Build GRCh38
rs574187183 Chr.6: 82364646-82364646 on Build GRCh38
rs1017770246 Chr.6: 82364316-82364316 on Build GRCh38
rs963332873 Chr.6: 82364605-82364605 on Build GRCh38
rs546805469 Chr.6: 82364452-82364452 on Build GRCh38
rs182948418 Chr.6: 82364454-82364454 on Build GRCh38
rs1020606032 Chr.6: 82364286-82364286 on Build GRCh38
rs911973586 Chr.6: 82364401-82364401 on Build GRCh38
rs980562548 Chr.6: 82364300-82364300 on Build GRCh38
rs1004638213 Chr.6: 82364760-82364760 on Build GRCh38
rs1001920819 Chr.6: 82364754-82364754 on Build GRCh38
rs375772233 Chr.6: 82364695-82364695 on Build GRCh38
rs530897752 Chr.6: 82364296-82364296 on Build GRCh38
rs1030034723 Chr.6: 82364750-82364750 on Build GRCh38
rs371418051 Chr.6: 82364557-82364557 on Build GRCh38
rs939739797 Chr.6: 82364731-82364731 on Build GRCh38
rs1039222180 Chr.6: 82364479-82364479 on Build GRCh38
rs980056675 Chr.6: 82364790-82364790 on Build GRCh38
rs1033480028 Chr.6: 82364611-82364611 on Build GRCh38
rs902734057 Chr.6: 82364643-82364643 on Build GRCh38
rs945352533 Chr.6: 82364373-82364373 on Build GRCh38
rs1048004454 Chr.6: 82364722-82364722 on Build GRCh38
rs913343571 Chr.6: 82364677-82364677 on Build GRCh38
rs997286966 Chr.6: 82364504-82364504 on Build GRCh38
rs1000933353 Chr.6: 82364478-82364478 on Build GRCh38
rs1046046820 Chr.6: 82364742-82364742 on Build GRCh38
rs573360720 Chr.6: 82364753-82364753 on Build GRCh38
rs552419434 Chr.6: 82364465-82364465 on Build GRCh38
rs537827290 Chr.6: 82364570-82364570 on Build GRCh38
rs770911 Chr.6: 82364648-82364648 on Build GRCh38
rs1046178693 Chr.6: 82364566-82364566 on Build GRCh38
rs1011151079 Chr.6: 82364786-82364786 on Build GRCh38
rs550730339 Chr.6: 82364334-82364334 on Build GRCh38
rs539104913 Chr.6: 82364705-82364706 on Build GRCh38
rs113229038 Chr.6: 82364651-82364651 on Build GRCh38
rs927231407 Chr.6: 82364800-82364800 on Build GRCh38
rs946194993 Chr.6: 82364680-82364680 on Build GRCh38
rs886802248 Chr.6: 82364525-82364525 on Build GRCh38
rs1007159714 Chr.6: 82364698-82364698 on Build GRCh38
rs964528008 Chr.6: 82364330-82364330 on Build GRCh38
rs553456323 Chr.6: 82364668-82364668 on Build GRCh38
rs987605004 Chr.6: 82364376-82364376 on Build GRCh38
rs950532980 Chr.6: 82364567-82364567 on Build GRCh38
rs757177631 Chr.6: 82364318-82364318 on Build GRCh38
rs1048421204 Chr.6: 82364524-82364524 on Build GRCh38
rs967031990 Chr.6: 82364823-82364823 on Build GRCh38
rs773100803 Chr.6: 82364325-82364325 on Build GRCh38
rs530262239 Chr.6: 82364411-82364411 on Build GRCh38
rs116233429 Chr.6: 82364603-82364603 on Build GRCh38
rs974797401 Chr.6: 82364713-82364713 on Build GRCh38
rs569091486 Chr.6: 82364564-82364564 on Build GRCh38
rs913923595 Chr.6: 82364359-82364359 on Build GRCh38
rs930787490 Chr.6: 82364720-82364720 on Build GRCh38
rs1043281722 Chr.6: 82364683-82364683 on Build GRCh38
rs963603408 Chr.6: 82364780-82364780 on Build GRCh38
rs989768246 Chr.6: 82364349-82364349 on Build GRCh38
rs1044434912 Chr.6: 82364358-82364358 on Build GRCh38
rs545290727 Chr.6: 82364758-82364758 on Build GRCh38

View Full Product Details