Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83057631-83058229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE3D
Entrez Gene ID
90025
Gene Name
ubiquitin protein ligase E3D
Gene Aliases
C6orf157, H10BH, UBE2CBP, YJR141W
UniGene
Hs.148609
Gene Chromosome Location
Chr.6: 82880834-83065841 on Build GRCh38
Transcripts
NM_001304437.1 NM_198920.2 XM_011536238.2 XM_011536239.2 XM_011536240.1 XM_011536241.2 XM_011536242.1 XM_017011458.1 XM_017011459.1 XM_017011460.1 XM_017011461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4453276 c.273A>T p.T91T Chr.6: 83057827-83057827 on Build GRCh38
1621975 c.103A>T p.M35L Chr.6: 83057997-83057997 on Build GRCh38
597461 c.105G>A p.M35I Chr.6: 83057995-83057995 on Build GRCh38
7422432 c.85A>T p.K29* Chr.6: 83058015-83058015 on Build GRCh38
1446135 c.178G>A p.E60K Chr.6: 83057922-83057922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188069755 Chr.6: 83058176-83058176 on Build GRCh38
rs905304446 Chr.6: 83058103-83058103 on Build GRCh38
rs529317738 Chr.6: 83058042-83058042 on Build GRCh38
rs113046065 Chr.6: 83058065-83058065 on Build GRCh38
rs1023768999 Chr.6: 83058188-83058188 on Build GRCh38
rs770710991 Chr.6: 83057985-83057985 on Build GRCh38
rs1013742494 Chr.6: 83057821-83057821 on Build GRCh38
rs960892136 Chr.6: 83057750-83057750 on Build GRCh38
rs761142679 Chr.6: 83057837-83057837 on Build GRCh38
rs765904900 Chr.6: 83057919-83057919 on Build GRCh38
rs368913478 Chr.6: 83058002-83058002 on Build GRCh38
rs911137872 Chr.6: 83058013-83058013 on Build GRCh38
rs1035011489 Chr.6: 83058142-83058142 on Build GRCh38
rs1025503020 Chr.6: 83057674-83057674 on Build GRCh38
rs781324476 Chr.6: 83057968-83057968 on Build GRCh38
rs780386712 Chr.6: 83057862-83057862 on Build GRCh38
rs998489592 Chr.6: 83057662-83057662 on Build GRCh38
rs771453519 Chr.6: 83057894-83057894 on Build GRCh38
rs771997693 Chr.6: 83058011-83058011 on Build GRCh38
rs751609273 Chr.6: 83057958-83057958 on Build GRCh38
rs372665243 Chr.6: 83058014-83058014 on Build GRCh38
rs549923372 Chr.6: 83057887-83057887 on Build GRCh38
rs765662214 Chr.6: 83058030-83058030 on Build GRCh38
rs758902367 Chr.6: 83057818-83057818 on Build GRCh38
rs776412790 Chr.6: 83057845-83057845 on Build GRCh38
rs988451424 Chr.6: 83058177-83058177 on Build GRCh38
rs145853060 Chr.6: 83057926-83057926 on Build GRCh38
rs1021618142 Chr.6: 83058184-83058184 on Build GRCh38
rs750546715 Chr.6: 83057856-83057856 on Build GRCh38
rs896233714 Chr.6: 83058125-83058125 on Build GRCh38
rs766255259 Chr.6: 83057794-83057794 on Build GRCh38
rs770858305 Chr.6: 83057881-83057881 on Build GRCh38
rs773814085 Chr.6: 83058031-83058031 on Build GRCh38
rs749093280 Chr.6: 83057971-83057971 on Build GRCh38
rs549184045 Chr.6: 83058058-83058058 on Build GRCh38
rs993909544 Chr.6: 83057692-83057692 on Build GRCh38
rs898241877 Chr.6: 83057678-83057678 on Build GRCh38
rs773881725 Chr.6: 83057806-83057806 on Build GRCh38
rs1038006784 Chr.6: 83057651-83057651 on Build GRCh38
rs113991038 Chr.6: 83057759-83057759 on Build GRCh38
rs925267089 Chr.6: 83057860-83057860 on Build GRCh38
rs779971163 Chr.6: 83057943-83057943 on Build GRCh38
rs755223808 Chr.6: 83057965-83057965 on Build GRCh38
rs140716565 Chr.6: 83057895-83057895 on Build GRCh38
rs762854726 Chr.6: 83057775-83057775 on Build GRCh38
rs764532079 Chr.6: 83057840-83057840 on Build GRCh38
rs769784096 Chr.6: 83058038-83058039 on Build GRCh38
rs1023931738 Chr.6: 83057851-83057851 on Build GRCh38
rs933868097 Chr.6: 83058001-83058001 on Build GRCh38
rs1016683970 Chr.6: 83058079-83058079 on Build GRCh38
rs571518732 Chr.6: 83058194-83058194 on Build GRCh38
rs184292340 Chr.6: 83057896-83057896 on Build GRCh38
rs375701422 Chr.6: 83058017-83058017 on Build GRCh38
rs572734870 Chr.6: 83057682-83057682 on Build GRCh38
rs758392896 Chr.6: 83057861-83057861 on Build GRCh38
rs1039407018 Chr.6: 83057969-83057969 on Build GRCh38
rs925702494 Chr.6: 83057948-83057948 on Build GRCh38
rs865955413 Chr.6: 83057900-83057900 on Build GRCh38
rs752289218 Chr.6: 83057836-83057836 on Build GRCh38
rs566915738 Chr.6: 83058122-83058122 on Build GRCh38
rs781134325 Chr.6: 83057864-83057864 on Build GRCh38
rs942389762 Chr.6: 83057961-83057961 on Build GRCh38
rs762214588 Chr.6: 83057901-83057901 on Build GRCh38
rs767260415 Chr.6: 83057829-83057829 on Build GRCh38
rs778863289 Chr.6: 83057997-83057997 on Build GRCh38
rs751595538 Chr.6: 83058063-83058063 on Build GRCh38
rs969410907 Chr.6: 83057769-83057769 on Build GRCh38
rs759154052 Chr.6: 83057890-83057890 on Build GRCh38
rs564623498 Chr.6: 83057727-83057727 on Build GRCh38
rs762331532 Chr.6: 83058024-83058024 on Build GRCh38
rs776412780 Chr.6: 83058018-83058018 on Build GRCh38
rs779224227 Chr.6: 83057762-83057762 on Build GRCh38
rs777566338 Chr.6: 83057869-83057869 on Build GRCh38
rs757874696 Chr.6: 83057848-83057848 on Build GRCh38
rs758542673 Chr.6: 83057934-83057934 on Build GRCh38
rs993819469 Chr.6: 83058069-83058069 on Build GRCh38
rs545029891 Chr.6: 83057724-83057724 on Build GRCh38
rs146891196 Chr.6: 83057853-83057853 on Build GRCh38
rs866363489 Chr.6: 83057693-83057693 on Build GRCh38
rs761484906 Chr.6: 83058021-83058021 on Build GRCh38
rs1005803206 Chr.6: 83058070-83058070 on Build GRCh38

View Full Product Details