Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99029577-99030112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927550
Entrez Gene ID
101927550
Gene Name
uncharacterized LOC101927550
Gene Aliases
-
UniGene
Hs.636663
Gene Chromosome Location
Chr.7: 99013165-99036492 on Build GRCh38
Transcripts
NR_110102.1 NM_001199847.1 NM_020429.2 NM_181349.2 XM_017012457.1

Gene Symbol
SMURF1
Entrez Gene ID
57154
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases
-
UniGene
Hs.189329
Gene Chromosome Location
Chr.7: 99027435-99144120 on Build GRCh38
Transcripts
NR_110102.1 NM_001199847.1 NM_020429.2 NM_181349.2 XM_017012457.1

SNP Detail

SNP ID Chromosome Location
rs888512889 Chr.7: 99029901-99029901 on Build GRCh38
rs943734520 Chr.7: 99029935-99029935 on Build GRCh38
rs777832981 Chr.7: 99029792-99029792 on Build GRCh38
rs565552073 Chr.7: 99030004-99030004 on Build GRCh38
rs554302124 Chr.7: 99029825-99029826 on Build GRCh38
rs936637534 Chr.7: 99029623-99029623 on Build GRCh38
rs965589960 Chr.7: 99029785-99029785 on Build GRCh38
rs34666591 Chr.7: 99029826-99029826 on Build GRCh38
rs925172570 Chr.7: 99029824-99029825 on Build GRCh38
rs886934738 Chr.7: 99030059-99030059 on Build GRCh38
rs1030038441 Chr.7: 99029985-99029985 on Build GRCh38
rs219810 Chr.7: 99029840-99029840 on Build GRCh38
rs772032001 Chr.7: 99029729-99029729 on Build GRCh38
rs772179761 Chr.7: 99029720-99029720 on Build GRCh38
rs190655030 Chr.7: 99029800-99029800 on Build GRCh38
rs146527669 Chr.7: 99029634-99029634 on Build GRCh38
rs912236506 Chr.7: 99029934-99029934 on Build GRCh38
rs539973732 Chr.7: 99030081-99030081 on Build GRCh38
rs970652036 Chr.7: 99030087-99030087 on Build GRCh38
rs895171547 Chr.7: 99029636-99029636 on Build GRCh38
rs899569977 Chr.7: 99029637-99029637 on Build GRCh38
rs773369335 Chr.7: 99029797-99029797 on Build GRCh38
rs960056133 Chr.7: 99029811-99029811 on Build GRCh38
rs568468826 Chr.7: 99029608-99029608 on Build GRCh38
rs554939708 Chr.7: 99029875-99029875 on Build GRCh38
rs567199372 Chr.7: 99029704-99029704 on Build GRCh38
rs1048938965 Chr.7: 99029893-99029893 on Build GRCh38
rs1004566582 Chr.7: 99030080-99030080 on Build GRCh38
rs968755624 Chr.7: 99029842-99029842 on Build GRCh38
rs1036135121 Chr.7: 99029783-99029783 on Build GRCh38
rs543841219 Chr.7: 99029979-99029979 on Build GRCh38
rs1007482196 Chr.7: 99029910-99029910 on Build GRCh38
rs957783879 Chr.7: 99030039-99030039 on Build GRCh38
rs925107820 Chr.7: 99029617-99029617 on Build GRCh38
rs548944728 Chr.7: 99029641-99029641 on Build GRCh38
rs183078715 Chr.7: 99030033-99030033 on Build GRCh38
rs1046354230 Chr.7: 99029684-99029684 on Build GRCh38
rs888599219 Chr.7: 99029761-99029764 on Build GRCh38
rs538410427 Chr.7: 99029837-99029837 on Build GRCh38
rs78501487 Chr.7: 99029873-99029873 on Build GRCh38
rs899601030 Chr.7: 99029997-99029997 on Build GRCh38
rs145442980 Chr.7: 99029624-99029624 on Build GRCh38
rs182630195 Chr.7: 99029708-99029708 on Build GRCh38
rs975753220 Chr.7: 99029788-99029788 on Build GRCh38
rs1008420480 Chr.7: 99029618-99029618 on Build GRCh38
rs930104241 Chr.7: 99029879-99029879 on Build GRCh38
rs966355974 Chr.7: 99029996-99029996 on Build GRCh38
rs1048257867 Chr.7: 99030050-99030050 on Build GRCh38
rs111233664 Chr.7: 99029862-99029862 on Build GRCh38
rs901429714 Chr.7: 99029955-99029955 on Build GRCh38
rs576463655 Chr.7: 99029926-99029926 on Build GRCh38
rs185803688 Chr.7: 99029751-99029751 on Build GRCh38
rs1007164489 Chr.7: 99029761-99029761 on Build GRCh38
rs1004131386 Chr.7: 99029709-99029709 on Build GRCh38
rs1026743808 Chr.7: 99029653-99029653 on Build GRCh38

View Full Product Details