Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 152136514-152137109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT2C
Entrez Gene ID
58508
Gene Name
lysine methyltransferase 2C
Gene Aliases
HALR, MLL3
UniGene
Hs.647120
Gene Chromosome Location
Chr.7: 152134925-152436642 on Build GRCh38
Transcripts
NM_170606.2 XM_005250025.4 XM_005250026.3 XM_005250027.4 XM_005250028.4 XM_005250031.4 XM_006716077.3 XM_006716078.3 XM_006716079.3 XM_011516450.2 XM_011516451.2 XM_011516452.2 XM_011516453.2 XM_011516454.2 XM_011516456.2 XM_017012480.1 XM_017012481.1 XM_017012482.1 XM_017012483.1 XM_017012484.1 XM_017012485.1 XM_017012486.1 XM_017012487.1 XM_017012488.1 XM_017012489.1 XM_017012490.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4808474 c.14815-6G>A p.? Chr.7: 152136930-152136930 on Build GRCh38
1184380 c.14651A>G p.Y4884C Chr.7: 152136917-152136917 on Build GRCh38
1673692 c.14836G>A p.D4946N Chr.7: 152136903-152136903 on Build GRCh38
1292465 c.14891G>A p.R4964Q Chr.7: 152136848-152136848 on Build GRCh38
7344545 c.14685C>A p.H4895Q Chr.7: 152136883-152136883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777081646 Chr.7: 152136804-152136804 on Build GRCh38
rs797012780 Chr.7: 152136563-152136567 on Build GRCh38
rs376468045 Chr.7: 152136848-152136848 on Build GRCh38
rs748426014 Chr.7: 152136917-152136917 on Build GRCh38
rs753404197 Chr.7: 152137023-152137023 on Build GRCh38
rs750203378 Chr.7: 152136578-152136579 on Build GRCh38
rs550646716 Chr.7: 152136679-152136679 on Build GRCh38
rs760061529 Chr.7: 152136960-152136960 on Build GRCh38
rs549717393 Chr.7: 152136598-152136598 on Build GRCh38
rs183136694 Chr.7: 152136625-152136625 on Build GRCh38
rs780161675 Chr.7: 152137073-152137073 on Build GRCh38
rs1037935167 Chr.7: 152137026-152137026 on Build GRCh38
rs190944263 Chr.7: 152136597-152136597 on Build GRCh38
rs377271632 Chr.7: 152136629-152136629 on Build GRCh38
rs6954722 Chr.7: 152137070-152137070 on Build GRCh38
rs775986465 Chr.7: 152136973-152136973 on Build GRCh38
rs538460804 Chr.7: 152136813-152136813 on Build GRCh38
rs1013951789 Chr.7: 152136780-152136780 on Build GRCh38
rs762659955 Chr.7: 152136768-152136768 on Build GRCh38
rs12333958 Chr.7: 152137023-152137023 on Build GRCh38
rs182850642 Chr.7: 152137075-152137075 on Build GRCh38
rs766726513 Chr.7: 152136798-152136798 on Build GRCh38
rs761180962 Chr.7: 152136793-152136793 on Build GRCh38
rs767781903 Chr.7: 152137028-152137028 on Build GRCh38
rs751955309 Chr.7: 152136977-152136977 on Build GRCh38
rs560899699 Chr.7: 152136565-152136565 on Build GRCh38
rs763756860 Chr.7: 152136827-152136827 on Build GRCh38
rs750905292 Chr.7: 152137040-152137040 on Build GRCh38
rs757587752 Chr.7: 152137006-152137006 on Build GRCh38
rs909382573 Chr.7: 152136579-152136579 on Build GRCh38
rs951001227 Chr.7: 152136552-152136552 on Build GRCh38
rs781675572 Chr.7: 152137065-152137065 on Build GRCh38
rs925886531 Chr.7: 152137071-152137071 on Build GRCh38
rs752440002 Chr.7: 152136812-152136812 on Build GRCh38
rs779818288 Chr.7: 152136909-152136909 on Build GRCh38
rs372314924 Chr.7: 152136803-152136803 on Build GRCh38
rs772205161 Chr.7: 152136784-152136784 on Build GRCh38
rs773266030 Chr.7: 152136786-152136786 on Build GRCh38
rs908129104 Chr.7: 152136641-152136641 on Build GRCh38
rs368866050 Chr.7: 152136875-152136875 on Build GRCh38
rs763142705 Chr.7: 152136616-152136616 on Build GRCh38
rs946001996 Chr.7: 152137051-152137051 on Build GRCh38
rs748338188 Chr.7: 152136783-152136783 on Build GRCh38
rs373607948 Chr.7: 152136932-152136932 on Build GRCh38
rs1050950636 Chr.7: 152136765-152136765 on Build GRCh38
rs765590829 Chr.7: 152136965-152136965 on Build GRCh38
rs762710731 Chr.7: 152136976-152136976 on Build GRCh38
rs186891530 Chr.7: 152136541-152136541 on Build GRCh38
rs145947780 Chr.7: 152136808-152136808 on Build GRCh38
rs751058113 Chr.7: 152136854-152136854 on Build GRCh38
rs1010853949 Chr.7: 152136825-152136825 on Build GRCh38
rs142171905 Chr.7: 152136741-152136741 on Build GRCh38
rs201223677 Chr.7: 152136566-152136567 on Build GRCh38
rs1016897630 Chr.7: 152136959-152136959 on Build GRCh38
rs866613310 Chr.7: 152136690-152136690 on Build GRCh38
rs558765564 Chr.7: 152136870-152136870 on Build GRCh38
rs759420858 Chr.7: 152136640-152136640 on Build GRCh38
rs553009136 Chr.7: 152136770-152136770 on Build GRCh38
rs369844615 Chr.7: 152137018-152137018 on Build GRCh38
rs12333453 Chr.7: 152137027-152137027 on Build GRCh38
rs546261970 Chr.7: 152136638-152136638 on Build GRCh38
rs1013898333 Chr.7: 152136550-152136550 on Build GRCh38
rs1008781964 Chr.7: 152136556-152136557 on Build GRCh38
rs983663512 Chr.7: 152136578-152136578 on Build GRCh38
rs377602852 Chr.7: 152136935-152136935 on Build GRCh38
rs747047595 Chr.7: 152136933-152136933 on Build GRCh38
rs537188192 Chr.7: 152136666-152136666 on Build GRCh38
rs567351628 Chr.7: 152136724-152136724 on Build GRCh38
rs528688034 Chr.7: 152136615-152136615 on Build GRCh38
rs1050488817 Chr.7: 152137068-152137068 on Build GRCh38
rs146009167 Chr.7: 152136874-152136874 on Build GRCh38
rs562774656 Chr.7: 152136551-152136551 on Build GRCh38
rs755902690 Chr.7: 152136883-152136883 on Build GRCh38
rs956598366 Chr.7: 152136809-152136809 on Build GRCh38
rs186692085 Chr.7: 152136748-152136748 on Build GRCh38
rs904370265 Chr.7: 152136667-152136667 on Build GRCh38
rs191450408 Chr.7: 152137017-152137017 on Build GRCh38
rs755323232 Chr.7: 152136805-152136805 on Build GRCh38
rs9969322 Chr.7: 152136607-152136607 on Build GRCh38
rs1055612862 Chr.7: 152136701-152136701 on Build GRCh38
rs777056437 Chr.7: 152136939-152136939 on Build GRCh38
rs542717961 Chr.7: 152136578-152136578 on Build GRCh38
rs886733481 Chr.7: 152136807-152136807 on Build GRCh38

View Full Product Details