Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169089784-169090177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
UniGene
Hs.744090
Gene Chromosome Location
Chr.3: 169083499-169663781 on Build GRCh38
Transcripts
NM_001105077.3 NM_001105078.3 NM_001163999.1 NM_001164000.1 NM_001205194.1 NM_004991.3 NM_005241.3 XM_005247213.3 XM_005247214.3 XM_005247215.3 XM_005247219.2 XM_005247220.2 XM_005247221.2 XM_005247223.2 XM_005247224.3 XM_005247225.3 XM_005247226.3 XM_011512546.2 XM_017005874.1 XM_017005875.1 XM_017005876.1 XM_017005877.1 XM_017005878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7082108 c.2906A>T p.E969V Chr.3: 169090126-169090126 on Build GRCh38
7082110 c.2905G>A p.E969K Chr.3: 169090127-169090127 on Build GRCh38
6020084 c.3256G>A p.D1086N Chr.3: 169090118-169090118 on Build GRCh38
6020083 c.2719G>A p.D907N Chr.3: 169090118-169090118 on Build GRCh38
3915307 c.2933G>A p.G978E Chr.3: 169090099-169090099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373451242 Chr.3: 169090008-169090008 on Build GRCh38
rs960303458 Chr.3: 169090068-169090068 on Build GRCh38
rs755701868 Chr.3: 169089981-169089981 on Build GRCh38
rs935326283 Chr.3: 169089808-169089808 on Build GRCh38
rs200202285 Chr.3: 169089899-169089899 on Build GRCh38
rs752722841 Chr.3: 169090079-169090079 on Build GRCh38
rs146772249 Chr.3: 169089898-169089899 on Build GRCh38
rs752530154 Chr.3: 169089961-169089961 on Build GRCh38
rs765564966 Chr.3: 169090048-169090048 on Build GRCh38
rs756715943 Chr.3: 169090117-169090117 on Build GRCh38
rs542695398 Chr.3: 169089854-169089854 on Build GRCh38
rs376175876 Chr.3: 169090035-169090035 on Build GRCh38
rs386668094 Chr.3: 169089898-169089901 on Build GRCh38
rs761930526 Chr.3: 169090047-169090047 on Build GRCh38
rs751417924 Chr.3: 169089950-169089950 on Build GRCh38
rs755989483 Chr.3: 169090088-169090088 on Build GRCh38
rs575196494 Chr.3: 169089839-169089839 on Build GRCh38
rs747612156 Chr.3: 169090147-169090147 on Build GRCh38
rs892380820 Chr.3: 169089840-169089840 on Build GRCh38
rs34194856 Chr.3: 169089903-169089903 on Build GRCh38
rs759427716 Chr.3: 169090062-169090062 on Build GRCh38
rs147951169 Chr.3: 169090143-169090143 on Build GRCh38
rs201639760 Chr.3: 169089898-169089898 on Build GRCh38
rs745493476 Chr.3: 169090138-169090138 on Build GRCh38
rs540848924 Chr.3: 169089942-169089942 on Build GRCh38
rs776563142 Chr.3: 169090039-169090039 on Build GRCh38
rs879846096 Chr.3: 169089949-169089949 on Build GRCh38
rs749026364 Chr.3: 169090004-169090004 on Build GRCh38
rs777142017 Chr.3: 169089990-169089990 on Build GRCh38
rs116089988 Chr.3: 169089901-169089901 on Build GRCh38
rs754753755 Chr.3: 169089953-169089953 on Build GRCh38
rs753349574 Chr.3: 169090100-169090100 on Build GRCh38
rs768849297 Chr.3: 169090038-169090038 on Build GRCh38
rs767195566 Chr.3: 169090072-169090072 on Build GRCh38
rs183812875 Chr.3: 169089865-169089865 on Build GRCh38
rs386352330 Chr.3: 169090087-169090087 on Build GRCh38
rs778418275 Chr.3: 169089897-169089898 on Build GRCh38
rs369006017 Chr.3: 169089954-169089954 on Build GRCh38
rs778554611 Chr.3: 169090133-169090133 on Build GRCh38
rs562893677 Chr.3: 169090140-169090140 on Build GRCh38
rs1052265559 Chr.3: 169089838-169089838 on Build GRCh38
rs150202188 Chr.3: 169090026-169090026 on Build GRCh38
rs996910323 Chr.3: 169089933-169089933 on Build GRCh38
rs777706418 Chr.3: 169090098-169090098 on Build GRCh38
rs1019209007 Chr.3: 169090095-169090095 on Build GRCh38
rs969559009 Chr.3: 169090120-169090120 on Build GRCh38
rs1015885653 Chr.3: 169090060-169090060 on Build GRCh38
rs188244708 Chr.3: 169090129-169090129 on Build GRCh38
rs1003850573 Chr.3: 169090078-169090078 on Build GRCh38
rs140918202 Chr.3: 169090018-169090018 on Build GRCh38
rs750634357 Chr.3: 169090058-169090058 on Build GRCh38
rs1045265789 Chr.3: 169089928-169089928 on Build GRCh38
rs778470697 Chr.3: 169090007-169090007 on Build GRCh38

View Full Product Details