Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13659129-13659615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
UniGene
Hs.330463
Gene Chromosome Location
Chr.10: 13643706-14330952 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3741577 c.1899-1G>A p.? Chr.10: 13659491-13659491 on Build GRCh38
87722 c.1934C>T p.A645V Chr.10: 13659455-13659455 on Build GRCh38
7145422 c.1925G>A p.G642E Chr.10: 13659464-13659464 on Build GRCh38
6666935 c.2033G>A p.R678Q Chr.10: 13659356-13659356 on Build GRCh38
244603 c.2032C>T p.R678W Chr.10: 13659357-13659357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867452670 Chr.10: 13659380-13659380 on Build GRCh38
rs775683016 Chr.10: 13659269-13659269 on Build GRCh38
rs1011535846 Chr.10: 13659445-13659445 on Build GRCh38
rs990334338 Chr.10: 13659205-13659205 on Build GRCh38
rs773309388 Chr.10: 13659517-13659517 on Build GRCh38
rs752099192 Chr.10: 13659430-13659430 on Build GRCh38
rs368099541 Chr.10: 13659259-13659259 on Build GRCh38
rs560390559 Chr.10: 13659332-13659332 on Build GRCh38
rs771302514 Chr.10: 13659367-13659367 on Build GRCh38
rs144479835 Chr.10: 13659424-13659424 on Build GRCh38
rs145178665 Chr.10: 13659356-13659356 on Build GRCh38
rs375459843 Chr.10: 13659361-13659361 on Build GRCh38
rs373948890 Chr.10: 13659511-13659511 on Build GRCh38
rs933684906 Chr.10: 13659151-13659151 on Build GRCh38
rs774809966 Chr.10: 13659444-13659444 on Build GRCh38
rs1031730680 Chr.10: 13659408-13659408 on Build GRCh38
rs1017648654 Chr.10: 13659160-13659160 on Build GRCh38
rs771517347 Chr.10: 13659290-13659290 on Build GRCh38
rs147549252 Chr.10: 13659400-13659400 on Build GRCh38
rs113485338 Chr.10: 13659451-13659451 on Build GRCh38
rs779625643 Chr.10: 13659346-13659346 on Build GRCh38
rs200519004 Chr.10: 13659447-13659447 on Build GRCh38
rs753680288 Chr.10: 13659412-13659412 on Build GRCh38
rs766341005 Chr.10: 13659462-13659462 on Build GRCh38
rs1007151998 Chr.10: 13659155-13659155 on Build GRCh38
rs368161853 Chr.10: 13659515-13659515 on Build GRCh38
rs139547009 Chr.10: 13659334-13659334 on Build GRCh38
rs755003146 Chr.10: 13659275-13659275 on Build GRCh38
rs779504081 Chr.10: 13659282-13659283 on Build GRCh38
rs761502052 Chr.10: 13659357-13659357 on Build GRCh38
rs776061907 Chr.10: 13659397-13659397 on Build GRCh38
rs964887006 Chr.10: 13659581-13659582 on Build GRCh38
rs151227469 Chr.10: 13659454-13659454 on Build GRCh38
rs1021851212 Chr.10: 13659214-13659214 on Build GRCh38
rs773914910 Chr.10: 13659289-13659289 on Build GRCh38
rs368644769 Chr.10: 13659368-13659368 on Build GRCh38
rs759343693 Chr.10: 13659473-13659473 on Build GRCh38
rs975358127 Chr.10: 13659583-13659583 on Build GRCh38
rs748748895 Chr.10: 13659284-13659284 on Build GRCh38
rs970212368 Chr.10: 13659250-13659250 on Build GRCh38
rs780738869 Chr.10: 13659505-13659505 on Build GRCh38
rs201770153 Chr.10: 13659374-13659374 on Build GRCh38
rs772200364 Chr.10: 13659457-13659457 on Build GRCh38
rs980687525 Chr.10: 13659252-13659252 on Build GRCh38
rs368935593 Chr.10: 13659409-13659409 on Build GRCh38
rs767309844 Chr.10: 13659403-13659403 on Build GRCh38
rs751081943 Chr.10: 13659485-13659485 on Build GRCh38
rs1048315456 Chr.10: 13659326-13659326 on Build GRCh38
rs763056639 Chr.10: 13659311-13659311 on Build GRCh38
rs942650524 Chr.10: 13659533-13659533 on Build GRCh38
rs758051382 Chr.10: 13659482-13659482 on Build GRCh38
rs764268341 Chr.10: 13659313-13659313 on Build GRCh38
rs372861237 Chr.10: 13659350-13659350 on Build GRCh38
rs753586892 Chr.10: 13659469-13659469 on Build GRCh38
rs749710776 Chr.10: 13659448-13659448 on Build GRCh38
rs1040864690 Chr.10: 13659545-13659545 on Build GRCh38
rs768960408 Chr.10: 13659453-13659453 on Build GRCh38
rs555463818 Chr.10: 13659195-13659195 on Build GRCh38
rs9664466 Chr.10: 13659458-13659458 on Build GRCh38
rs550587670 Chr.10: 13659467-13659467 on Build GRCh38
rs889701609 Chr.10: 13659337-13659337 on Build GRCh38
rs61833196 Chr.10: 13659340-13659340 on Build GRCh38
rs560340785 Chr.10: 13659363-13659363 on Build GRCh38
rs777135195 Chr.10: 13659161-13659161 on Build GRCh38
rs188397812 Chr.10: 13659169-13659169 on Build GRCh38
rs757939151 Chr.10: 13659434-13659434 on Build GRCh38
rs746379580 Chr.10: 13659442-13659442 on Build GRCh38
rs922399947 Chr.10: 13659574-13659574 on Build GRCh38
rs144235428 Chr.10: 13659576-13659576 on Build GRCh38
rs368485922 Chr.10: 13659325-13659325 on Build GRCh38
rs755960267 Chr.10: 13659336-13659336 on Build GRCh38
rs765324217 Chr.10: 13659306-13659306 on Build GRCh38
rs756659170 Chr.10: 13659495-13659495 on Build GRCh38
rs749624986 Chr.10: 13659507-13659507 on Build GRCh38
rs540147332 Chr.10: 13659351-13659351 on Build GRCh38
rs768169683 Chr.10: 13659285-13659285 on Build GRCh38
rs373760430 Chr.10: 13659316-13659316 on Build GRCh38
rs761744380 Chr.10: 13659455-13659455 on Build GRCh38
rs938471288 Chr.10: 13659274-13659274 on Build GRCh38
rs748400823 Chr.10: 13659512-13659512 on Build GRCh38
rs750389629 Chr.10: 13659410-13659410 on Build GRCh38
rs776562804 Chr.10: 13659536-13659536 on Build GRCh38
rs779713820 Chr.10: 13659280-13659280 on Build GRCh38
rs776829075 Chr.10: 13659294-13659294 on Build GRCh38
rs140711494 Chr.10: 13659402-13659402 on Build GRCh38
rs531553216 Chr.10: 13659373-13659373 on Build GRCh38
rs770802929 Chr.10: 13659531-13659531 on Build GRCh38
rs1050785976 Chr.10: 13659160-13659161 on Build GRCh38
rs767534496 Chr.10: 13659330-13659330 on Build GRCh38
rs11258505 Chr.10: 13659196-13659196 on Build GRCh38
rs973639888 Chr.10: 13659238-13659249 on Build GRCh38
rs74547111 Chr.10: 13659176-13659176 on Build GRCh38
rs376959985 Chr.10: 13659411-13659411 on Build GRCh38
rs374825261 Chr.10: 13659365-13659365 on Build GRCh38
rs369221320 Chr.10: 13659425-13659425 on Build GRCh38
rs536025126 Chr.10: 13659579-13659579 on Build GRCh38
rs759971697 Chr.10: 13659300-13659300 on Build GRCh38
rs752397976 Chr.10: 13659480-13659480 on Build GRCh38
rs564049987 Chr.10: 13659307-13659307 on Build GRCh38
rs200095341 Chr.10: 13659525-13659525 on Build GRCh38
rs371654662 Chr.10: 13659520-13659520 on Build GRCh38
rs570739185 Chr.10: 13659479-13659479 on Build GRCh38

View Full Product Details