Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6040678-6041270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRN4
Entrez Gene ID
164312
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
C20orf75, NLRR-4, NLRR4, dJ1056H1.1
UniGene
Hs.149133
Gene Chromosome Location
Chr.20: 6040546-6054080 on Build GRCh38
Transcripts
NM_152611.4 XM_011529183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2765879 c.2157G>A p.T719T Chr.20: 6041088-6041088 on Build GRCh38
2765878 c.2198C>T p.P733L Chr.20: 6041047-6041047 on Build GRCh38
7276451 c.2041G>A p.A681T Chr.20: 6041204-6041204 on Build GRCh38
6026566 c.2042C>T p.A681V Chr.20: 6041203-6041203 on Build GRCh38
1327521 c.2082_2083insTTG p.L695_A696insL Chr.20: 6041162-6041163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054056989 Chr.20: 6040773-6040773 on Build GRCh38
rs772315114 Chr.20: 6041074-6041074 on Build GRCh38
rs373031450 Chr.20: 6040928-6040928 on Build GRCh38
rs77819407 Chr.20: 6041122-6041122 on Build GRCh38
rs947755851 Chr.20: 6041159-6041159 on Build GRCh38
rs753390279 Chr.20: 6041167-6041167 on Build GRCh38
rs1012074445 Chr.20: 6040941-6040941 on Build GRCh38
rs766921913 Chr.20: 6041229-6041229 on Build GRCh38
rs748024804 Chr.20: 6041117-6041117 on Build GRCh38
rs778294325 Chr.20: 6040984-6040984 on Build GRCh38
rs1036425562 Chr.20: 6041179-6041179 on Build GRCh38
rs774505683 Chr.20: 6041205-6041205 on Build GRCh38
rs886072893 Chr.20: 6040878-6040878 on Build GRCh38
rs762301269 Chr.20: 6041152-6041152 on Build GRCh38
rs370963693 Chr.20: 6041087-6041087 on Build GRCh38
rs766834346 Chr.20: 6041165-6041165 on Build GRCh38
rs757812622 Chr.20: 6040994-6040994 on Build GRCh38
rs974814694 Chr.20: 6041080-6041080 on Build GRCh38
rs767305580 Chr.20: 6041241-6041247 on Build GRCh38
rs777448616 Chr.20: 6041250-6041250 on Build GRCh38
rs1018970438 Chr.20: 6041063-6041063 on Build GRCh38
rs370695742 Chr.20: 6041001-6041001 on Build GRCh38
rs752197478 Chr.20: 6041236-6041236 on Build GRCh38
rs769170358 Chr.20: 6041002-6041002 on Build GRCh38
rs547175698 Chr.20: 6040748-6040748 on Build GRCh38
rs765545570 Chr.20: 6040975-6040975 on Build GRCh38
rs754330860 Chr.20: 6041230-6041230 on Build GRCh38
rs113657531 Chr.20: 6041037-6041037 on Build GRCh38
rs769136433 Chr.20: 6040831-6040831 on Build GRCh38
rs750887295 Chr.20: 6041098-6041098 on Build GRCh38
rs747751685 Chr.20: 6041177-6041177 on Build GRCh38
rs930540270 Chr.20: 6040839-6040839 on Build GRCh38
rs758004420 Chr.20: 6041187-6041187 on Build GRCh38
rs923577514 Chr.20: 6041005-6041005 on Build GRCh38
rs367956344 Chr.20: 6040733-6040733 on Build GRCh38
rs764735385 Chr.20: 6041235-6041235 on Build GRCh38
rs765228395 Chr.20: 6041064-6041064 on Build GRCh38
rs776625819 Chr.20: 6041022-6041025 on Build GRCh38
rs932124830 Chr.20: 6040823-6040823 on Build GRCh38
rs756574056 Chr.20: 6041099-6041099 on Build GRCh38
rs751922632 Chr.20: 6040993-6040993 on Build GRCh38
rs376035754 Chr.20: 6041109-6041109 on Build GRCh38
rs763288561 Chr.20: 6041075-6041075 on Build GRCh38
rs778086094 Chr.20: 6040874-6040875 on Build GRCh38
rs773482346 Chr.20: 6041221-6041221 on Build GRCh38
rs116777692 Chr.20: 6041057-6041057 on Build GRCh38
rs959311648 Chr.20: 6040939-6040939 on Build GRCh38
rs995361869 Chr.20: 6040919-6040919 on Build GRCh38
rs1036333221 Chr.20: 6040856-6040856 on Build GRCh38
rs974574166 Chr.20: 6040776-6040776 on Build GRCh38
rs367873348 Chr.20: 6041174-6041174 on Build GRCh38
rs979441673 Chr.20: 6040724-6040724 on Build GRCh38
rs756604803 Chr.20: 6041164-6041164 on Build GRCh38
rs759504272 Chr.20: 6040972-6040972 on Build GRCh38
rs572479845 Chr.20: 6040934-6040934 on Build GRCh38
rs988872304 Chr.20: 6040754-6040754 on Build GRCh38
rs559212508 Chr.20: 6041151-6041151 on Build GRCh38
rs114148682 Chr.20: 6040762-6040762 on Build GRCh38
rs752955190 Chr.20: 6040976-6040976 on Build GRCh38
rs765428115 Chr.20: 6041151-6041151 on Build GRCh38
rs754660510 Chr.20: 6041113-6041113 on Build GRCh38
rs761952811 Chr.20: 6041211-6041211 on Build GRCh38
rs77984191 Chr.20: 6041231-6041231 on Build GRCh38
rs565580623 Chr.20: 6040760-6040760 on Build GRCh38
rs759674717 Chr.20: 6041078-6041080 on Build GRCh38
rs781749502 Chr.20: 6041089-6041089 on Build GRCh38
rs368800831 Chr.20: 6040847-6040847 on Build GRCh38
rs754466150 Chr.20: 6041172-6041172 on Build GRCh38
rs761559261 Chr.20: 6041047-6041047 on Build GRCh38
rs536716771 Chr.20: 6041055-6041055 on Build GRCh38
rs114994187 Chr.20: 6041137-6041137 on Build GRCh38
rs748750798 Chr.20: 6041107-6041107 on Build GRCh38
rs1032985188 Chr.20: 6040735-6040735 on Build GRCh38
rs912015125 Chr.20: 6040758-6040758 on Build GRCh38
rs779819405 Chr.20: 6041004-6041004 on Build GRCh38
rs747499668 Chr.20: 6040971-6040972 on Build GRCh38
rs767728329 Chr.20: 6041214-6041214 on Build GRCh38
rs781519437 Chr.20: 6040997-6040997 on Build GRCh38
rs370743397 Chr.20: 6041162-6041162 on Build GRCh38
rs767774106 Chr.20: 6041158-6041158 on Build GRCh38
rs772091784 Chr.20: 6041053-6041053 on Build GRCh38
rs922202647 Chr.20: 6040786-6040786 on Build GRCh38
rs921743162 Chr.20: 6041083-6041083 on Build GRCh38
rs142765128 Chr.20: 6041202-6041202 on Build GRCh38
rs940355166 Chr.20: 6040911-6040911 on Build GRCh38
rs761480246 Chr.20: 6041162-6041163 on Build GRCh38
rs761071116 Chr.20: 6041225-6041225 on Build GRCh38
rs552004462 Chr.20: 6040790-6040790 on Build GRCh38
rs41282952 Chr.20: 6040720-6040720 on Build GRCh38
rs775735336 Chr.20: 6041153-6041154 on Build GRCh38
rs113229963 Chr.20: 6041144-6041144 on Build GRCh38
rs529753609 Chr.20: 6041195-6041195 on Build GRCh38
rs550574687 Chr.20: 6040996-6040997 on Build GRCh38
rs199981644 Chr.20: 6041141-6041141 on Build GRCh38
rs944533255 Chr.20: 6040759-6040759 on Build GRCh38
rs774157248 Chr.20: 6041046-6041046 on Build GRCh38
rs896271397 Chr.20: 6040931-6040931 on Build GRCh38
rs541566350 Chr.20: 6040958-6040959 on Build GRCh38
rs758901189 Chr.20: 6040980-6040980 on Build GRCh38
rs758016285 Chr.20: 6041239-6041239 on Build GRCh38
rs570602300 Chr.20: 6040826-6040826 on Build GRCh38
rs529045077 Chr.20: 6041048-6041048 on Build GRCh38
rs899186785 Chr.20: 6040880-6040880 on Build GRCh38
rs765103253 Chr.20: 6041201-6041201 on Build GRCh38
rs1023215364 Chr.20: 6040704-6040704 on Build GRCh38
rs376508557 Chr.20: 6041076-6041076 on Build GRCh38
rs778509662 Chr.20: 6041114-6041114 on Build GRCh38
rs547838467 Chr.20: 6041198-6041198 on Build GRCh38
rs777444275 Chr.20: 6041192-6041192 on Build GRCh38
rs760763590 Chr.20: 6041123-6041123 on Build GRCh38
rs1025268311 Chr.20: 6040960-6040960 on Build GRCh38
rs763093671 Chr.20: 6041148-6041148 on Build GRCh38
rs774769459 Chr.20: 6041031-6041031 on Build GRCh38

View Full Product Details