Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227686958-227687473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC19A3
Entrez Gene ID
80704
Gene Name
solute carrier family 19 member 3
Gene Aliases
BBGD, THMD2, THTR2
UniGene
Hs.221597
Gene Chromosome Location
Chr.2: 227685210-227718030 on Build GRCh38
Transcripts
NM_025243.3 XM_005246874.3 XM_011511931.2 XM_011511932.1 XM_011511933.1 XM_017005030.1 XM_017005031.1 XM_017005032.1 XM_017005033.1 XM_017005034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1017403 c.1472T>C p.I491T Chr.2: 227687416-227687416 on Build GRCh38
5685327 c.1456A>C p.I486L Chr.2: 227687420-227687420 on Build GRCh38
6052859 c.1444delG p.E482fs*>11 Chr.2: 227687432-227687432 on Build GRCh38
5685326 c.1468A>C p.I490L Chr.2: 227687420-227687420 on Build GRCh38
362100 c.1452A>T p.P484P Chr.2: 227687436-227687436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150113016 Chr.2: 227687070-227687070 on Build GRCh38
rs886055761 Chr.2: 227687221-227687221 on Build GRCh38
rs934479506 Chr.2: 227687165-227687165 on Build GRCh38
rs924470002 Chr.2: 227687145-227687145 on Build GRCh38
rs371251177 Chr.2: 227687372-227687372 on Build GRCh38
rs988934111 Chr.2: 227687179-227687179 on Build GRCh38
rs1037322341 Chr.2: 227687097-227687097 on Build GRCh38
rs975129916 Chr.2: 227687390-227687390 on Build GRCh38
rs532167253 Chr.2: 227687007-227687007 on Build GRCh38
rs930690284 Chr.2: 227687018-227687018 on Build GRCh38
rs991508598 Chr.2: 227687041-227687043 on Build GRCh38
rs754918776 Chr.2: 227687449-227687449 on Build GRCh38
rs745399850 Chr.2: 227687358-227687358 on Build GRCh38
rs567092282 Chr.2: 227687161-227687161 on Build GRCh38
rs116590386 Chr.2: 227687294-227687294 on Build GRCh38
rs114116140 Chr.2: 227687131-227687131 on Build GRCh38
rs756936405 Chr.2: 227687094-227687094 on Build GRCh38
rs776336362 Chr.2: 227687210-227687210 on Build GRCh38
rs761295032 Chr.2: 227687380-227687380 on Build GRCh38
rs755717801 Chr.2: 227687436-227687436 on Build GRCh38
rs915952341 Chr.2: 227687357-227687357 on Build GRCh38
rs1003494762 Chr.2: 227687336-227687336 on Build GRCh38
rs550609221 Chr.2: 227687047-227687047 on Build GRCh38
rs192321676 Chr.2: 227687166-227687166 on Build GRCh38
rs1003396224 Chr.2: 227687169-227687169 on Build GRCh38
rs1051868414 Chr.2: 227687265-227687265 on Build GRCh38
rs890786407 Chr.2: 227687195-227687195 on Build GRCh38
rs779771123 Chr.2: 227687355-227687355 on Build GRCh38
rs148873152 Chr.2: 227687345-227687345 on Build GRCh38
rs940444405 Chr.2: 227687289-227687289 on Build GRCh38
rs750094260 Chr.2: 227687427-227687427 on Build GRCh38
rs886055763 Chr.2: 227687364-227687364 on Build GRCh38
rs886055760 Chr.2: 227687097-227687097 on Build GRCh38
rs761383691 Chr.2: 227687421-227687421 on Build GRCh38
rs749248666 Chr.2: 227687365-227687365 on Build GRCh38
rs192876801 Chr.2: 227687413-227687413 on Build GRCh38
rs898867925 Chr.2: 227687107-227687107 on Build GRCh38
rs188532608 Chr.2: 227687389-227687389 on Build GRCh38
rs745730487 Chr.2: 227687347-227687347 on Build GRCh38
rs1046056350 Chr.2: 227687140-227687140 on Build GRCh38
rs368717276 Chr.2: 227687447-227687447 on Build GRCh38
rs920691214 Chr.2: 227687016-227687016 on Build GRCh38
rs80307329 Chr.2: 227687194-227687194 on Build GRCh38
rs540774490 Chr.2: 227687425-227687425 on Build GRCh38
rs1023981577 Chr.2: 227687160-227687160 on Build GRCh38
rs922195196 Chr.2: 227687220-227687220 on Build GRCh38
rs562213489 Chr.2: 227687335-227687335 on Build GRCh38
rs183992017 Chr.2: 227687331-227687331 on Build GRCh38
rs769503014 Chr.2: 227687128-227687128 on Build GRCh38
rs1044966154 Chr.2: 227687333-227687333 on Build GRCh38
rs138554855 Chr.2: 227687258-227687258 on Build GRCh38
rs199499719 Chr.2: 227687381-227687381 on Build GRCh38
rs914538926 Chr.2: 227687190-227687190 on Build GRCh38
rs779422483 Chr.2: 227687446-227687446 on Build GRCh38
rs748591506 Chr.2: 227687413-227687414 on Build GRCh38
rs1009176543 Chr.2: 227687221-227687221 on Build GRCh38
rs886169023 Chr.2: 227687051-227687051 on Build GRCh38
rs112957132 Chr.2: 227687049-227687049 on Build GRCh38
rs560082192 Chr.2: 227687193-227687193 on Build GRCh38
rs763595744 Chr.2: 227687395-227687395 on Build GRCh38
rs886055762 Chr.2: 227687291-227687291 on Build GRCh38
rs377323228 Chr.2: 227687392-227687392 on Build GRCh38
rs863224205 Chr.2: 227687410-227687410 on Build GRCh38
rs892892723 Chr.2: 227687171-227687171 on Build GRCh38
rs776907842 Chr.2: 227687384-227687384 on Build GRCh38
rs549546166 Chr.2: 227687282-227687282 on Build GRCh38
rs764642918 Chr.2: 227687412-227687412 on Build GRCh38
rs956100048 Chr.2: 227687177-227687177 on Build GRCh38
rs898798043 Chr.2: 227687319-227687319 on Build GRCh38
rs933496714 Chr.2: 227686987-227686987 on Build GRCh38
rs755910828 Chr.2: 227687352-227687352 on Build GRCh38
rs991975369 Chr.2: 227687338-227687338 on Build GRCh38
rs570743421 Chr.2: 227687226-227687226 on Build GRCh38
rs1030864059 Chr.2: 227687174-227687174 on Build GRCh38

View Full Product Details