Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157414357-157414956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTIP
Entrez Gene ID
9595
Gene Name
cytohesin 1 interacting protein
Gene Aliases
B3-1, CASP, CYBR, CYTHIP, HE, PSCDBP
UniGene
Hs.270
Gene Chromosome Location
Chr.2: 157412781-157444092 on Build GRCh38
Transcripts
NM_004288.4 XM_017005386.1

SNP Detail

SNP ID Chromosome Location
rs916030168 Chr.2: 157414630-157414630 on Build GRCh38
rs908558385 Chr.2: 157414685-157414685 on Build GRCh38
rs770736365 Chr.2: 157414504-157414504 on Build GRCh38
rs903650689 Chr.2: 157414635-157414635 on Build GRCh38
rs138526669 Chr.2: 157414838-157414838 on Build GRCh38
rs1038597841 Chr.2: 157414699-157414699 on Build GRCh38
rs980264617 Chr.2: 157414654-157414654 on Build GRCh38
rs532567411 Chr.2: 157414917-157414917 on Build GRCh38
rs954266934 Chr.2: 157414585-157414586 on Build GRCh38
rs1032966007 Chr.2: 157414919-157414919 on Build GRCh38
rs1033411135 Chr.2: 157414598-157414599 on Build GRCh38
rs547628755 Chr.2: 157414931-157414931 on Build GRCh38
rs754267106 Chr.2: 157414565-157414566 on Build GRCh38
rs1000670294 Chr.2: 157414861-157414861 on Build GRCh38
rs1033072888 Chr.2: 157414903-157414903 on Build GRCh38
rs967725167 Chr.2: 157414553-157414553 on Build GRCh38
rs374489533 Chr.2: 157414784-157414784 on Build GRCh38
rs16841706 Chr.2: 157414727-157414727 on Build GRCh38
rs776655568 Chr.2: 157414530-157414530 on Build GRCh38
rs971491979 Chr.2: 157414633-157414633 on Build GRCh38
rs181761581 Chr.2: 157414771-157414771 on Build GRCh38
rs375611688 Chr.2: 157414719-157414719 on Build GRCh38
rs545472440 Chr.2: 157414421-157414421 on Build GRCh38
rs997942841 Chr.2: 157414484-157414484 on Build GRCh38
rs375394566 Chr.2: 157414582-157414585 on Build GRCh38
rs956248410 Chr.2: 157414904-157414904 on Build GRCh38
rs753156694 Chr.2: 157414752-157414752 on Build GRCh38
rs889904334 Chr.2: 157414859-157414859 on Build GRCh38
rs869275588 Chr.2: 157414605-157414616 on Build GRCh38
rs198688 Chr.2: 157414448-157414448 on Build GRCh38
rs561361117 Chr.2: 157414755-157414755 on Build GRCh38
rs565636252 Chr.2: 157414509-157414509 on Build GRCh38
rs756550961 Chr.2: 157414563-157414564 on Build GRCh38
rs1036920046 Chr.2: 157414796-157414796 on Build GRCh38
rs957193636 Chr.2: 157414610-157414610 on Build GRCh38
rs998481039 Chr.2: 157414918-157414918 on Build GRCh38
rs562510811 Chr.2: 157414389-157414389 on Build GRCh38
rs527827583 Chr.2: 157414697-157414697 on Build GRCh38
rs996936850 Chr.2: 157414447-157414447 on Build GRCh38
rs939988596 Chr.2: 157414694-157414694 on Build GRCh38
rs989081333 Chr.2: 157414615-157414615 on Build GRCh38
rs1009015050 Chr.2: 157414507-157414507 on Build GRCh38
rs770529027 Chr.2: 157414592-157414593 on Build GRCh38
rs926395372 Chr.2: 157414710-157414710 on Build GRCh38
rs888864505 Chr.2: 157414482-157414482 on Build GRCh38
rs5835666 Chr.2: 157414581-157414582 on Build GRCh38
rs971563241 Chr.2: 157414632-157414632 on Build GRCh38
rs776157434 Chr.2: 157414435-157414435 on Build GRCh38
rs200528113 Chr.2: 157414581-157414581 on Build GRCh38
rs920138969 Chr.2: 157414709-157414709 on Build GRCh38
rs1000233226 Chr.2: 157414589-157414590 on Build GRCh38
rs981872946 Chr.2: 157414664-157414664 on Build GRCh38
rs769958049 Chr.2: 157414564-157414565 on Build GRCh38
rs199685650 Chr.2: 157414578-157414578 on Build GRCh38
rs998794680 Chr.2: 157414558-157414558 on Build GRCh38
rs577777472 Chr.2: 157414414-157414414 on Build GRCh38
rs367701564 Chr.2: 157414582-157414589 on Build GRCh38
rs3072204 Chr.2: 157414606-157414607 on Build GRCh38
rs186878588 Chr.2: 157414819-157414819 on Build GRCh38
rs1004520815 Chr.2: 157414401-157414401 on Build GRCh38
rs898680782 Chr.2: 157414418-157414418 on Build GRCh38
rs1038566249 Chr.2: 157414412-157414412 on Build GRCh38
rs1049541722 Chr.2: 157414856-157414856 on Build GRCh38
rs397952418 Chr.2: 157414616-157414617 on Build GRCh38
rs901056392 Chr.2: 157414829-157414829 on Build GRCh38
rs1028365824 Chr.2: 157414577-157414577 on Build GRCh38
rs763503755 Chr.2: 157414833-157414833 on Build GRCh38
rs991316640 Chr.2: 157414614-157414614 on Build GRCh38
rs866786619 Chr.2: 157414580-157414580 on Build GRCh38

View Full Product Details