Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224400963-224401498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124B
Entrez Gene ID
79843
Gene Name
family with sequence similarity 124 member B
Gene Aliases
-
UniGene
Hs.147585
Gene Chromosome Location
Chr.2: 224378698-224401994 on Build GRCh38
Transcripts
NM_001122779.1 NM_024785.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442395 c.338A>G p.N113S Chr.2: 224401431-224401431 on Build GRCh38
224218 c.487G>A p.E163K Chr.2: 224401282-224401282 on Build GRCh38
3578268 c.404C>T p.S135F Chr.2: 224401365-224401365 on Build GRCh38
6658308 c.388C>T p.Q130* Chr.2: 224401381-224401381 on Build GRCh38
6034883 c.713G>A p.G238D Chr.2: 224401056-224401056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371374350 Chr.2: 224401161-224401161 on Build GRCh38
rs752187289 Chr.2: 224401329-224401329 on Build GRCh38
rs761291964 Chr.2: 224401308-224401309 on Build GRCh38
rs781535619 Chr.2: 224401000-224401000 on Build GRCh38
rs776107943 Chr.2: 224401475-224401475 on Build GRCh38
rs779112615 Chr.2: 224401036-224401036 on Build GRCh38
rs375430957 Chr.2: 224401066-224401066 on Build GRCh38
rs982165858 Chr.2: 224401174-224401174 on Build GRCh38
rs571017410 Chr.2: 224401192-224401192 on Build GRCh38
rs754512892 Chr.2: 224401211-224401211 on Build GRCh38
rs369601310 Chr.2: 224401388-224401388 on Build GRCh38
rs779739106 Chr.2: 224401088-224401088 on Build GRCh38
rs144448068 Chr.2: 224401364-224401364 on Build GRCh38
rs150631626 Chr.2: 224401436-224401436 on Build GRCh38
rs754381768 Chr.2: 224401207-224401207 on Build GRCh38
rs562831899 Chr.2: 224401383-224401383 on Build GRCh38
rs147930186 Chr.2: 224401176-224401176 on Build GRCh38
rs527479715 Chr.2: 224401426-224401426 on Build GRCh38
rs369547761 Chr.2: 224401239-224401239 on Build GRCh38
rs1055450886 Chr.2: 224401374-224401382 on Build GRCh38
rs764920776 Chr.2: 224401411-224401412 on Build GRCh38
rs201169903 Chr.2: 224401004-224401004 on Build GRCh38
rs745606630 Chr.2: 224401435-224401435 on Build GRCh38
rs769064165 Chr.2: 224401322-224401323 on Build GRCh38
rs368644434 Chr.2: 224401001-224401001 on Build GRCh38
rs367619277 Chr.2: 224401138-224401138 on Build GRCh38
rs761176688 Chr.2: 224401305-224401305 on Build GRCh38
rs569274440 Chr.2: 224401456-224401456 on Build GRCh38
rs886959439 Chr.2: 224401061-224401061 on Build GRCh38
rs780648923 Chr.2: 224401193-224401193 on Build GRCh38
rs1013250227 Chr.2: 224401449-224401449 on Build GRCh38
rs1039772945 Chr.2: 224401006-224401006 on Build GRCh38
rs765544185 Chr.2: 224401029-224401029 on Build GRCh38
rs746173534 Chr.2: 224401042-224401043 on Build GRCh38
rs764234329 Chr.2: 224401160-224401160 on Build GRCh38
rs529893167 Chr.2: 224401441-224401441 on Build GRCh38
rs746662202 Chr.2: 224401361-224401361 on Build GRCh38
rs768435478 Chr.2: 224401107-224401107 on Build GRCh38
rs769727078 Chr.2: 224401387-224401387 on Build GRCh38
rs771811630 Chr.2: 224401132-224401132 on Build GRCh38
rs757996538 Chr.2: 224401424-224401424 on Build GRCh38
rs755464550 Chr.2: 224401316-224401316 on Build GRCh38
rs756858453 Chr.2: 224401431-224401431 on Build GRCh38
rs773089408 Chr.2: 224401017-224401017 on Build GRCh38
rs200113424 Chr.2: 224401217-224401217 on Build GRCh38
rs1001233131 Chr.2: 224401341-224401342 on Build GRCh38
rs142573389 Chr.2: 224401310-224401310 on Build GRCh38
rs761233139 Chr.2: 224401410-224401411 on Build GRCh38
rs201191737 Chr.2: 224401422-224401422 on Build GRCh38
rs760006141 Chr.2: 224401469-224401469 on Build GRCh38
rs1022471227 Chr.2: 224401386-224401386 on Build GRCh38
rs201987132 Chr.2: 224401133-224401133 on Build GRCh38
rs775972675 Chr.2: 224401144-224401144 on Build GRCh38
rs1052712561 Chr.2: 224400995-224400995 on Build GRCh38
rs751266993 Chr.2: 224401430-224401430 on Build GRCh38
rs758625841 Chr.2: 224401043-224401043 on Build GRCh38
rs956920973 Chr.2: 224401242-224401242 on Build GRCh38
rs150435753 Chr.2: 224401323-224401323 on Build GRCh38
rs778455233 Chr.2: 224401213-224401213 on Build GRCh38
rs746749968 Chr.2: 224401231-224401231 on Build GRCh38
rs750775324 Chr.2: 224401173-224401173 on Build GRCh38
rs756690006 Chr.2: 224401190-224401190 on Build GRCh38
rs199522881 Chr.2: 224401098-224401098 on Build GRCh38
rs773052322 Chr.2: 224401424-224401426 on Build GRCh38
rs200327175 Chr.2: 224401432-224401432 on Build GRCh38
rs766918271 Chr.2: 224401246-224401246 on Build GRCh38
rs776874463 Chr.2: 224401468-224401468 on Build GRCh38
rs151315340 Chr.2: 224401169-224401169 on Build GRCh38
rs372107073 Chr.2: 224401010-224401010 on Build GRCh38
rs747779894 Chr.2: 224401214-224401214 on Build GRCh38
rs752735728 Chr.2: 224401033-224401033 on Build GRCh38
rs535835244 Chr.2: 224401045-224401045 on Build GRCh38
rs758001941 Chr.2: 224401339-224401339 on Build GRCh38
rs768671588 Chr.2: 224401283-224401283 on Build GRCh38
rs377743341 Chr.2: 224401462-224401462 on Build GRCh38
rs138469734 Chr.2: 224401172-224401172 on Build GRCh38
rs545145666 Chr.2: 224401394-224401394 on Build GRCh38
rs565742600 Chr.2: 224401286-224401286 on Build GRCh38
rs147767140 Chr.2: 224400999-224400999 on Build GRCh38
rs777097507 Chr.2: 224401411-224401411 on Build GRCh38
rs780946946 Chr.2: 224401377-224401377 on Build GRCh38
rs777463842 Chr.2: 224401346-224401346 on Build GRCh38
rs747832071 Chr.2: 224401117-224401117 on Build GRCh38
rs200319143 Chr.2: 224401019-224401019 on Build GRCh38
rs754436618 Chr.2: 224401313-224401313 on Build GRCh38
rs985072268 Chr.2: 224401476-224401476 on Build GRCh38
rs770789110 Chr.2: 224401139-224401139 on Build GRCh38
rs772484575 Chr.2: 224401131-224401131 on Build GRCh38
rs550079054 Chr.2: 224401402-224401402 on Build GRCh38
rs763081772 Chr.2: 224401148-224401148 on Build GRCh38
rs146039940 Chr.2: 224401392-224401392 on Build GRCh38
rs142663942 Chr.2: 224401011-224401011 on Build GRCh38
rs765719766 Chr.2: 224401421-224401421 on Build GRCh38
rs747266107 Chr.2: 224401458-224401458 on Build GRCh38
rs748198485 Chr.2: 224401390-224401390 on Build GRCh38
rs928020765 Chr.2: 224401179-224401179 on Build GRCh38
rs370141388 Chr.2: 224401218-224401218 on Build GRCh38
rs1050792964 Chr.2: 224401056-224401056 on Build GRCh38
rs375219740 Chr.2: 224401237-224401237 on Build GRCh38
rs769909652 Chr.2: 224401195-224401195 on Build GRCh38
rs894034647 Chr.2: 224400998-224400998 on Build GRCh38
rs547520726 Chr.2: 224401036-224401036 on Build GRCh38
rs776166992 Chr.2: 224401343-224401343 on Build GRCh38
rs760584262 Chr.2: 224401024-224401024 on Build GRCh38
rs933621297 Chr.2: 224401391-224401391 on Build GRCh38
rs775563751 Chr.2: 224401141-224401141 on Build GRCh38
rs750762038 Chr.2: 224401050-224401050 on Build GRCh38
rs149759174 Chr.2: 224401446-224401446 on Build GRCh38
rs748339760 Chr.2: 224401110-224401110 on Build GRCh38
rs373140603 Chr.2: 224401300-224401300 on Build GRCh38
rs765323647 Chr.2: 224401025-224401025 on Build GRCh38
rs376596730 Chr.2: 224401303-224401303 on Build GRCh38
rs780683002 Chr.2: 224401397-224401397 on Build GRCh38
rs749176681 Chr.2: 224401442-224401442 on Build GRCh38
rs200639231 Chr.2: 224401164-224401164 on Build GRCh38
rs370165264 Chr.2: 224401470-224401470 on Build GRCh38
rs767708285 Chr.2: 224401301-224401301 on Build GRCh38
rs549413818 Chr.2: 224401046-224401046 on Build GRCh38
rs139905737 Chr.2: 224401417-224401417 on Build GRCh38
rs13010549 Chr.2: 224401055-224401055 on Build GRCh38

View Full Product Details