Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 57205093-57205657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC5
Entrez Gene ID
10640
Gene Name
exocyst complex component 5
Gene Aliases
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P
UniGene
Hs.743987
Gene Chromosome Location
Chr.14: 57202476-57268899 on Build GRCh38
Transcripts
NM_006544.3 XM_005267272.3

SNP Detail

SNP ID Chromosome Location
rs553298995 Chr.14: 57205264-57205264 on Build GRCh38
rs902363238 Chr.14: 57205510-57205510 on Build GRCh38
rs1031732038 Chr.14: 57205268-57205268 on Build GRCh38
rs976054285 Chr.14: 57205370-57205370 on Build GRCh38
rs768463514 Chr.14: 57205159-57205159 on Build GRCh38
rs954334659 Chr.14: 57205404-57205404 on Build GRCh38
rs867731374 Chr.14: 57205234-57205234 on Build GRCh38
rs1012073835 Chr.14: 57205574-57205574 on Build GRCh38
rs568728430 Chr.14: 57205265-57205265 on Build GRCh38
rs74856420 Chr.14: 57205162-57205162 on Build GRCh38
rs972915605 Chr.14: 57205438-57205438 on Build GRCh38
rs773160531 Chr.14: 57205535-57205535 on Build GRCh38
rs575267898 Chr.14: 57205415-57205415 on Build GRCh38
rs529139903 Chr.14: 57205612-57205612 on Build GRCh38
rs772692300 Chr.14: 57205204-57205204 on Build GRCh38
rs919714522 Chr.14: 57205450-57205450 on Build GRCh38
rs1013504856 Chr.14: 57205205-57205205 on Build GRCh38
rs188950785 Chr.14: 57205157-57205157 on Build GRCh38
rs548848307 Chr.14: 57205173-57205173 on Build GRCh38
rs561842328 Chr.14: 57205547-57205547 on Build GRCh38
rs1028284193 Chr.14: 57205581-57205581 on Build GRCh38
rs778612812 Chr.14: 57205361-57205361 on Build GRCh38
rs186299949 Chr.14: 57205536-57205536 on Build GRCh38
rs574228768 Chr.14: 57205558-57205558 on Build GRCh38
rs954014005 Chr.14: 57205634-57205634 on Build GRCh38
rs557863048 Chr.14: 57205475-57205475 on Build GRCh38
rs535117397 Chr.14: 57205266-57205266 on Build GRCh38
rs898171284 Chr.14: 57205153-57205153 on Build GRCh38
rs986976315 Chr.14: 57205409-57205409 on Build GRCh38
rs1046317628 Chr.14: 57205504-57205504 on Build GRCh38
rs769757135 Chr.14: 57205478-57205478 on Build GRCh38
rs961541356 Chr.14: 57205422-57205422 on Build GRCh38
rs1024815483 Chr.14: 57205229-57205229 on Build GRCh38
rs925101064 Chr.14: 57205426-57205426 on Build GRCh38
rs916208489 Chr.14: 57205484-57205484 on Build GRCh38
rs1036750545 Chr.14: 57205149-57205149 on Build GRCh38
rs977991370 Chr.14: 57205416-57205416 on Build GRCh38
rs999358399 Chr.14: 57205259-57205259 on Build GRCh38
rs568646544 Chr.14: 57205174-57205174 on Build GRCh38
rs534868373 Chr.14: 57205253-57205253 on Build GRCh38
rs999389803 Chr.14: 57205537-57205537 on Build GRCh38
rs1028274314 Chr.14: 57205382-57205382 on Build GRCh38
rs964727470 Chr.14: 57205333-57205333 on Build GRCh38
rs990195695 Chr.14: 57205377-57205377 on Build GRCh38
rs181642324 Chr.14: 57205471-57205471 on Build GRCh38
rs200885116 Chr.14: 57205304-57205304 on Build GRCh38
rs985432088 Chr.14: 57205290-57205290 on Build GRCh38
rs8003535 Chr.14: 57205286-57205286 on Build GRCh38
rs573120105 Chr.14: 57205487-57205487 on Build GRCh38
rs893255531 Chr.14: 57205564-57205564 on Build GRCh38
rs894802719 Chr.14: 57205141-57205141 on Build GRCh38
rs764421216 Chr.14: 57205399-57205399 on Build GRCh38

View Full Product Details