Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102209458-102210005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR20
Entrez Gene ID
91833
Gene Name
WD repeat domain 20
Gene Aliases
DMR
UniGene
Hs.36859
Gene Chromosome Location
Chr.14: 102139518-102224686 on Build GRCh38
Transcripts
NM_001242414.1 NM_001242415.1 NM_001242416.1 NM_001242417.1 NM_001242418.1 NM_001320130.1 NM_144574.3 NM_181291.2 NM_181308.2 NR_038354.1 XM_006720308.1 XM_006720309.1 XM_006720310.1 XM_006720311.1 XM_006720313.2 XM_006720318.2 XM_006720320.3 XM_011537335.2 XM_011537336.2 XM_011537337.2 XM_011537338.1 XM_011537339.2 XM_011537340.2 XM_011537341.2 XM_011537343.1 XM_011537344.2 XM_011537345.1 XM_011537346.1 XM_011537347.1 XM_011537352.2 XM_011537353.2 XM_011537355.2 XM_011537356.2 XM_011537358.2 XM_017021766.1 XM_017021767.1 XM_017021768.1 XM_017021770.1 XM_017021771.1 XM_017021772.1 XM_017021773.1 XM_017021774.1 XM_017021775.1 XM_017021776.1 XM_017021777.1 XM_017021778.1 XM_017021779.1 XM_017021780.1 XM_017021781.1 XM_017021782.1 XM_017021783.1 XM_017021784.1 XM_017021785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568347 c.1427G>A p.S476N Chr.14: 102209504-102209504 on Build GRCh38
5568348 c.1334G>A p.S445N Chr.14: 102209504-102209504 on Build GRCh38
5479408 c.1507_1508AC>CG p.T503R Chr.14: 102209677-102209678 on Build GRCh38
5479407 c.1600_1601AC>CG p.T534R Chr.14: 102209677-102209678 on Build GRCh38
6869450 c.1576C>T p.L526L Chr.14: 102209653-102209653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767636814 Chr.14: 102209535-102209535 on Build GRCh38
rs777394061 Chr.14: 102209707-102209707 on Build GRCh38
rs750445266 Chr.14: 102209681-102209681 on Build GRCh38
rs140740176 Chr.14: 102209673-102209673 on Build GRCh38
rs758986382 Chr.14: 102209908-102209908 on Build GRCh38
rs373998004 Chr.14: 102209799-102209799 on Build GRCh38
rs199695106 Chr.14: 102209685-102209685 on Build GRCh38
rs111594639 Chr.14: 102209787-102209787 on Build GRCh38
rs766605234 Chr.14: 102209845-102209845 on Build GRCh38
rs772866138 Chr.14: 102209488-102209488 on Build GRCh38
rs1004017868 Chr.14: 102209916-102209916 on Build GRCh38
rs528060594 Chr.14: 102209920-102209920 on Build GRCh38
rs200502921 Chr.14: 102209618-102209618 on Build GRCh38
rs755507329 Chr.14: 102209600-102209600 on Build GRCh38
rs867562590 Chr.14: 102209527-102209527 on Build GRCh38
rs754220572 Chr.14: 102209694-102209694 on Build GRCh38
rs956851044 Chr.14: 102209930-102209930 on Build GRCh38
rs540991291 Chr.14: 102209642-102209642 on Build GRCh38
rs760995416 Chr.14: 102209683-102209683 on Build GRCh38
rs746069401 Chr.14: 102209792-102209792 on Build GRCh38
rs1005516354 Chr.14: 102209865-102209865 on Build GRCh38
rs879198791 Chr.14: 102209944-102209944 on Build GRCh38
rs747675612 Chr.14: 102209631-102209631 on Build GRCh38
rs768444741 Chr.14: 102209804-102209804 on Build GRCh38
rs780513433 Chr.14: 102209574-102209574 on Build GRCh38
rs879079286 Chr.14: 102209979-102209979 on Build GRCh38
rs758819721 Chr.14: 102209706-102209706 on Build GRCh38
rs141952409 Chr.14: 102209701-102209701 on Build GRCh38
rs1046961147 Chr.14: 102209752-102209752 on Build GRCh38
rs976898510 Chr.14: 102209974-102209974 on Build GRCh38
rs1018338839 Chr.14: 102209518-102209518 on Build GRCh38
rs779399464 Chr.14: 102209702-102209702 on Build GRCh38
rs1022516182 Chr.14: 102209937-102209937 on Build GRCh38
rs12888595 Chr.14: 102209500-102209500 on Build GRCh38
rs3742432 Chr.14: 102209929-102209929 on Build GRCh38
rs201001287 Chr.14: 102209717-102209717 on Build GRCh38
rs760395489 Chr.14: 102209498-102209498 on Build GRCh38
rs770648895 Chr.14: 102209726-102209726 on Build GRCh38
rs753997761 Chr.14: 102209852-102209852 on Build GRCh38
rs1046043858 Chr.14: 102209802-102209802 on Build GRCh38
rs745572420 Chr.14: 102209932-102209932 on Build GRCh38
rs144508370 Chr.14: 102209703-102209703 on Build GRCh38
rs553080667 Chr.14: 102209542-102209542 on Build GRCh38
rs900997681 Chr.14: 102209709-102209709 on Build GRCh38
rs542666708 Chr.14: 102209733-102209733 on Build GRCh38
rs200833496 Chr.14: 102209485-102209485 on Build GRCh38
rs976739709 Chr.14: 102209548-102209548 on Build GRCh38
rs771546202 Chr.14: 102209635-102209635 on Build GRCh38
rs58057317 Chr.14: 102209632-102209632 on Build GRCh38
rs774892208 Chr.14: 102209678-102209678 on Build GRCh38
rs747620730 Chr.14: 102209480-102209480 on Build GRCh38
rs148159105 Chr.14: 102209679-102209679 on Build GRCh38
rs751104081 Chr.14: 102209919-102209919 on Build GRCh38
rs765237151 Chr.14: 102209519-102209519 on Build GRCh38
rs769777369 Chr.14: 102209781-102209781 on Build GRCh38
rs199946793 Chr.14: 102209504-102209504 on Build GRCh38
rs45622942 Chr.14: 102209902-102209902 on Build GRCh38
rs910900527 Chr.14: 102209970-102209970 on Build GRCh38
rs367575817 Chr.14: 102209844-102209844 on Build GRCh38
rs376704374 Chr.14: 102209891-102209891 on Build GRCh38
rs1018890476 Chr.14: 102209963-102209963 on Build GRCh38
rs370645511 Chr.14: 102209663-102209663 on Build GRCh38
rs759752397 Chr.14: 102209853-102209853 on Build GRCh38
rs773368396 Chr.14: 102209839-102209839 on Build GRCh38
rs776608929 Chr.14: 102209655-102209655 on Build GRCh38
rs770470320 Chr.14: 102209654-102209654 on Build GRCh38
rs749617951 Chr.14: 102209575-102209575 on Build GRCh38
rs756273953 Chr.14: 102209545-102209545 on Build GRCh38
rs746069335 Chr.14: 102209950-102209950 on Build GRCh38
rs759366277 Chr.14: 102209658-102209658 on Build GRCh38
rs1024069329 Chr.14: 102209871-102209871 on Build GRCh38
rs368374786 Chr.14: 102209592-102209592 on Build GRCh38
rs112900408 Chr.14: 102209745-102209745 on Build GRCh38
rs956882273 Chr.14: 102209524-102209524 on Build GRCh38
rs942406889 Chr.14: 102209972-102209972 on Build GRCh38
rs759595649 Chr.14: 102209514-102209514 on Build GRCh38
rs374240865 Chr.14: 102209530-102209530 on Build GRCh38
rs915531099 Chr.14: 102209607-102209607 on Build GRCh38
rs200078288 Chr.14: 102209523-102209523 on Build GRCh38
rs778103159 Chr.14: 102209909-102209909 on Build GRCh38
rs978423902 Chr.14: 102209956-102209956 on Build GRCh38
rs557598274 Chr.14: 102209645-102209645 on Build GRCh38

View Full Product Details