Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78437883-78438447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IREB2
Entrez Gene ID
3658
Gene Name
iron responsive element binding protein 2
Gene Aliases
ACO3, IRP2, IRP2AD
UniGene
Hs.436031
Gene Chromosome Location
Chr.15: 78438176-78501456 on Build GRCh38
Transcripts
NM_001320941.1 NM_001320942.1 NM_001320943.1 NM_004136.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077980 c.10C>A p.P4T Chr.15: 78438347-78438347 on Build GRCh38
3969287 c.13A>G p.K5E Chr.15: 78438350-78438350 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904142758 Chr.15: 78438175-78438175 on Build GRCh38
rs560628141 Chr.15: 78438124-78438125 on Build GRCh38
rs889399787 Chr.15: 78438210-78438210 on Build GRCh38
rs1016022320 Chr.15: 78438344-78438344 on Build GRCh38
rs566288964 Chr.15: 78438060-78438060 on Build GRCh38
rs541886386 Chr.15: 78438263-78438263 on Build GRCh38
rs189535848 Chr.15: 78438330-78438330 on Build GRCh38
rs536718193 Chr.15: 78438082-78438082 on Build GRCh38
rs533378408 Chr.15: 78438413-78438413 on Build GRCh38
rs1006139458 Chr.15: 78438384-78438384 on Build GRCh38
rs984684218 Chr.15: 78437922-78437922 on Build GRCh38
rs530450756 Chr.15: 78437918-78437918 on Build GRCh38
rs935548540 Chr.15: 78438266-78438266 on Build GRCh38
rs948035734 Chr.15: 78438059-78438059 on Build GRCh38
rs565461124 Chr.15: 78438111-78438111 on Build GRCh38
rs961680450 Chr.15: 78438089-78438089 on Build GRCh38
rs768385913 Chr.15: 78438383-78438383 on Build GRCh38
rs984653173 Chr.15: 78438081-78438081 on Build GRCh38
rs1028795133 Chr.15: 78438071-78438071 on Build GRCh38
rs972064192 Chr.15: 78438418-78438418 on Build GRCh38
rs896259926 Chr.15: 78438305-78438305 on Build GRCh38
rs1029242788 Chr.15: 78437979-78437979 on Build GRCh38
rs997594275 Chr.15: 78438208-78438208 on Build GRCh38
rs999225027 Chr.15: 78438302-78438302 on Build GRCh38
rs745766339 Chr.15: 78438294-78438294 on Build GRCh38
rs2656070 Chr.15: 78437910-78437910 on Build GRCh38
rs572012121 Chr.15: 78437961-78437961 on Build GRCh38
rs960574370 Chr.15: 78438033-78438033 on Build GRCh38
rs1006269804 Chr.15: 78438307-78438307 on Build GRCh38
rs773625494 Chr.15: 78438314-78438314 on Build GRCh38
rs534611251 Chr.15: 78438227-78438227 on Build GRCh38
rs779130497 Chr.15: 78438398-78438398 on Build GRCh38
rs993408886 Chr.15: 78438370-78438370 on Build GRCh38
rs773390943 Chr.15: 78438408-78438408 on Build GRCh38
rs1023953697 Chr.15: 78438242-78438242 on Build GRCh38
rs373859045 Chr.15: 78438332-78438332 on Build GRCh38
rs909221725 Chr.15: 78438246-78438246 on Build GRCh38
rs570013420 Chr.15: 78438163-78438163 on Build GRCh38
rs758117313 Chr.15: 78438287-78438287 on Build GRCh38
rs909730113 Chr.15: 78438279-78438301 on Build GRCh38
rs779854722 Chr.15: 78438379-78438379 on Build GRCh38
rs112848882 Chr.15: 78438184-78438184 on Build GRCh38
rs992073361 Chr.15: 78438035-78438035 on Build GRCh38
rs202064813 Chr.15: 78438313-78438313 on Build GRCh38
rs768936705 Chr.15: 78438304-78438304 on Build GRCh38
rs895323115 Chr.15: 78438003-78438003 on Build GRCh38
rs962007312 Chr.15: 78438412-78438412 on Build GRCh38
rs969349733 Chr.15: 78438244-78438244 on Build GRCh38
rs552714324 Chr.15: 78438234-78438234 on Build GRCh38
rs903572303 Chr.15: 78438216-78438216 on Build GRCh38
rs948230010 Chr.15: 78438136-78438136 on Build GRCh38
rs748331818 Chr.15: 78438400-78438400 on Build GRCh38
rs975978254 Chr.15: 78437902-78437902 on Build GRCh38
rs774492226 Chr.15: 78438306-78438306 on Build GRCh38
rs769503701 Chr.15: 78438296-78438296 on Build GRCh38
rs916735715 Chr.15: 78438130-78438130 on Build GRCh38
rs1004988254 Chr.15: 78437952-78437952 on Build GRCh38
rs151337283 Chr.15: 78438217-78438217 on Build GRCh38
rs999780389 Chr.15: 78438179-78438179 on Build GRCh38
rs889340331 Chr.15: 78438276-78438276 on Build GRCh38
rs985165385 Chr.15: 78438245-78438245 on Build GRCh38
rs954144 Chr.15: 78438249-78438249 on Build GRCh38
rs923337467 Chr.15: 78438424-78438424 on Build GRCh38
rs780806654 Chr.15: 78438364-78438364 on Build GRCh38
rs1006945799 Chr.15: 78438213-78438213 on Build GRCh38
rs953361173 Chr.15: 78438074-78438074 on Build GRCh38
rs972118233 Chr.15: 78438256-78438256 on Build GRCh38
rs901861102 Chr.15: 78438198-78438198 on Build GRCh38
rs1036452943 Chr.15: 78438124-78438124 on Build GRCh38
rs971671456 Chr.15: 78438091-78438091 on Build GRCh38
rs901832590 Chr.15: 78438274-78438274 on Build GRCh38
rs938415992 Chr.15: 78438116-78438119 on Build GRCh38
rs897681685 Chr.15: 78438350-78438350 on Build GRCh38
rs182504945 Chr.15: 78438264-78438264 on Build GRCh38
rs969673760 Chr.15: 78438353-78438353 on Build GRCh38
rs947670826 Chr.15: 78438278-78438278 on Build GRCh38
rs759814732 Chr.15: 78438333-78438333 on Build GRCh38
rs984831326 Chr.15: 78438084-78438084 on Build GRCh38
rs1030847662 Chr.15: 78438027-78438027 on Build GRCh38
rs903519753 Chr.15: 78438284-78438284 on Build GRCh38
rs756759296 Chr.15: 78438288-78438288 on Build GRCh38
rs756122305 Chr.15: 78438377-78438377 on Build GRCh38
rs1022447161 Chr.15: 78438002-78438002 on Build GRCh38
rs559871633 Chr.15: 78438406-78438406 on Build GRCh38
rs1051105969 Chr.15: 78438190-78438190 on Build GRCh38
rs1030795886 Chr.15: 78438219-78438219 on Build GRCh38
rs902020207 Chr.15: 78438149-78438149 on Build GRCh38
rs766775395 Chr.15: 78438316-78438316 on Build GRCh38
rs535922618 Chr.15: 78438145-78438145 on Build GRCh38
rs546757706 Chr.15: 78437975-78437975 on Build GRCh38
rs781285310 Chr.15: 78438293-78438293 on Build GRCh38
rs754583759 Chr.15: 78438303-78438303 on Build GRCh38
rs991601164 Chr.15: 78438404-78438404 on Build GRCh38
rs770119221 Chr.15: 78437957-78437957 on Build GRCh38
rs953837635 Chr.15: 78437985-78437985 on Build GRCh38
rs1031086316 Chr.15: 78438036-78438036 on Build GRCh38
rs1006050421 Chr.15: 78438220-78438220 on Build GRCh38
rs796111080 Chr.15: 78438262-78438262 on Build GRCh38
rs887896745 Chr.15: 78437943-78437943 on Build GRCh38
rs940790810 Chr.15: 78438123-78438123 on Build GRCh38
rs1050531351 Chr.15: 78438281-78438281 on Build GRCh38
rs961901511 Chr.15: 78438224-78438224 on Build GRCh38
rs966113307 Chr.15: 78438058-78438058 on Build GRCh38
rs1006588743 Chr.15: 78437998-78437998 on Build GRCh38
rs774870147 Chr.15: 78438237-78438237 on Build GRCh38
rs558423307 Chr.15: 78438185-78438185 on Build GRCh38
rs923388446 Chr.15: 78438260-78438260 on Build GRCh38
rs200141558 Chr.15: 78438299-78438299 on Build GRCh38
rs1016054882 Chr.15: 78438222-78438222 on Build GRCh38
rs971967780 Chr.15: 78438228-78438228 on Build GRCh38
rs560690740 Chr.15: 78437950-78437950 on Build GRCh38
rs751197364 Chr.15: 78438347-78438347 on Build GRCh38
rs977891750 Chr.15: 78438011-78438011 on Build GRCh38
rs140614053 Chr.15: 78438282-78438282 on Build GRCh38
rs547966028 Chr.15: 78438010-78438010 on Build GRCh38
rs550480425 Chr.15: 78437958-78437958 on Build GRCh38
rs17483548 Chr.15: 78437971-78437971 on Build GRCh38
rs563267339 Chr.15: 78437914-78437914 on Build GRCh38
rs750845445 Chr.15: 78438268-78438268 on Build GRCh38
rs916535819 Chr.15: 78438054-78438054 on Build GRCh38
rs186821594 Chr.15: 78438265-78438265 on Build GRCh38
rs746059773 Chr.15: 78437936-78437936 on Build GRCh38
rs749865527 Chr.15: 78438369-78438369 on Build GRCh38
rs1023479485 Chr.15: 78438321-78438321 on Build GRCh38
rs902073942 Chr.15: 78437959-78437959 on Build GRCh38
rs909263738 Chr.15: 78438096-78438096 on Build GRCh38
rs749117384 Chr.15: 78438380-78438380 on Build GRCh38
rs756847270 Chr.15: 78438348-78438348 on Build GRCh38
rs1056597510 Chr.15: 78437997-78437997 on Build GRCh38
rs1021975048 Chr.15: 78438215-78438215 on Build GRCh38

View Full Product Details