Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49532220-49532783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCBTB1
Entrez Gene ID
55213
Gene Name
RCC1 and BTB domain containing protein 1
Gene Aliases
CLLD7, CLLL7, GLP
UniGene
Hs.508021
Gene Chromosome Location
Chr.13: 49531944-49585606 on Build GRCh38
Transcripts
NM_018191.3 XM_005266441.2 XM_011535133.1 XM_011535134.1 XM_011535135.2

SNP Detail

SNP ID Chromosome Location
rs937977522 Chr.13: 49532290-49532290 on Build GRCh38
rs878974985 Chr.13: 49532539-49532539 on Build GRCh38
rs936877692 Chr.13: 49532532-49532532 on Build GRCh38
rs898996285 Chr.13: 49532617-49532617 on Build GRCh38
rs566457601 Chr.13: 49532323-49532323 on Build GRCh38
rs41284784 Chr.13: 49532646-49532646 on Build GRCh38
rs950020100 Chr.13: 49532351-49532351 on Build GRCh38
rs41284782 Chr.13: 49532604-49532604 on Build GRCh38
rs762032989 Chr.13: 49532661-49532661 on Build GRCh38
rs963237168 Chr.13: 49532337-49532337 on Build GRCh38
rs562115260 Chr.13: 49532674-49532674 on Build GRCh38
rs979236185 Chr.13: 49532267-49532267 on Build GRCh38
rs925445550 Chr.13: 49532530-49532530 on Build GRCh38
rs978394309 Chr.13: 49532522-49532522 on Build GRCh38
rs1046034 Chr.13: 49532675-49532675 on Build GRCh38
rs992536835 Chr.13: 49532442-49532442 on Build GRCh38
rs573773665 Chr.13: 49532403-49532403 on Build GRCh38
rs61705134 Chr.13: 49532269-49532273 on Build GRCh38
rs866831490 Chr.13: 49532564-49532564 on Build GRCh38
rs917842303 Chr.13: 49532692-49532692 on Build GRCh38
rs902074217 Chr.13: 49532528-49532528 on Build GRCh38
rs925074994 Chr.13: 49532278-49532278 on Build GRCh38
rs959449416 Chr.13: 49532677-49532677 on Build GRCh38
rs997549505 Chr.13: 49532244-49532244 on Build GRCh38
rs567112014 Chr.13: 49532433-49532433 on Build GRCh38
rs1016492458 Chr.13: 49532331-49532331 on Build GRCh38
rs188565734 Chr.13: 49532266-49532266 on Build GRCh38
rs3186024 Chr.13: 49532448-49532448 on Build GRCh38
rs1001329923 Chr.13: 49532632-49532632 on Build GRCh38
rs921651375 Chr.13: 49532549-49532549 on Build GRCh38
rs897932216 Chr.13: 49532401-49532401 on Build GRCh38
rs534636863 Chr.13: 49532570-49532570 on Build GRCh38
rs398022717 Chr.13: 49532416-49532416 on Build GRCh38
rs1045736155 Chr.13: 49532367-49532367 on Build GRCh38
rs978746003 Chr.13: 49532709-49532709 on Build GRCh38
rs886128528 Chr.13: 49532598-49532598 on Build GRCh38
rs533818599 Chr.13: 49532420-49532420 on Build GRCh38
rs1007831241 Chr.13: 49532484-49532484 on Build GRCh38
rs1025379195 Chr.13: 49532697-49532697 on Build GRCh38
rs958288622 Chr.13: 49532755-49532755 on Build GRCh38
rs139202166 Chr.13: 49532268-49532272 on Build GRCh38
rs1005195916 Chr.13: 49532599-49532599 on Build GRCh38
rs886326981 Chr.13: 49532293-49532293 on Build GRCh38
rs573592323 Chr.13: 49532633-49532633 on Build GRCh38
rs933097908 Chr.13: 49532558-49532558 on Build GRCh38
rs997156497 Chr.13: 49532550-49532550 on Build GRCh38
rs1039178897 Chr.13: 49532544-49532544 on Build GRCh38
rs34222928 Chr.13: 49532415-49532415 on Build GRCh38
rs1032217024 Chr.13: 49532673-49532673 on Build GRCh38
rs1032514565 Chr.13: 49532731-49532731 on Build GRCh38
rs966767690 Chr.13: 49532699-49532699 on Build GRCh38
rs866497865 Chr.13: 49532478-49532478 on Build GRCh38
rs546456410 Chr.13: 49532595-49532595 on Build GRCh38
rs1013691634 Chr.13: 49532679-49532679 on Build GRCh38
rs9568240 Chr.13: 49532425-49532425 on Build GRCh38
rs1018258399 Chr.13: 49532485-49532485 on Build GRCh38
rs1037657080 Chr.13: 49532608-49532608 on Build GRCh38
rs967342499 Chr.13: 49532516-49532516 on Build GRCh38
rs537950147 Chr.13: 49532446-49532446 on Build GRCh38
rs995584810 Chr.13: 49532383-49532383 on Build GRCh38

View Full Product Details