Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 76427192-76427788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEE1
Entrez Gene ID
57692
Gene Name
MAGE family member E1
Gene Aliases
DAMAGE, HCA1
UniGene
Hs.8453
Gene Chromosome Location
Chr.X: 76427653-76431353 on Build GRCh38
Transcripts
NM_020932.2

SNP Detail

SNP ID Chromosome Location
rs1029106190 Chr.X: 76427347-76427347 on Build GRCh38
rs782207280 Chr.X: 76427438-76427438 on Build GRCh38
rs1044383431 Chr.X: 76427666-76427666 on Build GRCh38
rs995836393 Chr.X: 76427324-76427324 on Build GRCh38
rs368712644 Chr.X: 76427698-76427698 on Build GRCh38
rs1040598928 Chr.X: 76427712-76427712 on Build GRCh38
rs370433342 Chr.X: 76427284-76427284 on Build GRCh38
rs1027797345 Chr.X: 76427242-76427242 on Build GRCh38
rs138833005 Chr.X: 76427243-76427243 on Build GRCh38
rs996455365 Chr.X: 76427728-76427728 on Build GRCh38
rs927301753 Chr.X: 76427670-76427670 on Build GRCh38
rs782381195 Chr.X: 76427629-76427629 on Build GRCh38
rs947999235 Chr.X: 76427626-76427626 on Build GRCh38
rs1019209604 Chr.X: 76427290-76427290 on Build GRCh38
rs917831405 Chr.X: 76427615-76427615 on Build GRCh38
rs782243745 Chr.X: 76427561-76427561 on Build GRCh38
rs782566097 Chr.X: 76427305-76427305 on Build GRCh38
rs1049619625 Chr.X: 76427682-76427682 on Build GRCh38
rs1005050225 Chr.X: 76427705-76427705 on Build GRCh38
rs967806352 Chr.X: 76427359-76427359 on Build GRCh38
rs67821007 Chr.X: 76427551-76427551 on Build GRCh38
rs782662925 Chr.X: 76427218-76427218 on Build GRCh38
rs6647996 Chr.X: 76427234-76427234 on Build GRCh38
rs902084252 Chr.X: 76427727-76427727 on Build GRCh38
rs886567328 Chr.X: 76427691-76427691 on Build GRCh38
rs992000422 Chr.X: 76427572-76427572 on Build GRCh38
rs782411953 Chr.X: 76427522-76427523 on Build GRCh38
rs930714252 Chr.X: 76427675-76427675 on Build GRCh38
rs1005103969 Chr.X: 76427253-76427253 on Build GRCh38

View Full Product Details