Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150114771-150115343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDGFRB
Entrez Gene ID
5159
Gene Name
platelet derived growth factor receptor beta
Gene Aliases
CD140B, IBGC4, IMF1, JTK12, KOGS, PDGFR, PDGFR-1, PDGFR1, PENTT
UniGene
Hs.509067
Gene Chromosome Location
Chr.5: 150113839-150155884 on Build GRCh38
Transcripts
NM_002609.3 XM_005268464.2 XM_011537658.1 XM_011537659.1

SNP Detail

SNP ID Chromosome Location
rs189852053 Chr.5: 150115073-150115073 on Build GRCh38
rs144647473 Chr.5: 150114923-150114923 on Build GRCh38
rs1017375 Chr.5: 150114958-150114958 on Build GRCh38
rs765657661 Chr.5: 150114982-150114982 on Build GRCh38
rs917757168 Chr.5: 150114985-150114985 on Build GRCh38
rs576936094 Chr.5: 150115030-150115030 on Build GRCh38
rs529006877 Chr.5: 150114964-150114964 on Build GRCh38
rs904113288 Chr.5: 150115205-150115205 on Build GRCh38
rs574200760 Chr.5: 150115314-150115314 on Build GRCh38
rs947942447 Chr.5: 150115010-150115010 on Build GRCh38
rs985190342 Chr.5: 150114917-150114917 on Build GRCh38
rs1027836157 Chr.5: 150115196-150115196 on Build GRCh38
rs965135257 Chr.5: 150115301-150115301 on Build GRCh38
rs559602030 Chr.5: 150115170-150115170 on Build GRCh38
rs950148672 Chr.5: 150115077-150115077 on Build GRCh38
rs952339917 Chr.5: 150114914-150114914 on Build GRCh38
rs568575175 Chr.5: 150114880-150114880 on Build GRCh38
rs372138290 Chr.5: 150115079-150115079 on Build GRCh38
rs978662996 Chr.5: 150115020-150115020 on Build GRCh38
rs193195949 Chr.5: 150114802-150114802 on Build GRCh38
rs1019547212 Chr.5: 150114837-150114837 on Build GRCh38
rs7726051 Chr.5: 150115038-150115038 on Build GRCh38
rs919884847 Chr.5: 150115057-150115057 on Build GRCh38
rs956703507 Chr.5: 150115320-150115320 on Build GRCh38
rs925950739 Chr.5: 150115028-150115028 on Build GRCh38
rs1006537946 Chr.5: 150115274-150115274 on Build GRCh38
rs1055853585 Chr.5: 150114791-150114791 on Build GRCh38
rs138535908 Chr.5: 150115121-150115121 on Build GRCh38
rs945994690 Chr.5: 150114980-150114981 on Build GRCh38
rs897990056 Chr.5: 150115167-150115167 on Build GRCh38
rs533828413 Chr.5: 150114965-150114965 on Build GRCh38
rs531952172 Chr.5: 150114830-150114830 on Build GRCh38
rs888132273 Chr.5: 150115209-150115209 on Build GRCh38
rs552875539 Chr.5: 150114803-150114803 on Build GRCh38
rs867537698 Chr.5: 150114932-150114932 on Build GRCh38
rs996383143 Chr.5: 150115177-150115177 on Build GRCh38
rs564508884 Chr.5: 150114811-150114811 on Build GRCh38
rs995089693 Chr.5: 150114889-150114889 on Build GRCh38
rs899559712 Chr.5: 150115163-150115163 on Build GRCh38
rs558839956 Chr.5: 150115002-150115002 on Build GRCh38
rs1017885101 Chr.5: 150115287-150115287 on Build GRCh38
rs1029157384 Chr.5: 150115188-150115188 on Build GRCh38
rs886740269 Chr.5: 150115074-150115074 on Build GRCh38
rs931225770 Chr.5: 150115060-150115060 on Build GRCh38
rs185163183 Chr.5: 150114865-150114865 on Build GRCh38
rs901979961 Chr.5: 150114850-150114850 on Build GRCh38
rs535799011 Chr.5: 150114909-150114909 on Build GRCh38
rs1001077813 Chr.5: 150115283-150115283 on Build GRCh38
rs766404627 Chr.5: 150114955-150114955 on Build GRCh38
rs1027103849 Chr.5: 150114913-150114913 on Build GRCh38
rs555799472 Chr.5: 150115176-150115176 on Build GRCh38

View Full Product Details