Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16804576-16805101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGR2
Entrez Gene ID
10551
Gene Name
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2
UniGene
Hs.530009
Gene Chromosome Location
Chr.7: 16791811-16805114 on Build GRCh38
Transcripts
NM_006408.3 XM_005249581.4

SNP Detail

SNP ID Chromosome Location
rs3817506 Chr.7: 16804708-16804708 on Build GRCh38
rs927769331 Chr.7: 16804769-16804769 on Build GRCh38
rs960578458 Chr.7: 16805029-16805029 on Build GRCh38
rs1050016977 Chr.7: 16804964-16804964 on Build GRCh38
rs1005589740 Chr.7: 16804971-16804971 on Build GRCh38
rs553301716 Chr.7: 16804804-16804804 on Build GRCh38
rs998730097 Chr.7: 16804944-16804944 on Build GRCh38
rs978602361 Chr.7: 16804905-16804905 on Build GRCh38
rs920878944 Chr.7: 16805044-16805044 on Build GRCh38
rs925871105 Chr.7: 16804684-16804684 on Build GRCh38
rs3817508 Chr.7: 16804694-16804694 on Build GRCh38
rs189550968 Chr.7: 16805041-16805041 on Build GRCh38
rs774506538 Chr.7: 16805003-16805003 on Build GRCh38
rs186664031 Chr.7: 16804972-16804972 on Build GRCh38
rs141123982 Chr.7: 16804776-16804777 on Build GRCh38
rs557361426 Chr.7: 16804872-16804872 on Build GRCh38
rs186880333 Chr.7: 16804779-16804779 on Build GRCh38
rs979092709 Chr.7: 16804669-16804669 on Build GRCh38
rs201484466 Chr.7: 16805023-16805026 on Build GRCh38
rs946406880 Chr.7: 16804821-16804821 on Build GRCh38
rs548064472 Chr.7: 16805046-16805046 on Build GRCh38
rs927038679 Chr.7: 16805036-16805036 on Build GRCh38
rs536683644 Chr.7: 16804732-16804732 on Build GRCh38
rs979097032 Chr.7: 16805054-16805054 on Build GRCh38
rs3817507 Chr.7: 16804699-16804699 on Build GRCh38
rs954083341 Chr.7: 16804660-16804660 on Build GRCh38
rs552229239 Chr.7: 16804766-16804766 on Build GRCh38
rs779890961 Chr.7: 16804825-16804825 on Build GRCh38
rs374529180 Chr.7: 16804984-16804984 on Build GRCh38
rs764171706 Chr.7: 16804778-16804779 on Build GRCh38
rs3817510 Chr.7: 16804604-16804604 on Build GRCh38
rs545921374 Chr.7: 16804929-16804929 on Build GRCh38
rs980272151 Chr.7: 16805031-16805031 on Build GRCh38
rs706072 Chr.7: 16805039-16805039 on Build GRCh38
rs1017651815 Chr.7: 16804837-16804837 on Build GRCh38
rs934976271 Chr.7: 16805064-16805064 on Build GRCh38
rs3817505 Chr.7: 16804823-16804823 on Build GRCh38
rs1035075406 Chr.7: 16804994-16804994 on Build GRCh38
rs706073 Chr.7: 16805057-16805057 on Build GRCh38
rs961482642 Chr.7: 16804983-16804983 on Build GRCh38
rs564593214 Chr.7: 16804969-16804969 on Build GRCh38
rs899404745 Chr.7: 16804799-16804799 on Build GRCh38
rs80282641 Chr.7: 16804731-16804731 on Build GRCh38
rs748967988 Chr.7: 16804939-16804939 on Build GRCh38
rs1053242197 Chr.7: 16804888-16804888 on Build GRCh38
rs890577181 Chr.7: 16804834-16804834 on Build GRCh38
rs3817509 Chr.7: 16804664-16804664 on Build GRCh38
rs183141930 Chr.7: 16804916-16804916 on Build GRCh38
rs867052056 Chr.7: 16804625-16804625 on Build GRCh38
rs901365838 Chr.7: 16804920-16804920 on Build GRCh38
rs750570498 Chr.7: 16805021-16805024 on Build GRCh38
rs74997255 Chr.7: 16804656-16804656 on Build GRCh38
rs935071471 Chr.7: 16804835-16804835 on Build GRCh38
rs535211878 Chr.7: 16804790-16804790 on Build GRCh38
rs192073830 Chr.7: 16804805-16804805 on Build GRCh38
rs573644584 Chr.7: 16804624-16804624 on Build GRCh38
rs373406206 Chr.7: 16804612-16804612 on Build GRCh38
rs550687600 Chr.7: 16804955-16804955 on Build GRCh38
rs541172476 Chr.7: 16804978-16804978 on Build GRCh38

View Full Product Details