Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93443422-93444014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCR
Entrez Gene ID
799
Gene Name
calcitonin receptor
Gene Aliases
CRT, CT-R, CTR, CTR1
UniGene
Hs.489127
Gene Chromosome Location
Chr.7: 93424487-93574730 on Build GRCh38
Transcripts
NM_001164737.1 NM_001164738.1 NM_001742.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
747483 c.764T>C p.I255T Chr.7: 93443744-93443744 on Build GRCh38
3642418 c.832C>T p.L278F Chr.7: 93443676-93443676 on Build GRCh38
747484 c.662T>C p.I221T Chr.7: 93443744-93443744 on Build GRCh38
3929265 c.882G>C p.R294R Chr.7: 93443626-93443626 on Build GRCh38
2864821 c.776delT p.F259fs*5 Chr.7: 93443732-93443732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558278405 Chr.7: 93443982-93443982 on Build GRCh38
rs1043579927 Chr.7: 93443483-93443483 on Build GRCh38
rs1006907345 Chr.7: 93443694-93443694 on Build GRCh38
rs113190351 Chr.7: 93443773-93443773 on Build GRCh38
rs764187507 Chr.7: 93443633-93443633 on Build GRCh38
rs759481314 Chr.7: 93443809-93443809 on Build GRCh38
rs41278800 Chr.7: 93443892-93443892 on Build GRCh38
rs780711217 Chr.7: 93443666-93443666 on Build GRCh38
rs1020323738 Chr.7: 93443842-93443842 on Build GRCh38
rs867922665 Chr.7: 93443643-93443643 on Build GRCh38
rs945330509 Chr.7: 93443828-93443828 on Build GRCh38
rs893250593 Chr.7: 93443816-93443816 on Build GRCh38
rs568371015 Chr.7: 93443597-93443597 on Build GRCh38
rs896246996 Chr.7: 93443864-93443864 on Build GRCh38
rs200469954 Chr.7: 93443711-93443711 on Build GRCh38
rs368464231 Chr.7: 93443634-93443634 on Build GRCh38
rs777622964 Chr.7: 93443768-93443768 on Build GRCh38
rs201514395 Chr.7: 93443567-93443567 on Build GRCh38
rs540179321 Chr.7: 93443632-93443632 on Build GRCh38
rs144902080 Chr.7: 93443628-93443628 on Build GRCh38
rs572439533 Chr.7: 93443889-93443889 on Build GRCh38
rs960546457 Chr.7: 93443925-93443925 on Build GRCh38
rs370793228 Chr.7: 93443579-93443579 on Build GRCh38
rs536171395 Chr.7: 93443829-93443829 on Build GRCh38
rs886092619 Chr.7: 93443920-93443920 on Build GRCh38
rs752614232 Chr.7: 93443671-93443671 on Build GRCh38
rs748752860 Chr.7: 93443874-93443874 on Build GRCh38
rs758827884 Chr.7: 93443653-93443653 on Build GRCh38
rs2074123 Chr.7: 93443835-93443835 on Build GRCh38
rs989384628 Chr.7: 93443504-93443504 on Build GRCh38
rs199836961 Chr.7: 93443557-93443557 on Build GRCh38
rs1002392281 Chr.7: 93443515-93443515 on Build GRCh38
rs774313528 Chr.7: 93443801-93443801 on Build GRCh38
rs1013267250 Chr.7: 93443569-93443569 on Build GRCh38
rs903689518 Chr.7: 93443505-93443505 on Build GRCh38
rs966676351 Chr.7: 93443513-93443513 on Build GRCh38
rs781540723 Chr.7: 93443608-93443624 on Build GRCh38
rs73419385 Chr.7: 93443641-93443641 on Build GRCh38
rs945256631 Chr.7: 93443476-93443476 on Build GRCh38
rs774475094 Chr.7: 93443627-93443627 on Build GRCh38
rs758844860 Chr.7: 93443572-93443572 on Build GRCh38
rs954923867 Chr.7: 93443763-93443763 on Build GRCh38
rs768250423 Chr.7: 93443608-93443608 on Build GRCh38
rs937879138 Chr.7: 93443841-93443841 on Build GRCh38
rs746516889 Chr.7: 93443604-93443604 on Build GRCh38
rs747785006 Chr.7: 93443621-93443621 on Build GRCh38
rs2074124 Chr.7: 93443836-93443836 on Build GRCh38
rs140830339 Chr.7: 93443848-93443848 on Build GRCh38
rs975291322 Chr.7: 93443673-93443673 on Build GRCh38
rs769425672 Chr.7: 93443727-93443727 on Build GRCh38
rs751388015 Chr.7: 93443624-93443624 on Build GRCh38
rs548671354 Chr.7: 93443658-93443658 on Build GRCh38
rs892390543 Chr.7: 93443461-93443461 on Build GRCh38
rs371796364 Chr.7: 93443603-93443603 on Build GRCh38
rs375357765 Chr.7: 93443794-93443794 on Build GRCh38
rs755461448 Chr.7: 93443661-93443661 on Build GRCh38
rs750774000 Chr.7: 93443651-93443651 on Build GRCh38
rs990570544 Chr.7: 93443614-93443614 on Build GRCh38
rs1053500143 Chr.7: 93443460-93443460 on Build GRCh38
rs1033418572 Chr.7: 93443563-93443563 on Build GRCh38
rs923393974 Chr.7: 93443753-93443753 on Build GRCh38
rs563202976 Chr.7: 93443791-93443791 on Build GRCh38
rs554210532 Chr.7: 93443618-93443618 on Build GRCh38
rs149950588 Chr.7: 93443626-93443626 on Build GRCh38
rs781038636 Chr.7: 93443659-93443659 on Build GRCh38
rs769360550 Chr.7: 93443623-93443623 on Build GRCh38

View Full Product Details