Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154129329-154129878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
UniGene
Hs.240845
Gene Chromosome Location
Chr.3: 154121003-154257827 on Build GRCh38
Transcripts
NM_001251962.1 NM_001251963.1 NM_015595.3 XM_011512672.1 XM_017006160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1420084 c.1159G>A p.A387T Chr.3: 154129609-154129609 on Build GRCh38
3660268 c.1273A>T p.K425* Chr.3: 154129723-154129723 on Build GRCh38
4780396 c.1137G>A p.L379L Chr.3: 154129587-154129587 on Build GRCh38
4137003 c.1262T>C p.L421P Chr.3: 154129712-154129712 on Build GRCh38
4114610 c.1142A>G p.Q381R Chr.3: 154129592-154129592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779554378 Chr.3: 154129697-154129697 on Build GRCh38
rs759904925 Chr.3: 154129653-154129653 on Build GRCh38
rs528077497 Chr.3: 154129522-154129522 on Build GRCh38
rs774509781 Chr.3: 154129571-154129571 on Build GRCh38
rs535567953 Chr.3: 154129757-154129757 on Build GRCh38
rs780818199 Chr.3: 154129544-154129544 on Build GRCh38
rs16823679 Chr.3: 154129807-154129807 on Build GRCh38
rs945949693 Chr.3: 154129518-154129518 on Build GRCh38
rs935558255 Chr.3: 154129363-154129363 on Build GRCh38
rs770913818 Chr.3: 154129572-154129572 on Build GRCh38
rs900002002 Chr.3: 154129805-154129805 on Build GRCh38
rs759502710 Chr.3: 154129752-154129752 on Build GRCh38
rs777945173 Chr.3: 154129679-154129679 on Build GRCh38
rs367845420 Chr.3: 154129611-154129611 on Build GRCh38
rs376491851 Chr.3: 154129562-154129562 on Build GRCh38
rs1008388716 Chr.3: 154129372-154129372 on Build GRCh38
rs958394665 Chr.3: 154129376-154129376 on Build GRCh38
rs779888356 Chr.3: 154129529-154129531 on Build GRCh38
rs960634688 Chr.3: 154129542-154129542 on Build GRCh38
rs201024945 Chr.3: 154129718-154129718 on Build GRCh38
rs762803415 Chr.3: 154129719-154129719 on Build GRCh38
rs763801051 Chr.3: 154129756-154129756 on Build GRCh38
rs202240716 Chr.3: 154129762-154129762 on Build GRCh38
rs768144199 Chr.3: 154129688-154129689 on Build GRCh38
rs879608764 Chr.3: 154129403-154129403 on Build GRCh38
rs764898548 Chr.3: 154129625-154129625 on Build GRCh38
rs772339917 Chr.3: 154129595-154129595 on Build GRCh38
rs1019804030 Chr.3: 154129792-154129792 on Build GRCh38
rs546158318 Chr.3: 154129555-154129555 on Build GRCh38
rs747148864 Chr.3: 154129710-154129710 on Build GRCh38
rs1052476146 Chr.3: 154129715-154129715 on Build GRCh38
rs989492694 Chr.3: 154129479-154129479 on Build GRCh38
rs751565334 Chr.3: 154129637-154129637 on Build GRCh38
rs938483328 Chr.3: 154129557-154129557 on Build GRCh38
rs778232614 Chr.3: 154129561-154129561 on Build GRCh38
rs766220298 Chr.3: 154129722-154129722 on Build GRCh38
rs566784807 Chr.3: 154129404-154129404 on Build GRCh38
rs757766581 Chr.3: 154129690-154129690 on Build GRCh38
rs1028948036 Chr.3: 154129837-154129838 on Build GRCh38
rs772243711 Chr.3: 154129702-154129702 on Build GRCh38
rs767054994 Chr.3: 154129655-154129655 on Build GRCh38
rs1626280 Chr.3: 154129793-154129793 on Build GRCh38
rs769786046 Chr.3: 154129759-154129761 on Build GRCh38
rs1014781635 Chr.3: 154129499-154129499 on Build GRCh38
rs567827890 Chr.3: 154129558-154129558 on Build GRCh38
rs925284065 Chr.3: 154129362-154129362 on Build GRCh38
rs372771888 Chr.3: 154129745-154129745 on Build GRCh38
rs766308185 Chr.3: 154129635-154129635 on Build GRCh38
rs775828838 Chr.3: 154129609-154129609 on Build GRCh38
rs759687570 Chr.3: 154129576-154129576 on Build GRCh38
rs1027280146 Chr.3: 154129423-154129423 on Build GRCh38
rs773079785 Chr.3: 154129626-154129626 on Build GRCh38
rs879459535 Chr.3: 154129846-154129846 on Build GRCh38
rs752267103 Chr.3: 154129753-154129753 on Build GRCh38
rs544320181 Chr.3: 154129817-154129817 on Build GRCh38
rs188343550 Chr.3: 154129796-154129796 on Build GRCh38
rs11918824 Chr.3: 154129360-154129360 on Build GRCh38
rs746421883 Chr.3: 154129701-154129701 on Build GRCh38
rs747847166 Chr.3: 154129554-154129554 on Build GRCh38
rs749670829 Chr.3: 154129684-154129684 on Build GRCh38
rs891420494 Chr.3: 154129367-154129367 on Build GRCh38
rs755657875 Chr.3: 154129754-154129754 on Build GRCh38
rs991928659 Chr.3: 154129553-154129553 on Build GRCh38
rs768038426 Chr.3: 154129708-154129708 on Build GRCh38
rs771484932 Chr.3: 154129565-154129565 on Build GRCh38
rs540647263 Chr.3: 154129818-154129818 on Build GRCh38
rs574316844 Chr.3: 154129418-154129418 on Build GRCh38
rs1004586510 Chr.3: 154129498-154129498 on Build GRCh38
rs746636607 Chr.3: 154129603-154129614 on Build GRCh38
rs755814717 Chr.3: 154129678-154129678 on Build GRCh38
rs1053116302 Chr.3: 154129365-154129365 on Build GRCh38
rs776234509 Chr.3: 154129725-154129728 on Build GRCh38
rs752254191 Chr.3: 154129660-154129660 on Build GRCh38
rs761163589 Chr.3: 154129754-154129756 on Build GRCh38
rs762739266 Chr.3: 154129628-154129628 on Build GRCh38
rs951798161 Chr.3: 154129451-154129451 on Build GRCh38
rs1055670671 Chr.3: 154129568-154129568 on Build GRCh38

View Full Product Details