Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94491626-94492196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1429
Entrez Gene ID
25962
Gene Name
KIAA1429
Gene Aliases
MSTP054, fSAP121
UniGene
Hs.202238
Gene Chromosome Location
Chr.8: 94487777-94553518 on Build GRCh38
Transcripts
NM_015496.4 XM_017013306.1 XM_017013307.1 XM_017013308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530449 c.4825A>G p.I1609V Chr.8: 94491893-94491893 on Build GRCh38
1255709 c.4817C>T p.S1606F Chr.8: 94491901-94491901 on Build GRCh38
3996180 c.5073A>G p.S1691S Chr.8: 94491645-94491645 on Build GRCh38
3902147 c.4994T>C p.V1665A Chr.8: 94491724-94491724 on Build GRCh38
5482224 c.5060G>A p.R1687H Chr.8: 94491658-94491658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982836419 Chr.8: 94491682-94491682 on Build GRCh38
rs778621449 Chr.8: 94491882-94491882 on Build GRCh38
rs944371480 Chr.8: 94492107-94492107 on Build GRCh38
rs934422335 Chr.8: 94492027-94492027 on Build GRCh38
rs890161590 Chr.8: 94492083-94492083 on Build GRCh38
rs369018696 Chr.8: 94491665-94491665 on Build GRCh38
rs982466360 Chr.8: 94492118-94492118 on Build GRCh38
rs753864242 Chr.8: 94491796-94491796 on Build GRCh38
rs752822435 Chr.8: 94491777-94491777 on Build GRCh38
rs1033056672 Chr.8: 94492040-94492040 on Build GRCh38
rs140246840 Chr.8: 94492006-94492006 on Build GRCh38
rs746597322 Chr.8: 94491922-94491922 on Build GRCh38
rs766486921 Chr.8: 94491732-94491732 on Build GRCh38
rs540622209 Chr.8: 94492007-94492007 on Build GRCh38
rs1052787858 Chr.8: 94492036-94492036 on Build GRCh38
rs753865506 Chr.8: 94491837-94491837 on Build GRCh38
rs922743498 Chr.8: 94492065-94492065 on Build GRCh38
rs143450140 Chr.8: 94491844-94491844 on Build GRCh38
rs979003079 Chr.8: 94491998-94491998 on Build GRCh38
rs749962172 Chr.8: 94491884-94491884 on Build GRCh38
rs1014329250 Chr.8: 94491964-94491964 on Build GRCh38
rs975720149 Chr.8: 94492064-94492064 on Build GRCh38
rs778082605 Chr.8: 94491955-94491955 on Build GRCh38
rs879137027 Chr.8: 94491709-94491709 on Build GRCh38
rs759813402 Chr.8: 94491750-94491750 on Build GRCh38
rs557251478 Chr.8: 94491659-94491659 on Build GRCh38
rs3098725 Chr.8: 94491996-94491996 on Build GRCh38
rs1015632043 Chr.8: 94491692-94491692 on Build GRCh38
rs767795722 Chr.8: 94491765-94491765 on Build GRCh38
rs200618514 Chr.8: 94491669-94491669 on Build GRCh38
rs770018328 Chr.8: 94491698-94491698 on Build GRCh38
rs773793629 Chr.8: 94491703-94491703 on Build GRCh38
rs774412750 Chr.8: 94491741-94491741 on Build GRCh38
rs915805492 Chr.8: 94491977-94491977 on Build GRCh38
rs201808627 Chr.8: 94491693-94491693 on Build GRCh38
rs771832914 Chr.8: 94491678-94491678 on Build GRCh38
rs763431931 Chr.8: 94491704-94491704 on Build GRCh38
rs543344055 Chr.8: 94492152-94492152 on Build GRCh38
rs755056305 Chr.8: 94491915-94491916 on Build GRCh38
rs964638296 Chr.8: 94492063-94492063 on Build GRCh38
rs779790729 Chr.8: 94491919-94491919 on Build GRCh38
rs1056690867 Chr.8: 94491864-94491864 on Build GRCh38
rs560591414 Chr.8: 94492021-94492021 on Build GRCh38
rs756875731 Chr.8: 94491657-94491657 on Build GRCh38
rs755960033 Chr.8: 94491792-94491792 on Build GRCh38
rs770246067 Chr.8: 94491937-94491937 on Build GRCh38
rs199663643 Chr.8: 94491949-94491949 on Build GRCh38
rs200214954 Chr.8: 94491825-94491825 on Build GRCh38
rs997086627 Chr.8: 94492135-94492135 on Build GRCh38
rs757777053 Chr.8: 94491894-94491894 on Build GRCh38
rs749861015 Chr.8: 94491959-94491959 on Build GRCh38
rs748554192 Chr.8: 94491684-94491684 on Build GRCh38
rs28564036 Chr.8: 94492167-94492167 on Build GRCh38
rs763773678 Chr.8: 94491823-94491823 on Build GRCh38
rs78871083 Chr.8: 94492119-94492119 on Build GRCh38
rs900087987 Chr.8: 94492122-94492122 on Build GRCh38
rs577339919 Chr.8: 94491680-94491680 on Build GRCh38

View Full Product Details