Product Details
- Primer Sequences
- Log in to view
- Primer Chromosome Location
-
Chr.10:
46040225-46039910
on Build GRCh38
View in UCSC browser
- Format
- Each primer will be delivered in a single tube, dried-down
- Quick Reference Guide
- Primer Designer™ Tool Quick Reference
- Ampliseq ID's
Gene Details
- Gene Symbol
- MSMB
- Entrez Gene ID
-
4477
- Gene Name
- microseminoprotein beta
- Gene Aliases
-
HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94
- UniGene
- Hs.255462
- Gene Chromosome Location
-
Chr.10:
46033305-46046269
on Build GRCh38
- Transcripts
-
NM_002443.3
NM_138634.2
View Full Product Details