Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125836190-125836789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_078468.2 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Gene Symbol
DHX32
Entrez Gene ID
55760
Gene Name
DEAH-box helicase 32 (putative)
Gene Aliases
DDX32, DHLP1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125836337-125896483 on Build GRCh38
Transcripts
NM_016567.3 NM_078468.2 NM_078469.2 NM_018180.2 XM_017016404.1 XM_017016405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297048 c.2203G>A p.E735K Chr.10: 125836716-125836716 on Build GRCh38
4012520 c.2143G>A p.D715N Chr.10: 125836776-125836776 on Build GRCh38
7203953 c.927A>G p.K309K Chr.10: 125836256-125836256 on Build GRCh38
6023170 c.2195C>T p.T732M Chr.10: 125836724-125836724 on Build GRCh38
7175312 c.2209G>A p.E737K Chr.10: 125836710-125836710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779287657 Chr.10: 125836232-125836232 on Build GRCh38
rs765297092 Chr.10: 125836663-125836663 on Build GRCh38
rs767417807 Chr.10: 125836755-125836755 on Build GRCh38
rs1002158569 Chr.10: 125836560-125836560 on Build GRCh38
rs368995976 Chr.10: 125836710-125836710 on Build GRCh38
rs372141280 Chr.10: 125836680-125836680 on Build GRCh38
rs773988110 Chr.10: 125836613-125836613 on Build GRCh38
rs183040118 Chr.10: 125836362-125836362 on Build GRCh38
rs767375243 Chr.10: 125836678-125836678 on Build GRCh38
rs762783392 Chr.10: 125836588-125836588 on Build GRCh38
rs188432839 Chr.10: 125836418-125836418 on Build GRCh38
rs545907861 Chr.10: 125836607-125836607 on Build GRCh38
rs142010622 Chr.10: 125836732-125836732 on Build GRCh38
rs773018038 Chr.10: 125836761-125836761 on Build GRCh38
rs778339284 Chr.10: 125836249-125836249 on Build GRCh38
rs757203195 Chr.10: 125836218-125836218 on Build GRCh38
rs755174172 Chr.10: 125836762-125836763 on Build GRCh38
rs748920499 Chr.10: 125836733-125836733 on Build GRCh38
rs755176101 Chr.10: 125836224-125836224 on Build GRCh38
rs374910849 Chr.10: 125836697-125836697 on Build GRCh38
rs375257974 Chr.10: 125836656-125836656 on Build GRCh38
rs775132088 Chr.10: 125836320-125836320 on Build GRCh38
rs976026931 Chr.10: 125836662-125836662 on Build GRCh38
rs748457521 Chr.10: 125836234-125836234 on Build GRCh38
rs542077938 Chr.10: 125836599-125836599 on Build GRCh38
rs935044344 Chr.10: 125836520-125836520 on Build GRCh38
rs558654554 Chr.10: 125836441-125836441 on Build GRCh38
rs145409042 Chr.10: 125836223-125836223 on Build GRCh38
rs527851917 Chr.10: 125836605-125836605 on Build GRCh38
rs757734941 Chr.10: 125836723-125836723 on Build GRCh38
rs778249535 Chr.10: 125836702-125836702 on Build GRCh38
rs763866774 Chr.10: 125836657-125836657 on Build GRCh38
rs529488671 Chr.10: 125836535-125836535 on Build GRCh38
rs769429852 Chr.10: 125836319-125836319 on Build GRCh38
rs566626865 Chr.10: 125836376-125836376 on Build GRCh38
rs199520716 Chr.10: 125836730-125836730 on Build GRCh38
rs771384114 Chr.10: 125836285-125836285 on Build GRCh38
rs747526355 Chr.10: 125836255-125836255 on Build GRCh38
rs543896966 Chr.10: 125836504-125836504 on Build GRCh38
rs745394008 Chr.10: 125836306-125836306 on Build GRCh38
rs150024151 Chr.10: 125836561-125836561 on Build GRCh38
rs1034481427 Chr.10: 125836582-125836582 on Build GRCh38
rs745318505 Chr.10: 125836728-125836728 on Build GRCh38
rs951497071 Chr.10: 125836347-125836347 on Build GRCh38
rs896284434 Chr.10: 125836552-125836552 on Build GRCh38
rs964141130 Chr.10: 125836623-125836623 on Build GRCh38
rs1009884503 Chr.10: 125836506-125836506 on Build GRCh38
rs900299989 Chr.10: 125836460-125836460 on Build GRCh38
rs904765564 Chr.10: 125836547-125836547 on Build GRCh38
rs138878584 Chr.10: 125836724-125836724 on Build GRCh38
rs772571642 Chr.10: 125836743-125836743 on Build GRCh38
rs941963264 Chr.10: 125836608-125836608 on Build GRCh38
rs974635581 Chr.10: 125836673-125836673 on Build GRCh38
rs780113404 Chr.10: 125836570-125836570 on Build GRCh38
rs918604016 Chr.10: 125836286-125836286 on Build GRCh38
rs111628612 Chr.10: 125836480-125836480 on Build GRCh38
rs560318203 Chr.10: 125836600-125836600 on Build GRCh38
rs964195018 Chr.10: 125836467-125836467 on Build GRCh38
rs760722649 Chr.10: 125836762-125836762 on Build GRCh38
rs772560768 Chr.10: 125836235-125836235 on Build GRCh38
rs776168967 Chr.10: 125836305-125836305 on Build GRCh38
rs35941699 Chr.10: 125836703-125836703 on Build GRCh38
rs923666207 Chr.10: 125836508-125836508 on Build GRCh38
rs575071755 Chr.10: 125836579-125836579 on Build GRCh38
rs761768015 Chr.10: 125836669-125836669 on Build GRCh38
rs941892344 Chr.10: 125836437-125836437 on Build GRCh38
rs986913762 Chr.10: 125836389-125836389 on Build GRCh38
rs933077308 Chr.10: 125836493-125836493 on Build GRCh38
rs758741108 Chr.10: 125836698-125836698 on Build GRCh38
rs150831914 Chr.10: 125836737-125836737 on Build GRCh38
rs15281 Chr.10: 125836679-125836679 on Build GRCh38
rs909128683 Chr.10: 125836408-125836408 on Build GRCh38
rs78102986 Chr.10: 125836336-125836336 on Build GRCh38
rs984131005 Chr.10: 125836606-125836606 on Build GRCh38
rs774144111 Chr.10: 125836744-125836744 on Build GRCh38
rs1022331659 Chr.10: 125836538-125836538 on Build GRCh38
rs7320 Chr.10: 125836503-125836503 on Build GRCh38
rs761523347 Chr.10: 125836753-125836753 on Build GRCh38
rs983798171 Chr.10: 125836359-125836359 on Build GRCh38

View Full Product Details