Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 88149460-88150047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB38
Entrez Gene ID
23682
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
NY-MEL-1, rrGTPbp
UniGene
Hs.591975
Gene Chromosome Location
Chr.11: 87809490-88175505 on Build GRCh38
Transcripts
NM_022337.2 XM_017017455.1 XM_017017456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4037277 c.285A>G p.P95P Chr.11: 88149873-88149873 on Build GRCh38
5757798 c.224T>C p.M75T Chr.11: 88149934-88149934 on Build GRCh38
7093467 c.210A>C p.E70D Chr.11: 88149948-88149948 on Build GRCh38
2222032 c.457G>A p.V153I Chr.11: 88149701-88149701 on Build GRCh38
7190218 c.289A>G p.T97A Chr.11: 88149869-88149869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776366695 Chr.11: 88149614-88149614 on Build GRCh38
rs201341587 Chr.11: 88149781-88149781 on Build GRCh38
rs1037202963 Chr.11: 88149867-88149867 on Build GRCh38
rs372769467 Chr.11: 88149815-88149815 on Build GRCh38
rs978381301 Chr.11: 88149559-88149559 on Build GRCh38
rs191074221 Chr.11: 88149603-88149603 on Build GRCh38
rs78426990 Chr.11: 88149669-88149669 on Build GRCh38
rs1025101024 Chr.11: 88149537-88149537 on Build GRCh38
rs763437538 Chr.11: 88149947-88149947 on Build GRCh38
rs749806044 Chr.11: 88149665-88149665 on Build GRCh38
rs999870902 Chr.11: 88150014-88150014 on Build GRCh38
rs372563653 Chr.11: 88149825-88149825 on Build GRCh38
rs777987296 Chr.11: 88149767-88149767 on Build GRCh38
rs947311162 Chr.11: 88149499-88149499 on Build GRCh38
rs373661702 Chr.11: 88149922-88149922 on Build GRCh38
rs765412355 Chr.11: 88149744-88149744 on Build GRCh38
rs79353735 Chr.11: 88149528-88149528 on Build GRCh38
rs116516384 Chr.11: 88149702-88149702 on Build GRCh38
rs764496086 Chr.11: 88149951-88149951 on Build GRCh38
rs936922475 Chr.11: 88149610-88149610 on Build GRCh38
rs151139535 Chr.11: 88149638-88149638 on Build GRCh38
rs888480983 Chr.11: 88149841-88149841 on Build GRCh38
rs866417610 Chr.11: 88149940-88149940 on Build GRCh38
rs897368073 Chr.11: 88149677-88149677 on Build GRCh38
rs773331157 Chr.11: 88149684-88149684 on Build GRCh38
rs748342210 Chr.11: 88149678-88149678 on Build GRCh38
rs139725413 Chr.11: 88149742-88149742 on Build GRCh38
rs369454117 Chr.11: 88149879-88149879 on Build GRCh38
rs753236129 Chr.11: 88149943-88149943 on Build GRCh38
rs755424084 Chr.11: 88149974-88149974 on Build GRCh38
rs924475341 Chr.11: 88149553-88149553 on Build GRCh38
rs765888915 Chr.11: 88149935-88149935 on Build GRCh38
rs774036738 Chr.11: 88149812-88149812 on Build GRCh38
rs760114172 Chr.11: 88149931-88149931 on Build GRCh38
rs767112167 Chr.11: 88149927-88149927 on Build GRCh38
rs192780970 Chr.11: 88149904-88149904 on Build GRCh38
rs780468403 Chr.11: 88149801-88149801 on Build GRCh38
rs765696305 Chr.11: 88149845-88149845 on Build GRCh38
rs929768751 Chr.11: 88149706-88149706 on Build GRCh38
rs61758769 Chr.11: 88149803-88149803 on Build GRCh38
rs751174045 Chr.11: 88149718-88149718 on Build GRCh38
rs200000971 Chr.11: 88149949-88149949 on Build GRCh38
rs910441338 Chr.11: 88149495-88149495 on Build GRCh38
rs61749247 Chr.11: 88149743-88149743 on Build GRCh38
rs745778831 Chr.11: 88149800-88149800 on Build GRCh38
rs766837126 Chr.11: 88149900-88149900 on Build GRCh38
rs755523753 Chr.11: 88149745-88149745 on Build GRCh38
rs888679591 Chr.11: 88149633-88149633 on Build GRCh38
rs925504789 Chr.11: 88149593-88149593 on Build GRCh38
rs150190201 Chr.11: 88149930-88149930 on Build GRCh38
rs539231699 Chr.11: 88149565-88149565 on Build GRCh38
rs527546669 Chr.11: 88149831-88149831 on Build GRCh38
rs535940265 Chr.11: 88149482-88149482 on Build GRCh38
rs1044388552 Chr.11: 88149828-88149828 on Build GRCh38
rs761774745 Chr.11: 88149675-88149676 on Build GRCh38
rs143951992 Chr.11: 88149707-88149707 on Build GRCh38
rs757725742 Chr.11: 88149968-88149968 on Build GRCh38
rs747210498 Chr.11: 88149775-88149775 on Build GRCh38
rs139102451 Chr.11: 88149996-88149996 on Build GRCh38
rs756747428 Chr.11: 88149648-88149648 on Build GRCh38
rs781609607 Chr.11: 88149701-88149701 on Build GRCh38
rs140912752 Chr.11: 88149908-88149908 on Build GRCh38
rs529999144 Chr.11: 88149739-88149739 on Build GRCh38
rs985704446 Chr.11: 88149926-88149926 on Build GRCh38
rs993083322 Chr.11: 88149811-88149811 on Build GRCh38
rs771031060 Chr.11: 88149783-88149783 on Build GRCh38
rs774050658 Chr.11: 88149916-88149916 on Build GRCh38
rs758723669 Chr.11: 88149758-88149758 on Build GRCh38
rs986206697 Chr.11: 88149491-88149491 on Build GRCh38
rs753850141 Chr.11: 88149663-88149671 on Build GRCh38
rs779386796 Chr.11: 88149990-88149990 on Build GRCh38
rs1044810337 Chr.11: 88149623-88149623 on Build GRCh38
rs1051717924 Chr.11: 88149917-88149917 on Build GRCh38
rs757468293 Chr.11: 88149886-88149886 on Build GRCh38
rs751820797 Chr.11: 88149953-88149953 on Build GRCh38
rs769998112 Chr.11: 88149894-88149894 on Build GRCh38
rs976847985 Chr.11: 88150009-88150013 on Build GRCh38
rs748466802 Chr.11: 88149751-88149751 on Build GRCh38
rs561387314 Chr.11: 88149808-88149808 on Build GRCh38
rs917318545 Chr.11: 88149814-88149814 on Build GRCh38
rs752050923 Chr.11: 88149945-88149945 on Build GRCh38
rs377161063 Chr.11: 88149662-88149662 on Build GRCh38
rs151063326 Chr.11: 88149834-88149834 on Build GRCh38
rs541415439 Chr.11: 88149639-88149639 on Build GRCh38
rs781587258 Chr.11: 88149887-88149887 on Build GRCh38
rs1057178038 Chr.11: 88149601-88149601 on Build GRCh38
rs985797965 Chr.11: 88149733-88149733 on Build GRCh38
rs770922053 Chr.11: 88149697-88149697 on Build GRCh38
rs569757741 Chr.11: 88150023-88150023 on Build GRCh38
rs778058185 Chr.11: 88149876-88149876 on Build GRCh38
rs377211780 Chr.11: 88149836-88149836 on Build GRCh38
rs764154279 Chr.11: 88149659-88149660 on Build GRCh38
rs753151478 Chr.11: 88149860-88149860 on Build GRCh38
rs556812378 Chr.11: 88149516-88149516 on Build GRCh38
rs757412520 Chr.11: 88149607-88149607 on Build GRCh38
rs34326733 Chr.11: 88150016-88150016 on Build GRCh38
rs760769816 Chr.11: 88149685-88149685 on Build GRCh38
rs779231433 Chr.11: 88149750-88149750 on Build GRCh38
rs779321464 Chr.11: 88149672-88149672 on Build GRCh38
rs551154044 Chr.11: 88149786-88149786 on Build GRCh38
rs146840876 Chr.11: 88149708-88149708 on Build GRCh38
rs745901526 Chr.11: 88149888-88149888 on Build GRCh38
rs1017280522 Chr.11: 88150008-88150008 on Build GRCh38
rs781497218 Chr.11: 88149973-88149973 on Build GRCh38
rs563421881 Chr.11: 88149713-88149713 on Build GRCh38
rs772390244 Chr.11: 88149680-88149680 on Build GRCh38
rs934732999 Chr.11: 88149568-88149568 on Build GRCh38
rs959810633 Chr.11: 88149511-88149511 on Build GRCh38
rs200594259 Chr.11: 88149872-88149872 on Build GRCh38
rs780539986 Chr.11: 88149655-88149655 on Build GRCh38
rs878895282 Chr.11: 88149985-88149985 on Build GRCh38
rs756730383 Chr.11: 88149725-88149725 on Build GRCh38
rs762740402 Chr.11: 88149802-88149802 on Build GRCh38
rs536220520 Chr.11: 88150012-88150012 on Build GRCh38
rs1007756989 Chr.11: 88149941-88149941 on Build GRCh38

View Full Product Details