Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52810595-52811108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT4
Entrez Gene ID
3851
Gene Name
keratin 4
Gene Aliases
CK-4, CK4, CYK4, K4, WSN1
UniGene
Hs.654610
Gene Chromosome Location
Chr.12: 52806543-52814116 on Build GRCh38
Transcripts
NM_002272.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5963363 c.960+9C>T p.? Chr.12: 52810747-52810747 on Build GRCh38
1931994 c.922C>T p.R308C Chr.12: 52810794-52810794 on Build GRCh38
940848 c.923G>A p.R308H Chr.12: 52810793-52810793 on Build GRCh38
6014971 c.936G>A p.E312E Chr.12: 52810780-52810780 on Build GRCh38
940847 c.933C>T p.A311A Chr.12: 52810783-52810783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972743737 Chr.12: 52810885-52810885 on Build GRCh38
rs752793675 Chr.12: 52810731-52810731 on Build GRCh38
rs761048664 Chr.12: 52810708-52810708 on Build GRCh38
rs764594635 Chr.12: 52810862-52810863 on Build GRCh38
rs915739569 Chr.12: 52810945-52810945 on Build GRCh38
rs536036061 Chr.12: 52811011-52811011 on Build GRCh38
rs191198104 Chr.12: 52810728-52810728 on Build GRCh38
rs139700086 Chr.12: 52810831-52810831 on Build GRCh38
rs769622208 Chr.12: 52810851-52810851 on Build GRCh38
rs904294153 Chr.12: 52810985-52810985 on Build GRCh38
rs375246611 Chr.12: 52810794-52810794 on Build GRCh38
rs575752087 Chr.12: 52811079-52811079 on Build GRCh38
rs574874911 Chr.12: 52810983-52810983 on Build GRCh38
rs199930715 Chr.12: 52810735-52810735 on Build GRCh38
rs557491343 Chr.12: 52811071-52811071 on Build GRCh38
rs371095509 Chr.12: 52811054-52811054 on Build GRCh38
rs749252597 Chr.12: 52810860-52810860 on Build GRCh38
rs998438301 Chr.12: 52811019-52811019 on Build GRCh38
rs768144088 Chr.12: 52810861-52810861 on Build GRCh38
rs774011441 Chr.12: 52810782-52810782 on Build GRCh38
rs773365930 Chr.12: 52810998-52810998 on Build GRCh38
rs894506890 Chr.12: 52810896-52810896 on Build GRCh38
rs372766731 Chr.12: 52810746-52810746 on Build GRCh38
rs10783539 Chr.12: 52810973-52810973 on Build GRCh38
rs367879122 Chr.12: 52810925-52810925 on Build GRCh38
rs938468536 Chr.12: 52810703-52810703 on Build GRCh38
rs754240897 Chr.12: 52810795-52810795 on Build GRCh38
rs546100543 Chr.12: 52810783-52810783 on Build GRCh38
rs182844060 Chr.12: 52810825-52810825 on Build GRCh38
rs73306189 Chr.12: 52810819-52810819 on Build GRCh38
rs375835557 Chr.12: 52810720-52810720 on Build GRCh38
rs769721980 Chr.12: 52810754-52810754 on Build GRCh38
rs367660428 Chr.12: 52810751-52810751 on Build GRCh38
rs142604877 Chr.12: 52810856-52810856 on Build GRCh38
rs766448448 Chr.12: 52810879-52810881 on Build GRCh38
rs147271546 Chr.12: 52810747-52810747 on Build GRCh38
rs907293228 Chr.12: 52810913-52810913 on Build GRCh38
rs775442978 Chr.12: 52810765-52810765 on Build GRCh38
rs375353367 Chr.12: 52811055-52811055 on Build GRCh38
rs150595106 Chr.12: 52810889-52810889 on Build GRCh38
rs748107788 Chr.12: 52810873-52810873 on Build GRCh38
rs200221346 Chr.12: 52810834-52810834 on Build GRCh38
rs17119420 Chr.12: 52810740-52810740 on Build GRCh38
rs58438042 Chr.12: 52810894-52810894 on Build GRCh38
rs1038323114 Chr.12: 52810895-52810895 on Build GRCh38
rs202052680 Chr.12: 52810766-52810766 on Build GRCh38
rs934440110 Chr.12: 52810989-52810989 on Build GRCh38
rs887375384 Chr.12: 52810663-52810663 on Build GRCh38
rs771084245 Chr.12: 52810698-52810698 on Build GRCh38
rs371875636 Chr.12: 52810774-52810774 on Build GRCh38
rs1045793286 Chr.12: 52810908-52810908 on Build GRCh38
rs886049641 Chr.12: 52810768-52810768 on Build GRCh38
rs757064513 Chr.12: 52810624-52810624 on Build GRCh38
rs766683620 Chr.12: 52810717-52810717 on Build GRCh38
rs1036239131 Chr.12: 52810617-52810617 on Build GRCh38
rs535029412 Chr.12: 52810959-52810959 on Build GRCh38
rs1047289343 Chr.12: 52810629-52810629 on Build GRCh38
rs922365405 Chr.12: 52810878-52810878 on Build GRCh38
rs751217084 Chr.12: 52810707-52810707 on Build GRCh38
rs570829005 Chr.12: 52810957-52810957 on Build GRCh38
rs577345252 Chr.12: 52811009-52811009 on Build GRCh38
rs2307027 Chr.12: 52810830-52810830 on Build GRCh38
rs753280214 Chr.12: 52810823-52810823 on Build GRCh38
rs529778962 Chr.12: 52810833-52810833 on Build GRCh38
rs1013399696 Chr.12: 52810897-52810897 on Build GRCh38
rs1030879007 Chr.12: 52810926-52810926 on Build GRCh38
rs760019203 Chr.12: 52810809-52810809 on Build GRCh38
rs956603089 Chr.12: 52810969-52810969 on Build GRCh38
rs778005515 Chr.12: 52810739-52810739 on Build GRCh38
rs777722293 Chr.12: 52810826-52810826 on Build GRCh38
rs376461789 Chr.12: 52810748-52810748 on Build GRCh38
rs754302967 Chr.12: 52810719-52810719 on Build GRCh38
rs558891755 Chr.12: 52810940-52810940 on Build GRCh38
rs982909199 Chr.12: 52811075-52811075 on Build GRCh38
rs201529825 Chr.12: 52810839-52810839 on Build GRCh38
rs761374408 Chr.12: 52810787-52810787 on Build GRCh38
rs781098959 Chr.12: 52810633-52810633 on Build GRCh38

View Full Product Details