Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42455931-42456441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPCS
Entrez Gene ID
79717
Gene Name
phosphopantothenoylcysteine synthetase
Gene Aliases
-
UniGene
Hs.706662
Gene Chromosome Location
Chr.1: 42456341-42473381 on Build GRCh38
Transcripts
NM_001287506.1 NM_001287507.1 NM_001146192.1 NM_032257.4

Gene Symbol
ZMYND12
Entrez Gene ID
84217
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
UniGene
Hs.294009
Gene Chromosome Location
Chr.1: 42430329-42456267 on Build GRCh38
Transcripts
NM_001287506.1 NM_001287507.1 NM_001146192.1 NM_032257.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4742969 c.60C>T p.C20C Chr.1: 42455938-42455938 on Build GRCh38
4742970 c.20T>A p.L7Q Chr.1: 42455978-42455978 on Build GRCh38
4775616 c.9G>A p.V3V Chr.1: 42455989-42455989 on Build GRCh38
4913348 c.38G>A p.R13H Chr.1: 42455960-42455960 on Build GRCh38
4008046 c.25G>C p.V9L Chr.1: 42455973-42455973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373763715 Chr.1: 42456005-42456005 on Build GRCh38
rs759909447 Chr.1: 42455976-42455976 on Build GRCh38
rs917850052 Chr.1: 42456061-42456061 on Build GRCh38
rs746942511 Chr.1: 42455998-42455998 on Build GRCh38
rs1029245014 Chr.1: 42456410-42456410 on Build GRCh38
rs953067105 Chr.1: 42456203-42456203 on Build GRCh38
rs964468909 Chr.1: 42456218-42456218 on Build GRCh38
rs36121721 Chr.1: 42455953-42455953 on Build GRCh38
rs775975192 Chr.1: 42456027-42456027 on Build GRCh38
rs750703864 Chr.1: 42456045-42456045 on Build GRCh38
rs983027674 Chr.1: 42456251-42456251 on Build GRCh38
rs1049010796 Chr.1: 42456285-42456285 on Build GRCh38
rs755855113 Chr.1: 42456334-42456334 on Build GRCh38
rs753178264 Chr.1: 42455978-42455978 on Build GRCh38
rs1011433542 Chr.1: 42456190-42456190 on Build GRCh38
rs146773698 Chr.1: 42456239-42456239 on Build GRCh38
rs997760870 Chr.1: 42456170-42456170 on Build GRCh38
rs560176465 Chr.1: 42456315-42456315 on Build GRCh38
rs763497713 Chr.1: 42456114-42456114 on Build GRCh38
rs772858181 Chr.1: 42456015-42456015 on Build GRCh38
rs776156965 Chr.1: 42455966-42455966 on Build GRCh38
rs571194725 Chr.1: 42456109-42456109 on Build GRCh38
rs371699218 Chr.1: 42456028-42456028 on Build GRCh38
rs1036512077 Chr.1: 42456360-42456361 on Build GRCh38
rs767141433 Chr.1: 42456115-42456115 on Build GRCh38
rs772839967 Chr.1: 42455973-42455973 on Build GRCh38
rs765610907 Chr.1: 42455991-42455991 on Build GRCh38
rs113152212 Chr.1: 42455988-42455988 on Build GRCh38
rs1015126921 Chr.1: 42455967-42455967 on Build GRCh38
rs764724021 Chr.1: 42456031-42456031 on Build GRCh38
rs1045686595 Chr.1: 42456079-42456079 on Build GRCh38
rs74068443 Chr.1: 42456331-42456331 on Build GRCh38
rs941509985 Chr.1: 42456267-42456267 on Build GRCh38
rs908816844 Chr.1: 42456259-42456259 on Build GRCh38
rs753305531 Chr.1: 42456036-42456036 on Build GRCh38
rs779856286 Chr.1: 42455997-42455997 on Build GRCh38
rs540251372 Chr.1: 42456307-42456307 on Build GRCh38
rs766487286 Chr.1: 42456049-42456049 on Build GRCh38
rs979632060 Chr.1: 42456208-42456208 on Build GRCh38
rs975744786 Chr.1: 42455975-42455975 on Build GRCh38
rs764490300 Chr.1: 42455983-42455983 on Build GRCh38
rs1045803304 Chr.1: 42456320-42456320 on Build GRCh38
rs771976053 Chr.1: 42455952-42455952 on Build GRCh38
rs536418458 Chr.1: 42456238-42456238 on Build GRCh38
rs140060376 Chr.1: 42456009-42456009 on Build GRCh38
rs1046164210 Chr.1: 42456087-42456087 on Build GRCh38
rs16829550 Chr.1: 42456146-42456146 on Build GRCh38
rs1017867066 Chr.1: 42456198-42456198 on Build GRCh38
rs139504039 Chr.1: 42456246-42456246 on Build GRCh38
rs916083732 Chr.1: 42456304-42456304 on Build GRCh38
rs567391719 Chr.1: 42456206-42456206 on Build GRCh38
rs577566857 Chr.1: 42456301-42456301 on Build GRCh38
rs184134333 Chr.1: 42456008-42456008 on Build GRCh38
rs768081300 Chr.1: 42455963-42455963 on Build GRCh38
rs200378324 Chr.1: 42455970-42455970 on Build GRCh38
rs746894765 Chr.1: 42455961-42455961 on Build GRCh38
rs887984803 Chr.1: 42456122-42456122 on Build GRCh38
rs765515281 Chr.1: 42455969-42455969 on Build GRCh38
rs114459249 Chr.1: 42456022-42456022 on Build GRCh38
rs769238815 Chr.1: 42456014-42456014 on Build GRCh38
rs533780369 Chr.1: 42456128-42456128 on Build GRCh38
rs975831969 Chr.1: 42456232-42456232 on Build GRCh38
rs1015012368 Chr.1: 42456144-42456144 on Build GRCh38
rs112759046 Chr.1: 42456363-42456363 on Build GRCh38
rs531939190 Chr.1: 42456093-42456093 on Build GRCh38
rs758596635 Chr.1: 42456047-42456047 on Build GRCh38
rs892958952 Chr.1: 42456158-42456158 on Build GRCh38
rs927319407 Chr.1: 42456250-42456250 on Build GRCh38
rs766545158 Chr.1: 42455987-42455993 on Build GRCh38
rs143590939 Chr.1: 42456138-42456138 on Build GRCh38
rs907206105 Chr.1: 42456323-42456323 on Build GRCh38
rs887870252 Chr.1: 42456293-42456293 on Build GRCh38
rs141503615 Chr.1: 42455979-42455979 on Build GRCh38
rs777261371 Chr.1: 42456035-42456035 on Build GRCh38
rs868784939 Chr.1: 42456222-42456222 on Build GRCh38
rs956411374 Chr.1: 42456235-42456235 on Build GRCh38
rs551354010 Chr.1: 42456103-42456103 on Build GRCh38
rs974241647 Chr.1: 42456277-42456277 on Build GRCh38
rs748902366 Chr.1: 42456019-42456019 on Build GRCh38
rs11544999 Chr.1: 42456057-42456057 on Build GRCh38
rs1013433966 Chr.1: 42456078-42456078 on Build GRCh38
rs1011926034 Chr.1: 42456390-42456390 on Build GRCh38
rs750612160 Chr.1: 42455992-42455992 on Build GRCh38
rs754007687 Chr.1: 42455984-42455984 on Build GRCh38
rs758130985 Chr.1: 42455996-42455996 on Build GRCh38
rs765339650 Chr.1: 42456044-42456044 on Build GRCh38
rs1017397030 Chr.1: 42456395-42456395 on Build GRCh38
rs767859654 Chr.1: 42455977-42455977 on Build GRCh38

View Full Product Details