Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110341873-110342351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM15
Entrez Gene ID
64783
Gene Name
RNA binding motif protein 15
Gene Aliases
OTT, OTT1, SPEN
UniGene
Hs.435947
Gene Chromosome Location
Chr.1: 110338426-110373005 on Build GRCh38
Transcripts
NM_001201545.1 NM_022768.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1626299 c.2814G>C p.K938N Chr.1: 110342219-110342219 on Build GRCh38
4020516 c.2592T>C p.P864P Chr.1: 110341997-110341997 on Build GRCh38
5754129 c.2600C>T p.S867F Chr.1: 110342005-110342005 on Build GRCh38
6408130 c.2578C>T p.L860F Chr.1: 110341983-110341983 on Build GRCh38
414192 c.2602G>A p.D868N Chr.1: 110342007-110342007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745906657 Chr.1: 110342306-110342306 on Build GRCh38
rs749686792 Chr.1: 110342050-110342050 on Build GRCh38
rs767111502 Chr.1: 110342129-110342129 on Build GRCh38
rs778454048 Chr.1: 110342286-110342286 on Build GRCh38
rs759348845 Chr.1: 110341910-110341910 on Build GRCh38
rs201342657 Chr.1: 110342004-110342004 on Build GRCh38
rs776722641 Chr.1: 110342171-110342171 on Build GRCh38
rs751066660 Chr.1: 110341941-110341941 on Build GRCh38
rs752763239 Chr.1: 110341996-110341996 on Build GRCh38
rs769989160 Chr.1: 110342177-110342177 on Build GRCh38
rs774305240 Chr.1: 110342243-110342243 on Build GRCh38
rs759011847 Chr.1: 110342146-110342146 on Build GRCh38
rs749890067 Chr.1: 110342272-110342272 on Build GRCh38
rs757792294 Chr.1: 110342141-110342141 on Build GRCh38
rs756902716 Chr.1: 110341954-110341954 on Build GRCh38
rs766095804 Chr.1: 110342278-110342278 on Build GRCh38
rs201356376 Chr.1: 110342013-110342013 on Build GRCh38
rs373833752 Chr.1: 110342017-110342017 on Build GRCh38
rs973794308 Chr.1: 110342105-110342105 on Build GRCh38
rs761237379 Chr.1: 110342264-110342264 on Build GRCh38
rs765014074 Chr.1: 110342057-110342057 on Build GRCh38
rs779571058 Chr.1: 110341992-110341992 on Build GRCh38
rs752590664 Chr.1: 110341929-110341929 on Build GRCh38
rs761033295 Chr.1: 110341892-110341892 on Build GRCh38
rs961312675 Chr.1: 110342018-110342018 on Build GRCh38
rs778263276 Chr.1: 110342014-110342014 on Build GRCh38
rs751119834 Chr.1: 110342101-110342101 on Build GRCh38
rs370076418 Chr.1: 110342312-110342313 on Build GRCh38
rs558788724 Chr.1: 110342318-110342318 on Build GRCh38
rs780669160 Chr.1: 110342235-110342237 on Build GRCh38
rs867268749 Chr.1: 110341944-110341944 on Build GRCh38
rs952022673 Chr.1: 110341940-110341940 on Build GRCh38
rs777205172 Chr.1: 110341938-110341938 on Build GRCh38
rs139425937 Chr.1: 110342162-110342162 on Build GRCh38
rs914995610 Chr.1: 110342210-110342210 on Build GRCh38
rs769028870 Chr.1: 110341970-110341970 on Build GRCh38
rs765107744 Chr.1: 110341914-110341914 on Build GRCh38
rs775418481 Chr.1: 110342064-110342064 on Build GRCh38
rs763989651 Chr.1: 110342084-110342084 on Build GRCh38
rs768367912 Chr.1: 110342016-110342018 on Build GRCh38
rs553807881 Chr.1: 110342036-110342036 on Build GRCh38
rs768377734 Chr.1: 110342321-110342321 on Build GRCh38
rs981967324 Chr.1: 110342049-110342049 on Build GRCh38
rs753611997 Chr.1: 110341907-110341907 on Build GRCh38
rs770052907 Chr.1: 110342312-110342312 on Build GRCh38
rs778398647 Chr.1: 110342147-110342147 on Build GRCh38
rs747483026 Chr.1: 110342293-110342293 on Build GRCh38
rs755314262 Chr.1: 110342133-110342133 on Build GRCh38
rs775306039 Chr.1: 110342194-110342194 on Build GRCh38
rs756742730 Chr.1: 110342108-110342108 on Build GRCh38
rs868333253 Chr.1: 110342034-110342034 on Build GRCh38
rs371641830 Chr.1: 110342138-110342138 on Build GRCh38
rs747077640 Chr.1: 110342157-110342157 on Build GRCh38
rs780875157 Chr.1: 110341964-110341964 on Build GRCh38
rs375647275 Chr.1: 110342035-110342035 on Build GRCh38
rs867127927 Chr.1: 110342134-110342134 on Build GRCh38
rs61787372 Chr.1: 110342281-110342281 on Build GRCh38
rs370658386 Chr.1: 110342302-110342302 on Build GRCh38
rs747663988 Chr.1: 110342306-110342307 on Build GRCh38
rs756389203 Chr.1: 110342268-110342268 on Build GRCh38
rs140939757 Chr.1: 110342208-110342208 on Build GRCh38
rs987665017 Chr.1: 110342056-110342056 on Build GRCh38
rs774173247 Chr.1: 110342325-110342325 on Build GRCh38
rs762946925 Chr.1: 110342066-110342066 on Build GRCh38
rs748516713 Chr.1: 110342142-110342142 on Build GRCh38
rs757295449 Chr.1: 110342298-110342298 on Build GRCh38
rs374557071 Chr.1: 110342276-110342276 on Build GRCh38
rs747078751 Chr.1: 110342022-110342022 on Build GRCh38
rs368503888 Chr.1: 110342285-110342285 on Build GRCh38
rs766616667 Chr.1: 110341898-110341898 on Build GRCh38
rs75961595 Chr.1: 110342008-110342008 on Build GRCh38
rs757906873 Chr.1: 110341931-110341931 on Build GRCh38
rs201641173 Chr.1: 110342063-110342063 on Build GRCh38
rs771104153 Chr.1: 110342027-110342027 on Build GRCh38
rs749113649 Chr.1: 110342315-110342315 on Build GRCh38
rs764040852 Chr.1: 110342225-110342225 on Build GRCh38
rs367883725 Chr.1: 110342313-110342313 on Build GRCh38
rs929323035 Chr.1: 110342125-110342125 on Build GRCh38
rs200033561 Chr.1: 110342117-110342117 on Build GRCh38

View Full Product Details