Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156939423-156939975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF11
Entrez Gene ID
9826
Gene Name
Rho guanine nucleotide exchange factor 11
Gene Aliases
GTRAP48, PDZ-RHOGEF
UniGene
Hs.516954
Gene Chromosome Location
Chr.1: 156934840-157046903 on Build GRCh38
Transcripts
NM_014784.3 NM_198236.2 XM_005245629.4 XM_005245633.4 XM_006711659.2 XM_006711660.3 XM_006711661.2 XM_006711662.2 XM_006711663.3 XM_006711665.3 XM_011510185.2 XM_011510186.2 XM_011510187.2 XM_011510189.2 XM_017002920.1 XM_017002921.1 XM_017002922.1 XM_017002923.1 XM_017002924.1 XM_017000459.1

Gene Symbol
LRRC71
Entrez Gene ID
149499
Gene Name
leucine rich repeat containing 71
Gene Aliases
C1orf92
UniGene
Hs.491734
Gene Chromosome Location
Chr.1: 156920608-156941208 on Build GRCh38
Transcripts
NM_014784.3 NM_198236.2 XM_005245629.4 XM_005245633.4 XM_006711659.2 XM_006711660.3 XM_006711661.2 XM_006711662.2 XM_006711663.3 XM_006711665.3 XM_011510185.2 XM_011510186.2 XM_011510187.2 XM_011510189.2 XM_017002920.1 XM_017002921.1 XM_017002922.1 XM_017002923.1 XM_017002924.1 XM_017000459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4222984 c.1954C>T p.P652S Chr.1: 156939818-156939818 on Build GRCh38
3749005 c.1869C>T p.N623N Chr.1: 156939903-156939903 on Build GRCh38
6121699 c.3854C>T p.S1285F Chr.1: 156939790-156939790 on Build GRCh38
5844021 c.3823A>G p.T1275A Chr.1: 156939821-156939821 on Build GRCh38
4841766 c.4045G>A p.E1349K Chr.1: 156939599-156939599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968849731 Chr.1: 156939621-156939621 on Build GRCh38
rs750242942 Chr.1: 156939713-156939713 on Build GRCh38
rs1031537229 Chr.1: 156939731-156939731 on Build GRCh38
rs1021637337 Chr.1: 156939598-156939598 on Build GRCh38
rs757172810 Chr.1: 156939849-156939849 on Build GRCh38
rs372348613 Chr.1: 156939828-156939828 on Build GRCh38
rs372980428 Chr.1: 156939833-156939833 on Build GRCh38
rs755203907 Chr.1: 156939635-156939635 on Build GRCh38
rs188240595 Chr.1: 156939892-156939892 on Build GRCh38
rs749845760 Chr.1: 156939659-156939659 on Build GRCh38
rs770477824 Chr.1: 156939524-156939524 on Build GRCh38
rs746153844 Chr.1: 156939758-156939758 on Build GRCh38
rs946796172 Chr.1: 156939805-156939805 on Build GRCh38
rs765057222 Chr.1: 156939711-156939711 on Build GRCh38
rs201769707 Chr.1: 156939593-156939593 on Build GRCh38
rs781314175 Chr.1: 156939639-156939639 on Build GRCh38
rs560465848 Chr.1: 156939617-156939617 on Build GRCh38
rs111558417 Chr.1: 156939797-156939797 on Build GRCh38
rs3818807 Chr.1: 156939511-156939511 on Build GRCh38
rs773202369 Chr.1: 156939818-156939818 on Build GRCh38
rs567471201 Chr.1: 156939704-156939704 on Build GRCh38
rs746692819 Chr.1: 156939765-156939765 on Build GRCh38
rs368977976 Chr.1: 156939458-156939468 on Build GRCh38
rs751003197 Chr.1: 156939954-156939956 on Build GRCh38
rs749276417 Chr.1: 156939898-156939898 on Build GRCh38
rs756264591 Chr.1: 156939657-156939657 on Build GRCh38
rs748302129 Chr.1: 156939656-156939656 on Build GRCh38
rs751773190 Chr.1: 156939627-156939627 on Build GRCh38
rs115935778 Chr.1: 156939484-156939484 on Build GRCh38
rs994711414 Chr.1: 156939494-156939494 on Build GRCh38
rs761796808 Chr.1: 156939798-156939798 on Build GRCh38
rs774019160 Chr.1: 156939548-156939548 on Build GRCh38
rs373576989 Chr.1: 156939929-156939929 on Build GRCh38
rs145452145 Chr.1: 156939863-156939863 on Build GRCh38
rs779197564 Chr.1: 156939665-156939665 on Build GRCh38
rs532507970 Chr.1: 156939534-156939534 on Build GRCh38
rs532988298 Chr.1: 156939880-156939880 on Build GRCh38
rs753788298 Chr.1: 156939596-156939596 on Build GRCh38
rs545151254 Chr.1: 156939832-156939832 on Build GRCh38
rs889141711 Chr.1: 156939522-156939522 on Build GRCh38
rs377475140 Chr.1: 156939703-156939703 on Build GRCh38
rs779230294 Chr.1: 156939841-156939841 on Build GRCh38
rs772500421 Chr.1: 156939762-156939762 on Build GRCh38
rs992766463 Chr.1: 156939752-156939752 on Build GRCh38
rs928003294 Chr.1: 156939707-156939707 on Build GRCh38
rs115785833 Chr.1: 156939446-156939446 on Build GRCh38
rs781169434 Chr.1: 156939505-156939505 on Build GRCh38
rs767035441 Chr.1: 156939553-156939553 on Build GRCh38
rs550659351 Chr.1: 156939560-156939560 on Build GRCh38
rs375929380 Chr.1: 156939938-156939938 on Build GRCh38
rs774348649 Chr.1: 156939911-156939911 on Build GRCh38
rs757766993 Chr.1: 156939755-156939755 on Build GRCh38
rs959605895 Chr.1: 156939732-156939732 on Build GRCh38
rs776926403 Chr.1: 156939939-156939939 on Build GRCh38
rs771688817 Chr.1: 156939680-156939680 on Build GRCh38
rs375950621 Chr.1: 156939844-156939844 on Build GRCh38
rs767565368 Chr.1: 156939735-156939735 on Build GRCh38
rs762594137 Chr.1: 156939824-156939824 on Build GRCh38
rs772970922 Chr.1: 156939895-156939895 on Build GRCh38
rs746792104 Chr.1: 156939634-156939634 on Build GRCh38
rs765666978 Chr.1: 156939675-156939680 on Build GRCh38
rs1036932379 Chr.1: 156939899-156939899 on Build GRCh38
rs201030188 Chr.1: 156939678-156939678 on Build GRCh38
rs112685506 Chr.1: 156939454-156939464 on Build GRCh38
rs202191925 Chr.1: 156939827-156939827 on Build GRCh38
rs565426652 Chr.1: 156939658-156939658 on Build GRCh38
rs756211464 Chr.1: 156939745-156939745 on Build GRCh38
rs759156231 Chr.1: 156939550-156939550 on Build GRCh38
rs113239140 Chr.1: 156939685-156939685 on Build GRCh38
rs201904823 Chr.1: 156939532-156939532 on Build GRCh38
rs775129678 Chr.1: 156939682-156939682 on Build GRCh38
rs778042772 Chr.1: 156939747-156939747 on Build GRCh38
rs368998920 Chr.1: 156939944-156939944 on Build GRCh38
rs768179545 Chr.1: 156939686-156939686 on Build GRCh38
rs933557740 Chr.1: 156939923-156939923 on Build GRCh38
rs754042311 Chr.1: 156939835-156939835 on Build GRCh38
rs763954751 Chr.1: 156939586-156939586 on Build GRCh38
rs1017395055 Chr.1: 156939486-156939486 on Build GRCh38
rs752977682 Chr.1: 156939741-156939741 on Build GRCh38
rs963105303 Chr.1: 156939489-156939489 on Build GRCh38
rs370742614 Chr.1: 156939555-156939555 on Build GRCh38
rs780675400 Chr.1: 156939888-156939888 on Build GRCh38
rs771432445 Chr.1: 156939541-156939541 on Build GRCh38
rs1007586698 Chr.1: 156939594-156939594 on Build GRCh38
rs759795645 Chr.1: 156939733-156939733 on Build GRCh38
rs200157601 Chr.1: 156939738-156939738 on Build GRCh38
rs2275199 Chr.1: 156939903-156939903 on Build GRCh38
rs761894668 Chr.1: 156939537-156939540 on Build GRCh38
rs757362456 Chr.1: 156939836-156939836 on Build GRCh38
rs760602366 Chr.1: 156939562-156939562 on Build GRCh38
rs779635724 Chr.1: 156939763-156939763 on Build GRCh38
rs950671086 Chr.1: 156939529-156939529 on Build GRCh38
rs149737746 Chr.1: 156939748-156939748 on Build GRCh38
rs552902806 Chr.1: 156939769-156939769 on Build GRCh38
rs375668144 Chr.1: 156939812-156939812 on Build GRCh38
rs771298886 Chr.1: 156939662-156939662 on Build GRCh38
rs568941052 Chr.1: 156939599-156939599 on Build GRCh38
rs754683030 Chr.1: 156939884-156939884 on Build GRCh38

View Full Product Details