Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49903584-49904176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA2
Entrez Gene ID
9825
Gene Name
spermatogenesis associated 2
Gene Aliases
PD1, PPP1R145, tamo
UniGene
Hs.48513
Gene Chromosome Location
Chr.20: 49903391-49915543 on Build GRCh38
Transcripts
NM_001135773.1 NM_006038.3 XM_006723894.1 XM_011529116.1

SNP Detail

SNP ID Chromosome Location
rs954450708 Chr.20: 49904078-49904078 on Build GRCh38
rs922995262 Chr.20: 49903690-49903693 on Build GRCh38
rs1057069745 Chr.20: 49903615-49903615 on Build GRCh38
rs992676551 Chr.20: 49904143-49904143 on Build GRCh38
rs927533684 Chr.20: 49904151-49904151 on Build GRCh38
rs569208557 Chr.20: 49903951-49903952 on Build GRCh38
rs1026109293 Chr.20: 49903948-49903948 on Build GRCh38
rs1048953410 Chr.20: 49903932-49903932 on Build GRCh38
rs75624022 Chr.20: 49903623-49903623 on Build GRCh38
rs1019812086 Chr.20: 49903759-49903759 on Build GRCh38
rs6125806 Chr.20: 49903924-49903924 on Build GRCh38
rs868158049 Chr.20: 49903930-49903930 on Build GRCh38
rs6020121 Chr.20: 49903906-49903906 on Build GRCh38
rs1034500311 Chr.20: 49904140-49904140 on Build GRCh38
rs879570309 Chr.20: 49903904-49903905 on Build GRCh38
rs950645690 Chr.20: 49903632-49903633 on Build GRCh38
rs536371326 Chr.20: 49903659-49903659 on Build GRCh38
rs889982436 Chr.20: 49903951-49903951 on Build GRCh38
rs374830291 Chr.20: 49903903-49903924 on Build GRCh38
rs937321596 Chr.20: 49903847-49903847 on Build GRCh38
rs927189314 Chr.20: 49903850-49903850 on Build GRCh38
rs1020344450 Chr.20: 49903787-49903787 on Build GRCh38
rs1007131509 Chr.20: 49903737-49903737 on Build GRCh38
rs867092250 Chr.20: 49903953-49903953 on Build GRCh38
rs958549295 Chr.20: 49904142-49904142 on Build GRCh38
rs937147663 Chr.20: 49903882-49903882 on Build GRCh38
rs564621766 Chr.20: 49903772-49903772 on Build GRCh38
rs865848062 Chr.20: 49903928-49903928 on Build GRCh38
rs1019953844 Chr.20: 49904082-49904082 on Build GRCh38
rs1016585744 Chr.20: 49904015-49904015 on Build GRCh38
rs913368020 Chr.20: 49903740-49903740 on Build GRCh38
rs1011940029 Chr.20: 49903926-49903926 on Build GRCh38
rs750783039 Chr.20: 49904014-49904014 on Build GRCh38
rs1027162048 Chr.20: 49903718-49903718 on Build GRCh38
rs564109820 Chr.20: 49903662-49903662 on Build GRCh38
rs1015903241 Chr.20: 49903910-49903910 on Build GRCh38
rs995967037 Chr.20: 49903681-49903681 on Build GRCh38
rs965039082 Chr.20: 49903777-49903777 on Build GRCh38
rs1043677998 Chr.20: 49903819-49903819 on Build GRCh38
rs886250265 Chr.20: 49903971-49903971 on Build GRCh38
rs757566207 Chr.20: 49903661-49903661 on Build GRCh38
rs933512817 Chr.20: 49903711-49903711 on Build GRCh38
rs191126909 Chr.20: 49903658-49903658 on Build GRCh38
rs76401661 Chr.20: 49903835-49903835 on Build GRCh38
rs897256468 Chr.20: 49903671-49903671 on Build GRCh38
rs367694314 Chr.20: 49903761-49903761 on Build GRCh38
rs375814885 Chr.20: 49903638-49903639 on Build GRCh38
rs1019717687 Chr.20: 49904032-49904032 on Build GRCh38
rs895897744 Chr.20: 49903899-49903899 on Build GRCh38
rs978880362 Chr.20: 49903845-49903845 on Build GRCh38
rs1006995619 Chr.20: 49903954-49903954 on Build GRCh38
rs889911882 Chr.20: 49903731-49903731 on Build GRCh38
rs549355184 Chr.20: 49903707-49903707 on Build GRCh38
rs632376 Chr.20: 49904073-49904073 on Build GRCh38
rs7343604 Chr.20: 49903902-49903902 on Build GRCh38
rs1057185505 Chr.20: 49903867-49903867 on Build GRCh38
rs917151099 Chr.20: 49903923-49903923 on Build GRCh38
rs13038678 Chr.20: 49903982-49903982 on Build GRCh38
rs999984545 Chr.20: 49904122-49904122 on Build GRCh38
rs1006152774 Chr.20: 49904013-49904013 on Build GRCh38
rs1013370503 Chr.20: 49903606-49903606 on Build GRCh38
rs181295587 Chr.20: 49903780-49903780 on Build GRCh38
rs964429860 Chr.20: 49904045-49904045 on Build GRCh38
rs189549010 Chr.20: 49904047-49904047 on Build GRCh38
rs985075745 Chr.20: 49903645-49903645 on Build GRCh38
rs779147326 Chr.20: 49903672-49903672 on Build GRCh38
rs968788992 Chr.20: 49904090-49904090 on Build GRCh38
rs539507835 Chr.20: 49903933-49903952 on Build GRCh38
rs868473538 Chr.20: 49903947-49903947 on Build GRCh38
rs567233644 Chr.20: 49903663-49903663 on Build GRCh38

View Full Product Details