Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45729215-45729668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN10
Entrez Gene ID
25814
Gene Name
ataxin 10
Gene Aliases
E46L, HUMEEP, SCA10
UniGene
Hs.475125
Gene Chromosome Location
Chr.22: 45671798-45845307 on Build GRCh38
Transcripts
NM_001167621.1 NM_013236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7128744 c.729-1G>A p.? Chr.22: 45729424-45729424 on Build GRCh38
6608679 c.729-8delT p.? Chr.22: 45729417-45729417 on Build GRCh38
7285145 c.753A>G p.I251M Chr.22: 45729449-45729449 on Build GRCh38
4104804 c.764C>T p.T255M Chr.22: 45729460-45729460 on Build GRCh38
4104805 c.797C>G p.P266R Chr.22: 45729493-45729493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368814580 Chr.22: 45729581-45729581 on Build GRCh38
rs771885827 Chr.22: 45729452-45729452 on Build GRCh38
rs937532232 Chr.22: 45729434-45729434 on Build GRCh38
rs763991549 Chr.22: 45729512-45729512 on Build GRCh38
rs769181505 Chr.22: 45729539-45729539 on Build GRCh38
rs905302960 Chr.22: 45729303-45729303 on Build GRCh38
rs756105988 Chr.22: 45729551-45729552 on Build GRCh38
rs745375792 Chr.22: 45729527-45729527 on Build GRCh38
rs148992434 Chr.22: 45729566-45729566 on Build GRCh38
rs763121141 Chr.22: 45729486-45729486 on Build GRCh38
rs377292388 Chr.22: 45729634-45729634 on Build GRCh38
rs138453547 Chr.22: 45729562-45729562 on Build GRCh38
rs765408301 Chr.22: 45729479-45729479 on Build GRCh38
rs777263532 Chr.22: 45729464-45729464 on Build GRCh38
rs568068318 Chr.22: 45729478-45729478 on Build GRCh38
rs745873539 Chr.22: 45729633-45729633 on Build GRCh38
rs749939716 Chr.22: 45729617-45729617 on Build GRCh38
rs773185560 Chr.22: 45729484-45729484 on Build GRCh38
rs893766633 Chr.22: 45729328-45729330 on Build GRCh38
rs956404882 Chr.22: 45729319-45729319 on Build GRCh38
rs760443327 Chr.22: 45729583-45729583 on Build GRCh38
rs144613992 Chr.22: 45729416-45729416 on Build GRCh38
rs1032793819 Chr.22: 45729327-45729327 on Build GRCh38
rs954893372 Chr.22: 45729378-45729378 on Build GRCh38
rs994243932 Chr.22: 45729455-45729455 on Build GRCh38
rs752429212 Chr.22: 45729504-45729504 on Build GRCh38
rs746831904 Chr.22: 45729261-45729261 on Build GRCh38
rs758050698 Chr.22: 45729623-45729623 on Build GRCh38
rs1046213669 Chr.22: 45729265-45729265 on Build GRCh38
rs1057473358 Chr.22: 45729448-45729448 on Build GRCh38
rs763903931 Chr.22: 45729596-45729596 on Build GRCh38
rs200043904 Chr.22: 45729404-45729404 on Build GRCh38
rs369463744 Chr.22: 45729629-45729629 on Build GRCh38
rs376280245 Chr.22: 45729497-45729497 on Build GRCh38
rs988074604 Chr.22: 45729379-45729379 on Build GRCh38
rs761766866 Chr.22: 45729466-45729466 on Build GRCh38
rs987886768 Chr.22: 45729323-45729323 on Build GRCh38
rs375099471 Chr.22: 45729546-45729546 on Build GRCh38
rs757097542 Chr.22: 45729559-45729559 on Build GRCh38
rs745479298 Chr.22: 45729447-45729447 on Build GRCh38
rs946346630 Chr.22: 45729360-45729360 on Build GRCh38
rs117607950 Chr.22: 45729616-45729616 on Build GRCh38
rs368066531 Chr.22: 45729395-45729395 on Build GRCh38
rs914945318 Chr.22: 45729341-45729341 on Build GRCh38
rs756836344 Chr.22: 45729442-45729442 on Build GRCh38
rs141434930 Chr.22: 45729600-45729600 on Build GRCh38
rs747374366 Chr.22: 45729532-45729532 on Build GRCh38
rs149859910 Chr.22: 45729460-45729460 on Build GRCh38
rs202151442 Chr.22: 45729505-45729505 on Build GRCh38
rs373925397 Chr.22: 45729380-45729380 on Build GRCh38
rs145859375 Chr.22: 45729520-45729520 on Build GRCh38
rs1043333507 Chr.22: 45729370-45729370 on Build GRCh38
rs778412063 Chr.22: 45729444-45729444 on Build GRCh38
rs757155741 Chr.22: 45729612-45729612 on Build GRCh38
rs768152255 Chr.22: 45729551-45729551 on Build GRCh38
rs201298547 Chr.22: 45729558-45729558 on Build GRCh38
rs186780419 Chr.22: 45729582-45729582 on Build GRCh38
rs753866615 Chr.22: 45729519-45729519 on Build GRCh38
rs528589993 Chr.22: 45729413-45729413 on Build GRCh38
rs976964199 Chr.22: 45729239-45729239 on Build GRCh38
rs748731082 Chr.22: 45729428-45729428 on Build GRCh38
rs777697084 Chr.22: 45729427-45729427 on Build GRCh38
rs775931227 Chr.22: 45729454-45729454 on Build GRCh38
rs779910802 Chr.22: 45729531-45729531 on Build GRCh38
rs778449268 Chr.22: 45729521-45729521 on Build GRCh38
rs547165095 Chr.22: 45729425-45729425 on Build GRCh38
rs919601839 Chr.22: 45729500-45729500 on Build GRCh38
rs1021133966 Chr.22: 45729377-45729377 on Build GRCh38
rs577503398 Chr.22: 45729529-45729529 on Build GRCh38
rs922385417 Chr.22: 45729247-45729247 on Build GRCh38
rs1009782450 Chr.22: 45729363-45729363 on Build GRCh38
rs548270176 Chr.22: 45729461-45729461 on Build GRCh38
rs949316184 Chr.22: 45729548-45729548 on Build GRCh38
rs776980825 Chr.22: 45729543-45729543 on Build GRCh38
rs750697028 Chr.22: 45729393-45729393 on Build GRCh38
rs532419065 Chr.22: 45729279-45729279 on Build GRCh38

View Full Product Details