Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31599216-31599793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUBPL
Entrez Gene ID
80224
Gene Name
nucleotide binding protein like
Gene Aliases
C14orf127, IND1, huInd1
UniGene
Hs.288981
Gene Chromosome Location
Chr.14: 31561385-31861293 on Build GRCh38
Transcripts
NM_001201573.1 NM_025152.2 NR_120408.1 XM_011537181.2 XM_011537182.2 XM_011537183.2 XM_017021664.1 XM_017021665.1 XM_017021666.1 XM_017021667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574372 c.292+4A>T p.? Chr.14: 31599383-31599383 on Build GRCh38
4828141 c.316G>A p.V106M Chr.14: 31599313-31599313 on Build GRCh38
6731028 c.203C>A p.S68Y Chr.14: 31599290-31599290 on Build GRCh38
5848020 c.242C>T p.P81L Chr.14: 31599329-31599329 on Build GRCh38
5848019 c.332C>T p.P111L Chr.14: 31599329-31599329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056196909 Chr.14: 31599547-31599547 on Build GRCh38
rs748005300 Chr.14: 31599409-31599410 on Build GRCh38
rs747621208 Chr.14: 31599277-31599277 on Build GRCh38
rs376146035 Chr.14: 31599265-31599265 on Build GRCh38
rs762037874 Chr.14: 31599247-31599247 on Build GRCh38
rs769715753 Chr.14: 31599416-31599416 on Build GRCh38
rs397515440 Chr.14: 31599310-31599310 on Build GRCh38
rs774666357 Chr.14: 31599436-31599436 on Build GRCh38
rs749051189 Chr.14: 31599299-31599299 on Build GRCh38
rs748901975 Chr.14: 31599398-31599398 on Build GRCh38
rs777354820 Chr.14: 31599298-31599298 on Build GRCh38
rs979026893 Chr.14: 31599340-31599340 on Build GRCh38
rs992540043 Chr.14: 31599650-31599653 on Build GRCh38
rs944436446 Chr.14: 31599419-31599419 on Build GRCh38
rs776701145 Chr.14: 31599733-31599733 on Build GRCh38
rs769802615 Chr.14: 31599429-31599432 on Build GRCh38
rs930456783 Chr.14: 31599479-31599479 on Build GRCh38
rs771883848 Chr.14: 31599404-31599404 on Build GRCh38
rs748239635 Chr.14: 31599414-31599414 on Build GRCh38
rs996896251 Chr.14: 31599428-31599428 on Build GRCh38
rs1000329386 Chr.14: 31599589-31599589 on Build GRCh38
rs1033707531 Chr.14: 31599577-31599577 on Build GRCh38
rs949928537 Chr.14: 31599338-31599338 on Build GRCh38
rs772046824 Chr.14: 31599412-31599412 on Build GRCh38
rs1038148597 Chr.14: 31599750-31599750 on Build GRCh38
rs1020357302 Chr.14: 31599565-31599565 on Build GRCh38
rs949967336 Chr.14: 31599700-31599701 on Build GRCh38
rs775333603 Chr.14: 31599413-31599413 on Build GRCh38
rs765439746 Chr.14: 31599248-31599248 on Build GRCh38
rs941054113 Chr.14: 31599391-31599391 on Build GRCh38
rs778622069 Chr.14: 31599408-31599408 on Build GRCh38
rs773620793 Chr.14: 31599346-31599346 on Build GRCh38
rs775217395 Chr.14: 31599667-31599667 on Build GRCh38
rs1014360414 Chr.14: 31599560-31599560 on Build GRCh38
rs578191364 Chr.14: 31599700-31599704 on Build GRCh38
rs919106590 Chr.14: 31599463-31599463 on Build GRCh38
rs777457120 Chr.14: 31599382-31599382 on Build GRCh38
rs761869095 Chr.14: 31599332-31599332 on Build GRCh38
rs879277032 Chr.14: 31599644-31599644 on Build GRCh38
rs767966643 Chr.14: 31599379-31599379 on Build GRCh38
rs563071048 Chr.14: 31599636-31599636 on Build GRCh38
rs762323021 Chr.14: 31599694-31599694 on Build GRCh38
rs745892703 Chr.14: 31599265-31599267 on Build GRCh38
rs766807203 Chr.14: 31599353-31599353 on Build GRCh38
rs895928610 Chr.14: 31599552-31599552 on Build GRCh38
rs971902299 Chr.14: 31599441-31599441 on Build GRCh38
rs148107183 Chr.14: 31599508-31599508 on Build GRCh38
rs758877778 Chr.14: 31599260-31599260 on Build GRCh38
rs763968179 Chr.14: 31599720-31599720 on Build GRCh38
rs755482148 Chr.14: 31599366-31599366 on Build GRCh38
rs201430951 Chr.14: 31599308-31599308 on Build GRCh38
rs772038478 Chr.14: 31599277-31599278 on Build GRCh38
rs753868992 Chr.14: 31599242-31599242 on Build GRCh38
rs750799089 Chr.14: 31599258-31599258 on Build GRCh38
rs745740670 Chr.14: 31599311-31599311 on Build GRCh38
rs903353589 Chr.14: 31599575-31599575 on Build GRCh38
rs979526818 Chr.14: 31599576-31599576 on Build GRCh38
rs190646587 Chr.14: 31599603-31599603 on Build GRCh38
rs765470010 Chr.14: 31599342-31599342 on Build GRCh38
rs187351005 Chr.14: 31599502-31599502 on Build GRCh38
rs965656676 Chr.14: 31599710-31599710 on Build GRCh38
rs760560491 Chr.14: 31599676-31599676 on Build GRCh38
rs755569494 Chr.14: 31599267-31599267 on Build GRCh38
rs745720385 Chr.14: 31599411-31599411 on Build GRCh38
rs763292714 Chr.14: 31599347-31599347 on Build GRCh38
rs863224125 Chr.14: 31599373-31599373 on Build GRCh38
rs745333245 Chr.14: 31599365-31599365 on Build GRCh38
rs755777861 Chr.14: 31599297-31599297 on Build GRCh38
rs1020450662 Chr.14: 31599274-31599274 on Build GRCh38
rs1021047203 Chr.14: 31599543-31599543 on Build GRCh38
rs755618596 Chr.14: 31599380-31599380 on Build GRCh38
rs776758619 Chr.14: 31599321-31599321 on Build GRCh38
rs554314704 Chr.14: 31599243-31599243 on Build GRCh38
rs182466765 Chr.14: 31599684-31599684 on Build GRCh38
rs775690714 Chr.14: 31599400-31599402 on Build GRCh38
rs991445609 Chr.14: 31599264-31599264 on Build GRCh38
rs937393360 Chr.14: 31599531-31599531 on Build GRCh38
rs773455069 Chr.14: 31599431-31599431 on Build GRCh38
rs780682882 Chr.14: 31599273-31599273 on Build GRCh38
rs771917774 Chr.14: 31599315-31599315 on Build GRCh38
rs1029939949 Chr.14: 31599444-31599444 on Build GRCh38
rs56188755 Chr.14: 31599700-31599700 on Build GRCh38

View Full Product Details