Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103710926-103711518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC107984648
Entrez Gene ID
107984648
Gene Name
uncharacterized LOC107984648
Gene Aliases
-
UniGene
Hs.733412
Gene Chromosome Location
Chr.14: 103711245-103712868 on Build GRCh38
Transcripts
XM_017021849.1 NM_001100118.1 NM_001100119.1 NM_005432.3 XM_005268046.2 XM_011537138.2

Gene Symbol
XRCC3
Entrez Gene ID
7517
Gene Name
X-ray repair cross complementing 3
Gene Aliases
CMM6
UniGene
Hs.592325
Gene Chromosome Location
Chr.14: 103697611-103715486 on Build GRCh38
Transcripts
XM_017021849.1 NM_001100118.1 NM_001100119.1 NM_005432.3 XM_005268046.2 XM_011537138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116736 c.36T>G p.I12M Chr.14: 103711052-103711052 on Build GRCh38
4150726 c.61A>G p.K21E Chr.14: 103711423-103711423 on Build GRCh38
5037004 c.14A>C p.N5T Chr.14: 103711376-103711376 on Build GRCh38
6015632 c.96C>T p.P32P Chr.14: 103711458-103711458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778459700 Chr.14: 103711019-103711019 on Build GRCh38
rs900138898 Chr.14: 103711263-103711263 on Build GRCh38
rs61995795 Chr.14: 103711279-103711279 on Build GRCh38
rs764138700 Chr.14: 103711067-103711067 on Build GRCh38
rs978700071 Chr.14: 103711205-103711205 on Build GRCh38
rs1004713158 Chr.14: 103711343-103711343 on Build GRCh38
rs751635577 Chr.14: 103711069-103711069 on Build GRCh38
rs373587799 Chr.14: 103710960-103710960 on Build GRCh38
rs749766789 Chr.14: 103711006-103711006 on Build GRCh38
rs918023041 Chr.14: 103711143-103711143 on Build GRCh38
rs542685042 Chr.14: 103711425-103711425 on Build GRCh38
rs948325350 Chr.14: 103711487-103711487 on Build GRCh38
rs755493221 Chr.14: 103711126-103711126 on Build GRCh38
rs935627734 Chr.14: 103711165-103711165 on Build GRCh38
rs750792518 Chr.14: 103710995-103710995 on Build GRCh38
rs533312810 Chr.14: 103711444-103711444 on Build GRCh38
rs1047553801 Chr.14: 103711233-103711233 on Build GRCh38
rs777590068 Chr.14: 103711351-103711351 on Build GRCh38
rs370388004 Chr.14: 103711123-103711123 on Build GRCh38
rs990818314 Chr.14: 103711446-103711446 on Build GRCh38
rs948292788 Chr.14: 103711194-103711194 on Build GRCh38
rs750598578 Chr.14: 103711094-103711094 on Build GRCh38
rs768095504 Chr.14: 103711011-103711011 on Build GRCh38
rs1018276047 Chr.14: 103711243-103711243 on Build GRCh38
rs529025680 Chr.14: 103711344-103711344 on Build GRCh38
rs866773973 Chr.14: 103711220-103711220 on Build GRCh38
rs995140193 Chr.14: 103711402-103711402 on Build GRCh38
rs199632866 Chr.14: 103711473-103711473 on Build GRCh38
rs780451323 Chr.14: 103711005-103711005 on Build GRCh38
rs778350647 Chr.14: 103711132-103711132 on Build GRCh38
rs781604798 Chr.14: 103710994-103710994 on Build GRCh38
rs746619890 Chr.14: 103711364-103711364 on Build GRCh38
rs771648753 Chr.14: 103711326-103711326 on Build GRCh38
rs1034298334 Chr.14: 103711426-103711426 on Build GRCh38
rs949550766 Chr.14: 103711190-103711190 on Build GRCh38
rs746679221 Chr.14: 103710992-103710992 on Build GRCh38
rs1018163117 Chr.14: 103711007-103711007 on Build GRCh38
rs751777770 Chr.14: 103710991-103710991 on Build GRCh38
rs760558474 Chr.14: 103711031-103711031 on Build GRCh38
rs924140814 Chr.14: 103711213-103711213 on Build GRCh38
rs571569002 Chr.14: 103711492-103711492 on Build GRCh38
rs1014842828 Chr.14: 103711074-103711074 on Build GRCh38
rs56018633 Chr.14: 103711112-103711112 on Build GRCh38
rs551369420 Chr.14: 103711491-103711491 on Build GRCh38
rs770709968 Chr.14: 103711037-103711037 on Build GRCh38
rs373800330 Chr.14: 103711133-103711133 on Build GRCh38
rs1006002468 Chr.14: 103711234-103711234 on Build GRCh38
rs1055386143 Chr.14: 103711167-103711167 on Build GRCh38
rs1029515596 Chr.14: 103711318-103711318 on Build GRCh38
rs756415470 Chr.14: 103711003-103711003 on Build GRCh38
rs1025005386 Chr.14: 103711458-103711458 on Build GRCh38
rs995903699 Chr.14: 103710976-103710976 on Build GRCh38
rs977547501 Chr.14: 103711142-103711142 on Build GRCh38
rs528107761 Chr.14: 103711489-103711489 on Build GRCh38
rs1051493024 Chr.14: 103711295-103711295 on Build GRCh38
rs564381445 Chr.14: 103711490-103711490 on Build GRCh38
rs936878819 Chr.14: 103711225-103711225 on Build GRCh38
rs768454791 Chr.14: 103711030-103711030 on Build GRCh38
rs963660984 Chr.14: 103711085-103711085 on Build GRCh38
rs564460234 Chr.14: 103711183-103711183 on Build GRCh38
rs56314050 Chr.14: 103711309-103711309 on Build GRCh38
rs759485767 Chr.14: 103711061-103711061 on Build GRCh38
rs756505421 Chr.14: 103711108-103711108 on Build GRCh38
rs1054059081 Chr.14: 103711226-103711226 on Build GRCh38
rs776306927 Chr.14: 103711182-103711182 on Build GRCh38
rs376582164 Chr.14: 103711093-103711093 on Build GRCh38
rs56377012 Chr.14: 103711073-103711073 on Build GRCh38
rs774462522 Chr.14: 103711488-103711488 on Build GRCh38
rs563894235 Chr.14: 103711342-103711342 on Build GRCh38
rs776463599 Chr.14: 103711046-103711046 on Build GRCh38
rs577867073 Chr.14: 103711217-103711217 on Build GRCh38
rs771843136 Chr.14: 103711029-103711029 on Build GRCh38
rs75660546 Chr.14: 103711246-103711246 on Build GRCh38
rs1033216887 Chr.14: 103711408-103711408 on Build GRCh38
rs747858149 Chr.14: 103711028-103711028 on Build GRCh38

View Full Product Details