Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100648184-100648782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB7
Entrez Gene ID
140460
Gene Name
ankyrin repeat and SOCS box containing 7
Gene Aliases
-
UniGene
Hs.31845
Gene Chromosome Location
Chr.15: 100602550-100651701 on Build GRCh38
Transcripts
NM_198243.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4053447 c.867T>C p.C289C Chr.15: 100648372-100648372 on Build GRCh38
2200350 c.827G>A p.R276K Chr.15: 100648332-100648332 on Build GRCh38
959580 c.892C>T p.L298F Chr.15: 100648397-100648397 on Build GRCh38
959579 c.859C>T p.R287* Chr.15: 100648364-100648364 on Build GRCh38
6588756 c.954C>T p.I318I Chr.15: 100648459-100648459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770267756 Chr.15: 100648351-100648351 on Build GRCh38
rs74039473 Chr.15: 100648396-100648396 on Build GRCh38
rs966953568 Chr.15: 100648444-100648444 on Build GRCh38
rs118046496 Chr.15: 100648288-100648288 on Build GRCh38
rs111349661 Chr.15: 100648237-100648238 on Build GRCh38
rs772564230 Chr.15: 100648492-100648496 on Build GRCh38
rs28716928 Chr.15: 100648324-100648324 on Build GRCh38
rs184263340 Chr.15: 100648335-100648335 on Build GRCh38
rs560376901 Chr.15: 100648523-100648523 on Build GRCh38
rs377385107 Chr.15: 100648702-100648702 on Build GRCh38
rs1010978960 Chr.15: 100648516-100648516 on Build GRCh38
rs376501102 Chr.15: 100648223-100648223 on Build GRCh38
rs751732525 Chr.15: 100648452-100648452 on Build GRCh38
rs929229000 Chr.15: 100648666-100648666 on Build GRCh38
rs770216391 Chr.15: 100648681-100648681 on Build GRCh38
rs954201399 Chr.15: 100648637-100648637 on Build GRCh38
rs781590488 Chr.15: 100648460-100648460 on Build GRCh38
rs188895527 Chr.15: 100648531-100648531 on Build GRCh38
rs758427918 Chr.15: 100648433-100648433 on Build GRCh38
rs567825028 Chr.15: 100648669-100648669 on Build GRCh38
rs781537692 Chr.15: 100648542-100648542 on Build GRCh38
rs887820302 Chr.15: 100648727-100648727 on Build GRCh38
rs943194795 Chr.15: 100648261-100648261 on Build GRCh38
rs188412002 Chr.15: 100648224-100648224 on Build GRCh38
rs537744530 Chr.15: 100648508-100648508 on Build GRCh38
rs750239887 Chr.15: 100648412-100648412 on Build GRCh38
rs758732883 Chr.15: 100648279-100648279 on Build GRCh38
rs761260973 Chr.15: 100648748-100648748 on Build GRCh38
rs766387162 Chr.15: 100648420-100648420 on Build GRCh38
rs998329670 Chr.15: 100648598-100648601 on Build GRCh38
rs770329063 Chr.15: 100648465-100648465 on Build GRCh38
rs919161674 Chr.15: 100648390-100648390 on Build GRCh38
rs753785778 Chr.15: 100648425-100648425 on Build GRCh38
rs149517755 Chr.15: 100648408-100648408 on Build GRCh38
rs150230229 Chr.15: 100648373-100648373 on Build GRCh38
rs748216432 Chr.15: 100648274-100648274 on Build GRCh38
rs777887254 Chr.15: 100648285-100648285 on Build GRCh38
rs943098177 Chr.15: 100648557-100648557 on Build GRCh38
rs924310022 Chr.15: 100648466-100648466 on Build GRCh38
rs1055261890 Chr.15: 100648477-100648477 on Build GRCh38
rs572147712 Chr.15: 100648262-100648262 on Build GRCh38
rs371262240 Chr.15: 100648345-100648345 on Build GRCh38
rs746433010 Chr.15: 100648487-100648489 on Build GRCh38
rs749748915 Chr.15: 100648473-100648473 on Build GRCh38
rs762422274 Chr.15: 100648370-100648370 on Build GRCh38
rs901768396 Chr.15: 100648270-100648270 on Build GRCh38
rs12913166 Chr.15: 100648209-100648209 on Build GRCh38
rs888989192 Chr.15: 100648255-100648255 on Build GRCh38
rs574533283 Chr.15: 100648643-100648643 on Build GRCh38
rs746214212 Chr.15: 100648464-100648464 on Build GRCh38
rs368010823 Chr.15: 100648435-100648435 on Build GRCh38
rs902736135 Chr.15: 100648561-100648561 on Build GRCh38
rs746399671 Chr.15: 100648650-100648650 on Build GRCh38
rs181636288 Chr.15: 100648604-100648604 on Build GRCh38
rs757493272 Chr.15: 100648456-100648456 on Build GRCh38
rs375149482 Chr.15: 100648383-100648383 on Build GRCh38
rs558498445 Chr.15: 100648700-100648700 on Build GRCh38
rs778003836 Chr.15: 100648271-100648271 on Build GRCh38
rs929178178 Chr.15: 100648729-100648729 on Build GRCh38
rs775033832 Chr.15: 100648369-100648369 on Build GRCh38
rs560703767 Chr.15: 100648289-100648289 on Build GRCh38
rs941341220 Chr.15: 100648348-100648348 on Build GRCh38
rs533154753 Chr.15: 100648417-100648417 on Build GRCh38
rs1047663078 Chr.15: 100648673-100648673 on Build GRCh38
rs1017433165 Chr.15: 100648647-100648647 on Build GRCh38
rs577061223 Chr.15: 100648589-100648589 on Build GRCh38
rs181146719 Chr.15: 100648227-100648227 on Build GRCh38
rs973111683 Chr.15: 100648660-100648660 on Build GRCh38
rs895236318 Chr.15: 100648754-100648754 on Build GRCh38
rs773877900 Chr.15: 100648739-100648739 on Build GRCh38
rs12907140 Chr.15: 100648215-100648215 on Build GRCh38
rs551790926 Chr.15: 100648496-100648496 on Build GRCh38

View Full Product Details