Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15664575-15665151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM27
Entrez Gene ID
57393
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, NX17
UniGene
Hs.129614
Gene Chromosome Location
Chr.X: 15627316-15675624 on Build GRCh38
Transcripts
NM_020665.5 XM_017029680.1 XM_017029681.1

SNP Detail

SNP ID Chromosome Location
rs774111506 Chr.X: 15664754-15664754 on Build GRCh38
rs189753422 Chr.X: 15664633-15664633 on Build GRCh38
rs781004153 Chr.X: 15665027-15665027 on Build GRCh38
rs773727107 Chr.X: 15664742-15664742 on Build GRCh38
rs139660495 Chr.X: 15664980-15664980 on Build GRCh38
rs180868712 Chr.X: 15664885-15664885 on Build GRCh38
rs757129417 Chr.X: 15664804-15664804 on Build GRCh38
rs989906763 Chr.X: 15665076-15665076 on Build GRCh38
rs995544567 Chr.X: 15664859-15664859 on Build GRCh38
rs763021603 Chr.X: 15664934-15664934 on Build GRCh38
rs759778337 Chr.X: 15664824-15664824 on Build GRCh38
rs368563346 Chr.X: 15664760-15664760 on Build GRCh38
rs747101375 Chr.X: 15664655-15664655 on Build GRCh38
rs768909556 Chr.X: 15664704-15664704 on Build GRCh38
rs958154096 Chr.X: 15665028-15665028 on Build GRCh38
rs945660139 Chr.X: 15664730-15664730 on Build GRCh38
rs939895630 Chr.X: 15664860-15664860 on Build GRCh38
rs1054162452 Chr.X: 15664684-15664684 on Build GRCh38
rs781446459 Chr.X: 15664823-15664823 on Build GRCh38
rs935742077 Chr.X: 15664657-15664657 on Build GRCh38
rs868480022 Chr.X: 15664753-15664763 on Build GRCh38
rs1025652502 Chr.X: 15664864-15664864 on Build GRCh38
rs755882876 Chr.X: 15665096-15665096 on Build GRCh38
rs891013305 Chr.X: 15664795-15664795 on Build GRCh38
rs1006745966 Chr.X: 15664805-15664805 on Build GRCh38
rs898611808 Chr.X: 15664846-15664846 on Build GRCh38
rs1049861492 Chr.X: 15664999-15664999 on Build GRCh38
rs758937659 Chr.X: 15664755-15664755 on Build GRCh38
rs762764857 Chr.X: 15664807-15664808 on Build GRCh38
rs770238468 Chr.X: 15664741-15664741 on Build GRCh38
rs1011755204 Chr.X: 15665119-15665119 on Build GRCh38
rs201844811 Chr.X: 15664763-15664763 on Build GRCh38
rs1025250864 Chr.X: 15664901-15664901 on Build GRCh38
rs891192772 Chr.X: 15665018-15665018 on Build GRCh38
rs763808409 Chr.X: 15664783-15664783 on Build GRCh38
rs898301700 Chr.X: 15664951-15664951 on Build GRCh38
rs753569016 Chr.X: 15664788-15664788 on Build GRCh38
rs1056526965 Chr.X: 15664830-15664830 on Build GRCh38
rs763825926 Chr.X: 15664808-15664809 on Build GRCh38
rs370084733 Chr.X: 15664690-15664690 on Build GRCh38
rs912955895 Chr.X: 15665084-15665084 on Build GRCh38
rs751086329 Chr.X: 15665022-15665022 on Build GRCh38
rs926348933 Chr.X: 15664819-15664819 on Build GRCh38
rs747042827 Chr.X: 15664697-15664697 on Build GRCh38
rs964455163 Chr.X: 15664608-15664608 on Build GRCh38
rs765141540 Chr.X: 15664877-15664877 on Build GRCh38
rs780386458 Chr.X: 15664695-15664695 on Build GRCh38
rs769732293 Chr.X: 15664752-15664753 on Build GRCh38
rs184918998 Chr.X: 15664604-15664604 on Build GRCh38
rs771128575 Chr.X: 15664796-15664796 on Build GRCh38
rs16980061 Chr.X: 15665127-15665127 on Build GRCh38
rs759445205 Chr.X: 15664748-15664748 on Build GRCh38
rs751218884 Chr.X: 15664809-15664809 on Build GRCh38
rs1032087405 Chr.X: 15665077-15665077 on Build GRCh38
rs375643105 Chr.X: 15664751-15664751 on Build GRCh38
rs747926688 Chr.X: 15665081-15665081 on Build GRCh38
rs770486623 Chr.X: 15664752-15664752 on Build GRCh38
rs781757399 Chr.X: 15664719-15664719 on Build GRCh38
rs28422048 Chr.X: 15664603-15664603 on Build GRCh38
rs748698931 Chr.X: 15664738-15664738 on Build GRCh38
rs908496684 Chr.X: 15664840-15664840 on Build GRCh38
rs186056383 Chr.X: 15665066-15665066 on Build GRCh38
rs780121073 Chr.X: 15664808-15664808 on Build GRCh38
rs773315151 Chr.X: 15664726-15664727 on Build GRCh38
rs201874670 Chr.X: 15664819-15664819 on Build GRCh38

View Full Product Details