Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12919280-12919854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR8
Entrez Gene ID
51311
Gene Name
toll like receptor 8
Gene Aliases
CD288
UniGene
Hs.660543
Gene Chromosome Location
Chr.X: 12906620-12923169 on Build GRCh38
Transcripts
NM_016610.3 NM_138636.5 XM_011545529.1 XM_011545530.2 NR_030727.1

Gene Symbol
TLR8-AS1
Entrez Gene ID
349408
Gene Name
TLR8 antisense RNA 1
Gene Aliases
GS1-324M7.6
UniGene
Hs.685035
Gene Chromosome Location
Chr.X: 12902817-12943300 on Build GRCh38
Transcripts
NM_016610.3 NM_138636.5 XM_011545529.1 XM_011545530.2 NR_030727.1

SNP Detail

SNP ID Chromosome Location
rs139006461 Chr.X: 12919793-12919793 on Build GRCh38
rs761066050 Chr.X: 12919565-12919565 on Build GRCh38
rs779613268 Chr.X: 12919638-12919638 on Build GRCh38
rs767811815 Chr.X: 12919549-12919549 on Build GRCh38
rs868494026 Chr.X: 12919660-12919660 on Build GRCh38
rs779846666 Chr.X: 12919457-12919457 on Build GRCh38
rs774944401 Chr.X: 12919361-12919361 on Build GRCh38
rs146700498 Chr.X: 12919326-12919326 on Build GRCh38
rs576185229 Chr.X: 12919373-12919373 on Build GRCh38
rs768871999 Chr.X: 12919754-12919754 on Build GRCh38
rs750358899 Chr.X: 12919384-12919384 on Build GRCh38
rs772600126 Chr.X: 12919427-12919427 on Build GRCh38
rs749440648 Chr.X: 12919526-12919526 on Build GRCh38
rs376689229 Chr.X: 12919512-12919512 on Build GRCh38
rs757616604 Chr.X: 12919713-12919713 on Build GRCh38
rs878907333 Chr.X: 12919364-12919364 on Build GRCh38
rs146163865 Chr.X: 12919751-12919751 on Build GRCh38
rs776057581 Chr.X: 12919600-12919600 on Build GRCh38
rs994032756 Chr.X: 12919337-12919337 on Build GRCh38
rs2159377 Chr.X: 12919394-12919394 on Build GRCh38
rs775845314 Chr.X: 12919742-12919742 on Build GRCh38
rs745489278 Chr.X: 12919422-12919422 on Build GRCh38
rs752683730 Chr.X: 12919567-12919567 on Build GRCh38
rs778696383 Chr.X: 12919406-12919406 on Build GRCh38
rs749370882 Chr.X: 12919301-12919301 on Build GRCh38
rs749938444 Chr.X: 12919802-12919802 on Build GRCh38
rs373184358 Chr.X: 12919352-12919352 on Build GRCh38
rs5744079 Chr.X: 12919601-12919601 on Build GRCh38
rs889236034 Chr.X: 12919714-12919714 on Build GRCh38
rs781255488 Chr.X: 12919676-12919676 on Build GRCh38
rs5744080 Chr.X: 12919685-12919685 on Build GRCh38
rs200981848 Chr.X: 12919339-12919339 on Build GRCh38
rs183801068 Chr.X: 12919532-12919532 on Build GRCh38
rs776006574 Chr.X: 12919554-12919554 on Build GRCh38
rs771088322 Chr.X: 12919693-12919693 on Build GRCh38
rs774608026 Chr.X: 12919696-12919696 on Build GRCh38
rs759806873 Chr.X: 12919535-12919535 on Build GRCh38
rs756834432 Chr.X: 12919589-12919589 on Build GRCh38
rs976964102 Chr.X: 12919636-12919636 on Build GRCh38
rs934606168 Chr.X: 12919616-12919616 on Build GRCh38
rs750589747 Chr.X: 12919688-12919689 on Build GRCh38
rs762250935 Chr.X: 12919776-12919776 on Build GRCh38
rs374687057 Chr.X: 12919452-12919452 on Build GRCh38
rs773370542 Chr.X: 12919311-12919311 on Build GRCh38
rs148046494 Chr.X: 12919637-12919637 on Build GRCh38
rs762362421 Chr.X: 12919519-12919519 on Build GRCh38
rs768640826 Chr.X: 12919497-12919497 on Build GRCh38
rs777245589 Chr.X: 12919773-12919773 on Build GRCh38
rs754297567 Chr.X: 12919585-12919585 on Build GRCh38
rs772440962 Chr.X: 12919737-12919737 on Build GRCh38
rs942023200 Chr.X: 12919715-12919715 on Build GRCh38
rs915691659 Chr.X: 12919353-12919353 on Build GRCh38
rs746819165 Chr.X: 12919640-12919640 on Build GRCh38
rs746766277 Chr.X: 12919490-12919490 on Build GRCh38
rs1035870242 Chr.X: 12919338-12919338 on Build GRCh38
rs754712471 Chr.X: 12919644-12919644 on Build GRCh38
rs955257968 Chr.X: 12919400-12919400 on Build GRCh38
rs932494695 Chr.X: 12919645-12919645 on Build GRCh38
rs764443515 Chr.X: 12919382-12919382 on Build GRCh38
rs376034872 Chr.X: 12919602-12919602 on Build GRCh38

View Full Product Details