Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54750639-54751202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH6
Entrez Gene ID
347365
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Gene Aliases
ITIH5L, UNQ6369, dJ14O9.1
UniGene
Hs.454272
Gene Chromosome Location
Chr.X: 54748899-54798240 on Build GRCh38
Transcripts
NM_198510.2

SNP Detail

SNP ID Chromosome Location
rs763669704 Chr.X: 54751141-54751141 on Build GRCh38
rs77585128 Chr.X: 54750953-54750953 on Build GRCh38
rs747237077 Chr.X: 54750865-54750865 on Build GRCh38
rs755739603 Chr.X: 54751063-54751063 on Build GRCh38
rs769797963 Chr.X: 54750992-54750992 on Build GRCh38
rs774235260 Chr.X: 54751110-54751110 on Build GRCh38
rs902508439 Chr.X: 54750772-54750772 on Build GRCh38
rs1032114879 Chr.X: 54750719-54750719 on Build GRCh38
rs1014707549 Chr.X: 54750906-54750906 on Build GRCh38
rs770993032 Chr.X: 54751120-54751120 on Build GRCh38
rs1056493437 Chr.X: 54751098-54751098 on Build GRCh38
rs758446366 Chr.X: 54750827-54750827 on Build GRCh38
rs769942710 Chr.X: 54751102-54751102 on Build GRCh38
rs374595010 Chr.X: 54750771-54750771 on Build GRCh38
rs909998215 Chr.X: 54751012-54751012 on Build GRCh38
rs775175752 Chr.X: 54751134-54751134 on Build GRCh38
rs142183856 Chr.X: 54751014-54751014 on Build GRCh38
rs779162145 Chr.X: 54750714-54750714 on Build GRCh38
rs201616304 Chr.X: 54751027-54751027 on Build GRCh38
rs370438285 Chr.X: 54750713-54750713 on Build GRCh38
rs758502825 Chr.X: 54750768-54750768 on Build GRCh38
rs368352151 Chr.X: 54750961-54750961 on Build GRCh38
rs201473014 Chr.X: 54751049-54751049 on Build GRCh38
rs371340681 Chr.X: 54751146-54751146 on Build GRCh38
rs1003586170 Chr.X: 54750796-54750796 on Build GRCh38
rs1042506642 Chr.X: 54750738-54750738 on Build GRCh38
rs149957596 Chr.X: 54751142-54751142 on Build GRCh38
rs781390419 Chr.X: 54751096-54751096 on Build GRCh38
rs945936029 Chr.X: 54751046-54751046 on Build GRCh38
rs375393223 Chr.X: 54751095-54751095 on Build GRCh38
rs775058785 Chr.X: 54751133-54751133 on Build GRCh38
rs755565017 Chr.X: 54751181-54751181 on Build GRCh38
rs781696686 Chr.X: 54751183-54751183 on Build GRCh38
rs753288851 Chr.X: 54751064-54751064 on Build GRCh38
rs975721497 Chr.X: 54750936-54750936 on Build GRCh38
rs756898377 Chr.X: 54751144-54751144 on Build GRCh38
rs965679713 Chr.X: 54750919-54750919 on Build GRCh38
rs773189468 Chr.X: 54751044-54751044 on Build GRCh38
rs759392609 Chr.X: 54751051-54751051 on Build GRCh38
rs183915186 Chr.X: 54750826-54750826 on Build GRCh38
rs150775337 Chr.X: 54751053-54751053 on Build GRCh38
rs756773471 Chr.X: 54751066-54751066 on Build GRCh38
rs200494632 Chr.X: 54751122-54751122 on Build GRCh38
rs1054702998 Chr.X: 54750672-54750672 on Build GRCh38
rs954228138 Chr.X: 54750889-54750889 on Build GRCh38
rs1028258752 Chr.X: 54750841-54750841 on Build GRCh38
rs747517641 Chr.X: 54751094-54751094 on Build GRCh38
rs768920518 Chr.X: 54750950-54750950 on Build GRCh38
rs200075050 Chr.X: 54751152-54751152 on Build GRCh38
rs769966096 Chr.X: 54751039-54751039 on Build GRCh38
rs532806724 Chr.X: 54750926-54750926 on Build GRCh38
rs919161637 Chr.X: 54751060-54751060 on Build GRCh38
rs368934269 Chr.X: 54751127-54751127 on Build GRCh38
rs139183502 Chr.X: 54751117-54751117 on Build GRCh38
rs924025929 Chr.X: 54751065-54751065 on Build GRCh38
rs760299702 Chr.X: 54751135-54751135 on Build GRCh38
rs769134048 Chr.X: 54751015-54751015 on Build GRCh38
rs1035232632 Chr.X: 54750797-54750797 on Build GRCh38
rs780767581 Chr.X: 54751076-54751076 on Build GRCh38
rs1028406690 Chr.X: 54750886-54750886 on Build GRCh38
rs750634235 Chr.X: 54750744-54750744 on Build GRCh38
rs749027057 Chr.X: 54751009-54751009 on Build GRCh38
rs772942032 Chr.X: 54750977-54750977 on Build GRCh38
rs535285789 Chr.X: 54751101-54751101 on Build GRCh38
rs752161857 Chr.X: 54751178-54751178 on Build GRCh38
rs752439019 Chr.X: 54751056-54751056 on Build GRCh38
rs780141307 Chr.X: 54750879-54750879 on Build GRCh38
rs1009266264 Chr.X: 54750811-54750811 on Build GRCh38
rs763049826 Chr.X: 54751048-54751048 on Build GRCh38

View Full Product Details