Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71110046-71110476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RG
Entrez Gene ID
3561
Gene Name
interleukin 2 receptor subunit gamma
Gene Aliases
CD132, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1
UniGene
Hs.84
Gene Chromosome Location
Chr.X: 71107404-71111631 on Build GRCh38
Transcripts
NM_000206.2

SNP Detail

SNP ID Chromosome Location
rs150560580 Chr.X: 71110291-71110291 on Build GRCh38
rs979698157 Chr.X: 71110316-71110316 on Build GRCh38
rs754007832 Chr.X: 71110215-71110215 on Build GRCh38
rs1009255987 Chr.X: 71110259-71110259 on Build GRCh38
rs773342053 Chr.X: 71110183-71110183 on Build GRCh38
rs760755004 Chr.X: 71110182-71110182 on Build GRCh38
rs767280606 Chr.X: 71110153-71110153 on Build GRCh38
rs757827108 Chr.X: 71110299-71110299 on Build GRCh38
rs768114423 Chr.X: 71110312-71110312 on Build GRCh38
rs1034305744 Chr.X: 71110358-71110358 on Build GRCh38
rs775037876 Chr.X: 71110125-71110125 on Build GRCh38
rs763359860 Chr.X: 71110239-71110239 on Build GRCh38
rs193922348 Chr.X: 71110295-71110295 on Build GRCh38
rs747781307 Chr.X: 71110317-71110317 on Build GRCh38
rs748820536 Chr.X: 71110305-71110305 on Build GRCh38
rs199593676 Chr.X: 71110252-71110252 on Build GRCh38
rs762420284 Chr.X: 71110126-71110126 on Build GRCh38
rs201665163 Chr.X: 71110119-71110119 on Build GRCh38
rs141707292 Chr.X: 71110236-71110236 on Build GRCh38
rs761545792 Chr.X: 71110141-71110141 on Build GRCh38
rs766505134 Chr.X: 71110337-71110337 on Build GRCh38
rs758693125 Chr.X: 71110256-71110256 on Build GRCh38
rs1005449113 Chr.X: 71110139-71110139 on Build GRCh38
rs751943366 Chr.X: 71110421-71110421 on Build GRCh38
rs769306246 Chr.X: 71110121-71110121 on Build GRCh38
rs959572366 Chr.X: 71110414-71110414 on Build GRCh38
rs752921124 Chr.X: 71110246-71110246 on Build GRCh38
rs111033621 Chr.X: 71110292-71110292 on Build GRCh38
rs762043391 Chr.X: 71110081-71110084 on Build GRCh38
rs773880797 Chr.X: 71110314-71110314 on Build GRCh38
rs781649645 Chr.X: 71110303-71110303 on Build GRCh38
rs200797436 Chr.X: 71110115-71110115 on Build GRCh38
rs778140826 Chr.X: 71110257-71110257 on Build GRCh38
rs768349207 Chr.X: 71110140-71110140 on Build GRCh38
rs750345322 Chr.X: 71110175-71110175 on Build GRCh38
rs992703553 Chr.X: 71110415-71110415 on Build GRCh38
rs1041213698 Chr.X: 71110216-71110216 on Build GRCh38
rs1020615876 Chr.X: 71110263-71110263 on Build GRCh38
rs878923825 Chr.X: 71110193-71110193 on Build GRCh38

View Full Product Details