Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15980272-15980843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROM1
Entrez Gene ID
8842
Gene Name
prominin 1
Gene Aliases
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
UniGene
Hs.614734
Gene Chromosome Location
Chr.4: 15968226-16084100 on Build GRCh38
Transcripts
NM_001145847.1 NM_001145848.1 NM_001145849.1 NM_001145850.1 NM_001145851.1 NM_001145852.1 NM_006017.2 XM_005248195.4 XM_005248196.4 XM_006713974.3 XM_011513890.1 XM_011513892.2 XM_011513893.2 XM_011513894.2 XM_011513895.2 XM_011513896.2 XM_011513897.2 XM_011513898.2 XM_011513899.2 XM_011513900.2 XM_011513902.2 XM_011513903.2 XM_017008799.1 XM_017008800.1 XM_017008801.1 XM_017008802.1 XM_017008803.1 XM_017008804.1 XM_017008805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6893146 c.2435G>A p.R812Q Chr.4: 15980449-15980449 on Build GRCh38
3702550 c.2409G>A p.A803A Chr.4: 15980475-15980475 on Build GRCh38
3714827 c.2461G>C p.D821H Chr.4: 15980423-15980423 on Build GRCh38
5941241 c.2368G>A p.G790R Chr.4: 15980516-15980516 on Build GRCh38
6860817 c.2463-3_2463-2insT p.? Chr.4: 15980303-15980304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34269395 Chr.4: 15980540-15980541 on Build GRCh38
rs773542265 Chr.4: 15980550-15980550 on Build GRCh38
rs774262417 Chr.4: 15980433-15980433 on Build GRCh38
rs368346472 Chr.4: 15980607-15980613 on Build GRCh38
rs975534422 Chr.4: 15980787-15980787 on Build GRCh38
rs780541802 Chr.4: 15980321-15980321 on Build GRCh38
rs73230220 Chr.4: 15980759-15980759 on Build GRCh38
rs372945014 Chr.4: 15980714-15980714 on Build GRCh38
rs755393305 Chr.4: 15980326-15980326 on Build GRCh38
rs369355817 Chr.4: 15980321-15980321 on Build GRCh38
rs200645062 Chr.4: 15980607-15980612 on Build GRCh38
rs375216918 Chr.4: 15980686-15980686 on Build GRCh38
rs779244085 Chr.4: 15980330-15980330 on Build GRCh38
rs567214704 Chr.4: 15980624-15980630 on Build GRCh38
rs746667815 Chr.4: 15980303-15980304 on Build GRCh38
rs368515078 Chr.4: 15980465-15980465 on Build GRCh38
rs796468740 Chr.4: 15980320-15980320 on Build GRCh38
rs752012414 Chr.4: 15980403-15980403 on Build GRCh38
rs1013388720 Chr.4: 15980611-15980611 on Build GRCh38
rs370736469 Chr.4: 15980449-15980449 on Build GRCh38
rs369431929 Chr.4: 15980700-15980700 on Build GRCh38
rs1014692401 Chr.4: 15980689-15980689 on Build GRCh38
rs779675775 Chr.4: 15980489-15980489 on Build GRCh38
rs6449209 Chr.4: 15980543-15980543 on Build GRCh38
rs376574674 Chr.4: 15980320-15980321 on Build GRCh38
rs975461967 Chr.4: 15980784-15980784 on Build GRCh38
rs1014666024 Chr.4: 15980342-15980342 on Build GRCh38
rs370477050 Chr.4: 15980542-15980543 on Build GRCh38
rs376134203 Chr.4: 15980524-15980524 on Build GRCh38
rs766357803 Chr.4: 15980450-15980450 on Build GRCh38
rs768449418 Chr.4: 15980304-15980305 on Build GRCh38
rs912649689 Chr.4: 15980769-15980774 on Build GRCh38
rs776374839 Chr.4: 15980609-15980614 on Build GRCh38
rs894500089 Chr.4: 15980595-15980595 on Build GRCh38
rs768616945 Chr.4: 15980427-15980427 on Build GRCh38
rs760021491 Chr.4: 15980319-15980320 on Build GRCh38
rs371838461 Chr.4: 15980576-15980576 on Build GRCh38
rs941353631 Chr.4: 15980582-15980582 on Build GRCh38
rs879196518 Chr.4: 15980607-15980607 on Build GRCh38
rs370967396 Chr.4: 15980606-15980609 on Build GRCh38
rs372746499 Chr.4: 15980439-15980439 on Build GRCh38
rs760761831 Chr.4: 15980325-15980326 on Build GRCh38
rs377213813 Chr.4: 15980441-15980441 on Build GRCh38
rs754214373 Chr.4: 15980325-15980325 on Build GRCh38
rs200008077 Chr.4: 15980518-15980518 on Build GRCh38
rs201460480 Chr.4: 15980613-15980613 on Build GRCh38
rs757702736 Chr.4: 15980503-15980503 on Build GRCh38
rs886059198 Chr.4: 15980431-15980431 on Build GRCh38
rs377669778 Chr.4: 15980600-15980601 on Build GRCh38
rs374854589 Chr.4: 15980715-15980715 on Build GRCh38
rs58036643 Chr.4: 15980304-15980304 on Build GRCh38
rs561412660 Chr.4: 15980606-15980607 on Build GRCh38
rs768356107 Chr.4: 15980304-15980306 on Build GRCh38
rs200874380 Chr.4: 15980379-15980379 on Build GRCh38
rs766089651 Chr.4: 15980607-15980614 on Build GRCh38
rs1032654718 Chr.4: 15980733-15980733 on Build GRCh38
rs146434364 Chr.4: 15980426-15980426 on Build GRCh38
rs771462940 Chr.4: 15980316-15980321 on Build GRCh38
rs771942110 Chr.4: 15980606-15980612 on Build GRCh38
rs887073099 Chr.4: 15980362-15980362 on Build GRCh38
rs746933638 Chr.4: 15980608-15980614 on Build GRCh38
rs956636567 Chr.4: 15980731-15980731 on Build GRCh38
rs942448504 Chr.4: 15980531-15980531 on Build GRCh38
rs922733416 Chr.4: 15980545-15980545 on Build GRCh38
rs771546267 Chr.4: 15980606-15980611 on Build GRCh38
rs906321716 Chr.4: 15980681-15980681 on Build GRCh38
rs931618095 Chr.4: 15980514-15980514 on Build GRCh38
rs371444761 Chr.4: 15980476-15980476 on Build GRCh38
rs989127104 Chr.4: 15980753-15980753 on Build GRCh38
rs979411892 Chr.4: 15980728-15980728 on Build GRCh38
rs570545320 Chr.4: 15980774-15980774 on Build GRCh38
rs770569204 Chr.4: 15980346-15980346 on Build GRCh38
rs746713193 Chr.4: 15980337-15980337 on Build GRCh38
rs768670104 Chr.4: 15980497-15980497 on Build GRCh38
rs751856109 Chr.4: 15980567-15980567 on Build GRCh38
rs199871818 Chr.4: 15980614-15980614 on Build GRCh38
rs755446696 Chr.4: 15980453-15980453 on Build GRCh38
rs745319992 Chr.4: 15980380-15980380 on Build GRCh38
rs912618452 Chr.4: 15980455-15980455 on Build GRCh38
rs1005509145 Chr.4: 15980376-15980376 on Build GRCh38
rs115191060 Chr.4: 15980553-15980553 on Build GRCh38
rs550431575 Chr.4: 15980623-15980630 on Build GRCh38
rs745830890 Chr.4: 15980295-15980296 on Build GRCh38
rs77314058 Chr.4: 15980639-15980639 on Build GRCh38
rs147880953 Chr.4: 15980457-15980457 on Build GRCh38
rs969451501 Chr.4: 15980718-15980718 on Build GRCh38
rs1024404197 Chr.4: 15980612-15980612 on Build GRCh38
rs766577396 Chr.4: 15980475-15980475 on Build GRCh38
rs1047320414 Chr.4: 15980359-15980359 on Build GRCh38
rs7668131 Chr.4: 15980607-15980607 on Build GRCh38
rs764185270 Chr.4: 15980397-15980397 on Build GRCh38
rs371997460 Chr.4: 15980819-15980819 on Build GRCh38
rs549596143 Chr.4: 15980305-15980305 on Build GRCh38
rs754410636 Chr.4: 15980452-15980452 on Build GRCh38
rs542424952 Chr.4: 15980391-15980391 on Build GRCh38
rs774901208 Chr.4: 15980317-15980321 on Build GRCh38
rs775630400 Chr.4: 15980415-15980415 on Build GRCh38
rs866191488 Chr.4: 15980432-15980432 on Build GRCh38
rs551323235 Chr.4: 15980584-15980584 on Build GRCh38
rs373738555 Chr.4: 15980788-15980788 on Build GRCh38
rs575302651 Chr.4: 15980440-15980440 on Build GRCh38
rs370154531 Chr.4: 15980402-15980402 on Build GRCh38
rs398124217 Chr.4: 15980543-15980543 on Build GRCh38
rs1013258756 Chr.4: 15980702-15980702 on Build GRCh38
rs564113318 Chr.4: 15980491-15980491 on Build GRCh38
rs982406072 Chr.4: 15980447-15980447 on Build GRCh38
rs780837948 Chr.4: 15980466-15980466 on Build GRCh38

View Full Product Details