Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119498595-119499087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC645513
Entrez Gene ID
645513
Gene Name
septin 7 pseudogene
Gene Aliases
-
UniGene
Hs.677546
Gene Chromosome Location
Chr.4: 119454783-119499592 on Build GRCh38
Transcripts
NR_037630.1 NM_001083.3 NM_033430.2 NM_033437.3

Gene Symbol
PDE5A
Entrez Gene ID
8654
Gene Name
phosphodiesterase 5A
Gene Aliases
CGB-PDE, CN5A, PDE5
UniGene
Hs.647971
Gene Chromosome Location
Chr.4: 119494395-119629080 on Build GRCh38
Transcripts
NR_037630.1 NM_001083.3 NM_033430.2 NM_033437.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3599562 c.2473G>A p.G825R Chr.4: 119498630-119498630 on Build GRCh38
7383366 c.2599G>C p.G867R Chr.4: 119498630-119498630 on Build GRCh38
6166150 c.2484C>A p.G828G Chr.4: 119498619-119498619 on Build GRCh38
339816 c.2509G>A p.E837K Chr.4: 119498720-119498720 on Build GRCh38
6166149 c.2610C>A p.G870G Chr.4: 119498619-119498619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899991279 Chr.4: 119498836-119498836 on Build GRCh38
rs775695855 Chr.4: 119498736-119498736 on Build GRCh38
rs199872368 Chr.4: 119498774-119498774 on Build GRCh38
rs991583571 Chr.4: 119498989-119498989 on Build GRCh38
rs566897432 Chr.4: 119498777-119498777 on Build GRCh38
rs570838498 Chr.4: 119498628-119498628 on Build GRCh38
rs201183225 Chr.4: 119498742-119498742 on Build GRCh38
rs143408453 Chr.4: 119498622-119498622 on Build GRCh38
rs757023722 Chr.4: 119498717-119498717 on Build GRCh38
rs752263592 Chr.4: 119498697-119498697 on Build GRCh38
rs969173857 Chr.4: 119499005-119499005 on Build GRCh38
rs1029835996 Chr.4: 119498921-119498921 on Build GRCh38
rs893521490 Chr.4: 119498743-119498743 on Build GRCh38
rs765488746 Chr.4: 119498778-119498778 on Build GRCh38
rs906449549 Chr.4: 119498716-119498716 on Build GRCh38
rs769151564 Chr.4: 119498759-119498759 on Build GRCh38
rs762201093 Chr.4: 119498776-119498776 on Build GRCh38
rs201689484 Chr.4: 119498620-119498620 on Build GRCh38
rs968528925 Chr.4: 119498825-119498825 on Build GRCh38
rs201716853 Chr.4: 119498648-119498648 on Build GRCh38
rs761541983 Chr.4: 119498646-119498648 on Build GRCh38
rs779232858 Chr.4: 119498726-119498726 on Build GRCh38
rs141662936 Chr.4: 119498824-119498825 on Build GRCh38
rs746052731 Chr.4: 119498727-119498727 on Build GRCh38
rs202223380 Chr.4: 119498633-119498633 on Build GRCh38
rs373215155 Chr.4: 119498721-119498721 on Build GRCh38
rs766493005 Chr.4: 119498787-119498787 on Build GRCh38
rs367914889 Chr.4: 119498711-119498711 on Build GRCh38
rs749053188 Chr.4: 119498707-119498707 on Build GRCh38
rs567535136 Chr.4: 119499043-119499044 on Build GRCh38
rs916405645 Chr.4: 119498649-119498649 on Build GRCh38
rs747067152 Chr.4: 119498746-119498746 on Build GRCh38
rs527806558 Chr.4: 119498936-119498936 on Build GRCh38
rs910519401 Chr.4: 119498960-119498960 on Build GRCh38
rs374933707 Chr.4: 119498991-119498991 on Build GRCh38
rs977370140 Chr.4: 119498898-119498898 on Build GRCh38
rs764742810 Chr.4: 119499038-119499038 on Build GRCh38
rs772469633 Chr.4: 119498733-119498733 on Build GRCh38
rs761239684 Chr.4: 119498631-119498631 on Build GRCh38
rs76051661 Chr.4: 119498992-119498992 on Build GRCh38
rs200504615 Chr.4: 119498783-119498783 on Build GRCh38
rs144055854 Chr.4: 119498698-119498698 on Build GRCh38
rs140289122 Chr.4: 119498651-119498651 on Build GRCh38
rs751248499 Chr.4: 119498642-119498642 on Build GRCh38
rs774728491 Chr.4: 119498619-119498619 on Build GRCh38
rs111615296 Chr.4: 119498994-119498994 on Build GRCh38
rs998347904 Chr.4: 119498905-119498905 on Build GRCh38
rs61729962 Chr.4: 119498621-119498621 on Build GRCh38
rs943489097 Chr.4: 119498975-119498975 on Build GRCh38
rs761282823 Chr.4: 119499018-119499018 on Build GRCh38
rs963063839 Chr.4: 119498976-119498976 on Build GRCh38
rs753921669 Chr.4: 119498798-119498798 on Build GRCh38
rs1052118351 Chr.4: 119498738-119498738 on Build GRCh38
rs776990088 Chr.4: 119498638-119498638 on Build GRCh38
rs754972736 Chr.4: 119498941-119498941 on Build GRCh38
rs200438658 Chr.4: 119498639-119498639 on Build GRCh38
rs1016918418 Chr.4: 119498956-119498956 on Build GRCh38
rs777574814 Chr.4: 119498706-119498706 on Build GRCh38
rs761154403 Chr.4: 119498767-119498767 on Build GRCh38
rs752536773 Chr.4: 119498740-119498741 on Build GRCh38
rs199886141 Chr.4: 119498745-119498745 on Build GRCh38
rs138544693 Chr.4: 119498673-119498673 on Build GRCh38

View Full Product Details