Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173316058-173316606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STC2
Entrez Gene ID
8614
Gene Name
stanniocalcin 2
Gene Aliases
STC-2, STCRP
UniGene
Hs.233160
Gene Chromosome Location
Chr.5: 173314723-173329503 on Build GRCh38
Transcripts
NM_003714.2

SNP Detail

SNP ID Chromosome Location
rs867411075 Chr.5: 173316285-173316285 on Build GRCh38
rs375180856 Chr.5: 173316218-173316218 on Build GRCh38
rs986583244 Chr.5: 173316278-173316278 on Build GRCh38
rs964559970 Chr.5: 173316098-173316098 on Build GRCh38
rs954681985 Chr.5: 173316362-173316363 on Build GRCh38
rs752513616 Chr.5: 173316424-173316424 on Build GRCh38
rs749603364 Chr.5: 173316297-173316297 on Build GRCh38
rs993494103 Chr.5: 173316400-173316400 on Build GRCh38
rs758578577 Chr.5: 173316100-173316100 on Build GRCh38
rs540247451 Chr.5: 173316110-173316110 on Build GRCh38
rs925408569 Chr.5: 173316455-173316455 on Build GRCh38
rs987555379 Chr.5: 173316456-173316456 on Build GRCh38
rs942539827 Chr.5: 173316287-173316287 on Build GRCh38
rs936327563 Chr.5: 173316293-173316293 on Build GRCh38
rs975671954 Chr.5: 173316542-173316542 on Build GRCh38
rs1002496407 Chr.5: 173316146-173316146 on Build GRCh38
rs866936308 Chr.5: 173316087-173316087 on Build GRCh38
rs370360827 Chr.5: 173316292-173316292 on Build GRCh38
rs1055307108 Chr.5: 173316509-173316509 on Build GRCh38
rs974604162 Chr.5: 173316353-173316354 on Build GRCh38
rs969623516 Chr.5: 173316145-173316145 on Build GRCh38
rs988721338 Chr.5: 173316387-173316387 on Build GRCh38
rs139938653 Chr.5: 173316388-173316388 on Build GRCh38
rs1013377799 Chr.5: 173316299-173316299 on Build GRCh38
rs558362571 Chr.5: 173316562-173316562 on Build GRCh38
rs74992533 Chr.5: 173316502-173316502 on Build GRCh38
rs913894507 Chr.5: 173316230-173316230 on Build GRCh38
rs1016928610 Chr.5: 173316198-173316198 on Build GRCh38
rs1017651412 Chr.5: 173316489-173316489 on Build GRCh38
rs527684983 Chr.5: 173316328-173316328 on Build GRCh38
rs569110291 Chr.5: 173316124-173316124 on Build GRCh38
rs945480502 Chr.5: 173316255-173316255 on Build GRCh38
rs750609417 Chr.5: 173316082-173316082 on Build GRCh38
rs573716581 Chr.5: 173316222-173316222 on Build GRCh38
rs564815291 Chr.5: 173316340-173316340 on Build GRCh38
rs578067513 Chr.5: 173316097-173316097 on Build GRCh38
rs896822713 Chr.5: 173316298-173316298 on Build GRCh38
rs905320036 Chr.5: 173316342-173316342 on Build GRCh38
rs1135097 Chr.5: 173316557-173316557 on Build GRCh38
rs150673127 Chr.5: 173316273-173316273 on Build GRCh38
rs569620175 Chr.5: 173316543-173316543 on Build GRCh38
rs191266094 Chr.5: 173316362-173316362 on Build GRCh38
rs369676744 Chr.5: 173316259-173316259 on Build GRCh38
rs777524951 Chr.5: 173316390-173316390 on Build GRCh38
rs531392067 Chr.5: 173316294-173316294 on Build GRCh38
rs919325629 Chr.5: 173316256-173316256 on Build GRCh38
rs142905526 Chr.5: 173316132-173316132 on Build GRCh38
rs972486152 Chr.5: 173316223-173316223 on Build GRCh38
rs529726142 Chr.5: 173316415-173316415 on Build GRCh38
rs923228257 Chr.5: 173316143-173316143 on Build GRCh38
rs979398220 Chr.5: 173316407-173316407 on Build GRCh38
rs757462061 Chr.5: 173316092-173316092 on Build GRCh38
rs750524683 Chr.5: 173316134-173316134 on Build GRCh38
rs938205267 Chr.5: 173316279-173316279 on Build GRCh38
rs538030509 Chr.5: 173316552-173316552 on Build GRCh38
rs933192556 Chr.5: 173316170-173316170 on Build GRCh38
rs771211001 Chr.5: 173316350-173316357 on Build GRCh38
rs1002790283 Chr.5: 173316344-173316344 on Build GRCh38
rs770550351 Chr.5: 173316550-173316550 on Build GRCh38
rs1039590063 Chr.5: 173316288-173316288 on Build GRCh38
rs567226887 Chr.5: 173316403-173316403 on Build GRCh38

View Full Product Details