Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99493697-99494239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF394
Entrez Gene ID
84124
Gene Name
zinc finger protein 394
Gene Aliases
ZKSCAN14, ZSCAN46
UniGene
Hs.386324
Gene Chromosome Location
Chr.7: 99486519-99500302 on Build GRCh38
Transcripts
NM_032164.2 XM_017012710.1 XM_017012711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286365 c.1422C>T p.C474C Chr.7: 99493793-99493793 on Build GRCh38
2867218 c.1292G>A p.R431H Chr.7: 99493923-99493923 on Build GRCh38
7286463 c.1047G>T p.Q349H Chr.7: 99494168-99494168 on Build GRCh38
5960883 c.1251G>C p.L417L Chr.7: 99493964-99493964 on Build GRCh38
3982305 c.1239G>A p.P413P Chr.7: 99493976-99493976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771554441 Chr.7: 99494193-99494193 on Build GRCh38
rs761934979 Chr.7: 99493788-99493788 on Build GRCh38
rs776005371 Chr.7: 99494147-99494147 on Build GRCh38
rs369951167 Chr.7: 99493793-99493793 on Build GRCh38
rs777024156 Chr.7: 99493785-99493785 on Build GRCh38
rs1019252335 Chr.7: 99493866-99493866 on Build GRCh38
rs763504677 Chr.7: 99493794-99493794 on Build GRCh38
rs911125493 Chr.7: 99494219-99494219 on Build GRCh38
rs751929204 Chr.7: 99493733-99493733 on Build GRCh38
rs781093559 Chr.7: 99494099-99494099 on Build GRCh38
rs753454166 Chr.7: 99493997-99493997 on Build GRCh38
rs990258391 Chr.7: 99494072-99494072 on Build GRCh38
rs761987977 Chr.7: 99493718-99493718 on Build GRCh38
rs749886513 Chr.7: 99494210-99494210 on Build GRCh38
rs751885306 Chr.7: 99493799-99493799 on Build GRCh38
rs754566877 Chr.7: 99493736-99493736 on Build GRCh38
rs61737538 Chr.7: 99493796-99493796 on Build GRCh38
rs78336331 Chr.7: 99493976-99493976 on Build GRCh38
rs142908024 Chr.7: 99493985-99493985 on Build GRCh38
rs755367126 Chr.7: 99493802-99493802 on Build GRCh38
rs923612428 Chr.7: 99494186-99494186 on Build GRCh38
rs549168586 Chr.7: 99493994-99493994 on Build GRCh38
rs145840389 Chr.7: 99493970-99493970 on Build GRCh38
rs762584008 Chr.7: 99494216-99494216 on Build GRCh38
rs772894359 Chr.7: 99493961-99493961 on Build GRCh38
rs768798762 Chr.7: 99494045-99494049 on Build GRCh38
rs772496774 Chr.7: 99494141-99494141 on Build GRCh38
rs780828334 Chr.7: 99493741-99493741 on Build GRCh38
rs763665378 Chr.7: 99493885-99493885 on Build GRCh38
rs1038338636 Chr.7: 99494008-99494008 on Build GRCh38
rs377441684 Chr.7: 99494067-99494067 on Build GRCh38
rs755775562 Chr.7: 99493884-99493884 on Build GRCh38
rs745778788 Chr.7: 99493910-99493910 on Build GRCh38
rs868740141 Chr.7: 99494166-99494166 on Build GRCh38
rs148228058 Chr.7: 99493850-99493850 on Build GRCh38
rs756981612 Chr.7: 99494001-99494001 on Build GRCh38
rs150696777 Chr.7: 99494107-99494107 on Build GRCh38
rs749506038 Chr.7: 99494022-99494040 on Build GRCh38
rs769320625 Chr.7: 99493754-99493754 on Build GRCh38
rs760089445 Chr.7: 99493870-99493870 on Build GRCh38
rs140280438 Chr.7: 99493951-99493951 on Build GRCh38
rs773287043 Chr.7: 99493849-99493849 on Build GRCh38
rs377630090 Chr.7: 99493874-99493874 on Build GRCh38
rs746814350 Chr.7: 99493924-99493924 on Build GRCh38
rs768929770 Chr.7: 99493784-99493784 on Build GRCh38
rs766870050 Chr.7: 99493863-99493863 on Build GRCh38
rs201390152 Chr.7: 99494106-99494106 on Build GRCh38
rs942487729 Chr.7: 99493905-99493905 on Build GRCh38
rs765039143 Chr.7: 99494200-99494200 on Build GRCh38
rs753318022 Chr.7: 99493893-99493893 on Build GRCh38
rs770839119 Chr.7: 99493833-99493833 on Build GRCh38
rs779009136 Chr.7: 99494054-99494056 on Build GRCh38
rs768073512 Chr.7: 99493975-99493975 on Build GRCh38
rs748259596 Chr.7: 99494058-99494066 on Build GRCh38
rs749221141 Chr.7: 99493758-99493758 on Build GRCh38
rs772014677 Chr.7: 99493844-99493844 on Build GRCh38
rs770836611 Chr.7: 99493773-99493773 on Build GRCh38
rs758447840 Chr.7: 99493916-99493916 on Build GRCh38
rs879129210 Chr.7: 99493777-99493777 on Build GRCh38
rs1019304065 Chr.7: 99494190-99494190 on Build GRCh38
rs750019483 Chr.7: 99494058-99494058 on Build GRCh38
rs899362645 Chr.7: 99494016-99494016 on Build GRCh38
rs761048588 Chr.7: 99494167-99494167 on Build GRCh38
rs770933535 Chr.7: 99494125-99494126 on Build GRCh38
rs774864258 Chr.7: 99493865-99493865 on Build GRCh38
rs758355027 Chr.7: 99494060-99494060 on Build GRCh38
rs760948373 Chr.7: 99493984-99493984 on Build GRCh38
rs114413880 Chr.7: 99493835-99493835 on Build GRCh38
rs749059749 Chr.7: 99493931-99493931 on Build GRCh38
rs574670480 Chr.7: 99493778-99493778 on Build GRCh38
rs868812712 Chr.7: 99494035-99494035 on Build GRCh38
rs780464083 Chr.7: 99493731-99493731 on Build GRCh38
rs769969218 Chr.7: 99493847-99493847 on Build GRCh38
rs779990254 Chr.7: 99493920-99493920 on Build GRCh38
rs753130219 Chr.7: 99493882-99493882 on Build GRCh38
rs374077648 Chr.7: 99493841-99493841 on Build GRCh38
rs760036321 Chr.7: 99493964-99493964 on Build GRCh38
rs780342924 Chr.7: 99493931-99493932 on Build GRCh38
rs773339323 Chr.7: 99493947-99493947 on Build GRCh38
rs775473049 Chr.7: 99493846-99493846 on Build GRCh38
rs768878441 Chr.7: 99494175-99494175 on Build GRCh38
rs764808339 Chr.7: 99494017-99494017 on Build GRCh38
rs777091972 Chr.7: 99494181-99494181 on Build GRCh38
rs758876923 Chr.7: 99493730-99493730 on Build GRCh38
rs755806041 Chr.7: 99493813-99493813 on Build GRCh38
rs754753153 Chr.7: 99494084-99494084 on Build GRCh38
rs866604275 Chr.7: 99494077-99494077 on Build GRCh38
rs777335435 Chr.7: 99493820-99493820 on Build GRCh38
rs765336766 Chr.7: 99493792-99493792 on Build GRCh38
rs748804580 Chr.7: 99493827-99493827 on Build GRCh38
rs376637716 Chr.7: 99493774-99493774 on Build GRCh38
rs201864076 Chr.7: 99493746-99493746 on Build GRCh38
rs145046863 Chr.7: 99493751-99493751 on Build GRCh38
rs1034532141 Chr.7: 99493989-99493989 on Build GRCh38
rs778860202 Chr.7: 99493896-99493896 on Build GRCh38
rs977831559 Chr.7: 99494174-99494174 on Build GRCh38
rs780777534 Chr.7: 99493809-99493809 on Build GRCh38
rs749443143 Chr.7: 99494102-99494102 on Build GRCh38
rs756745586 Chr.7: 99493894-99493894 on Build GRCh38
rs750960609 Chr.7: 99493726-99493726 on Build GRCh38
rs575999191 Chr.7: 99493873-99493873 on Build GRCh38
rs376629185 Chr.7: 99493945-99493945 on Build GRCh38
rs75476549 Chr.7: 99494028-99494028 on Build GRCh38
rs938728584 Chr.7: 99493936-99493936 on Build GRCh38
rs933642901 Chr.7: 99494191-99494191 on Build GRCh38
rs770077558 Chr.7: 99494132-99494134 on Build GRCh38
rs371418453 Chr.7: 99493725-99493725 on Build GRCh38
rs746781878 Chr.7: 99494083-99494083 on Build GRCh38
rs749341211 Chr.7: 99493950-99493950 on Build GRCh38

View Full Product Details